Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 179836357-179836848 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SQSTM1
Entrez Gene ID
8878
Gene Name
sequestosome 1
Gene Aliases
A170, FTDALS3, OSIL, PDB3, ZIP3, p60, p62, p62B
UniGene
Hs.587290
Gene Chromosome Location
Chr.5: 179806388-179838078 on Build GRCh38
Transcripts
NM_001142298.1 NM_001142299.1 NM_003900.4 XM_017010010.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6790011 c.1305G>A p.K435K Chr.5: 179836575-179836575 on Build GRCh38
3776577 c.1282C>T p.L428L Chr.5: 179836552-179836552 on Build GRCh38
1567750 c.1277C>T p.A426V Chr.5: 179836547-179836547 on Build GRCh38
7221231 c.1245G>A p.R415R Chr.5: 179836515-179836515 on Build GRCh38
4988730 c.1175C>T p.P392L Chr.5: 179836445-179836445 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1030474241 Chr.5: 179836759-179836759 on Build GRCh38
rs753600779 Chr.5: 179836590-179836590 on Build GRCh38
rs775798698 Chr.5: 179836406-179836408 on Build GRCh38
rs368933511 Chr.5: 179836485-179836485 on Build GRCh38
rs762322293 Chr.5: 179836430-179836430 on Build GRCh38
rs75700262 Chr.5: 179836446-179836446 on Build GRCh38
rs765613974 Chr.5: 179836431-179836431 on Build GRCh38
rs539942101 Chr.5: 179836447-179836447 on Build GRCh38
rs143511494 Chr.5: 179836501-179836501 on Build GRCh38
rs1014179726 Chr.5: 179836761-179836762 on Build GRCh38
rs1006467602 Chr.5: 179836696-179836696 on Build GRCh38
rs373148815 Chr.5: 179836631-179836631 on Build GRCh38
rs979372280 Chr.5: 179836783-179836783 on Build GRCh38
rs200551825 Chr.5: 179836448-179836448 on Build GRCh38
rs753608025 Chr.5: 179836650-179836650 on Build GRCh38
rs779531319 Chr.5: 179836610-179836610 on Build GRCh38
rs781300399 Chr.5: 179836706-179836707 on Build GRCh38
rs757311547 Chr.5: 179836676-179836676 on Build GRCh38
rs550096373 Chr.5: 179836382-179836382 on Build GRCh38
rs756095586 Chr.5: 179836455-179836455 on Build GRCh38
rs777601802 Chr.5: 179836540-179836540 on Build GRCh38
rs758468329 Chr.5: 179836617-179836618 on Build GRCh38
rs922763648 Chr.5: 179836787-179836787 on Build GRCh38
rs538853972 Chr.5: 179836420-179836423 on Build GRCh38
rs11548622 Chr.5: 179836675-179836675 on Build GRCh38
rs989864881 Chr.5: 179836778-179836779 on Build GRCh38
rs186996560 Chr.5: 179836768-179836768 on Build GRCh38
rs749217241 Chr.5: 179836458-179836458 on Build GRCh38
rs201795943 Chr.5: 179836471-179836471 on Build GRCh38
rs375943902 Chr.5: 179836397-179836397 on Build GRCh38
rs745585269 Chr.5: 179836476-179836476 on Build GRCh38
rs577638223 Chr.5: 179836652-179836652 on Build GRCh38
rs987821927 Chr.5: 179836592-179836592 on Build GRCh38
rs10688915 Chr.5: 179836781-179836782 on Build GRCh38
rs765122407 Chr.5: 179836606-179836607 on Build GRCh38
rs1052389844 Chr.5: 179836408-179836408 on Build GRCh38
rs143977783 Chr.5: 179836548-179836548 on Build GRCh38
rs958008519 Chr.5: 179836770-179836770 on Build GRCh38
rs1015277726 Chr.5: 179836742-179836742 on Build GRCh38
rs771468767 Chr.5: 179836702-179836702 on Build GRCh38
rs771657338 Chr.5: 179836477-179836477 on Build GRCh38
rs778010441 Chr.5: 179836623-179836623 on Build GRCh38
rs757212984 Chr.5: 179836543-179836543 on Build GRCh38
rs770396685 Chr.5: 179836464-179836464 on Build GRCh38
rs750331363 Chr.5: 179836601-179836601 on Build GRCh38
rs370200075 Chr.5: 179836636-179836636 on Build GRCh38
rs747890873 Chr.5: 179836452-179836452 on Build GRCh38
rs766437927 Chr.5: 179836500-179836500 on Build GRCh38
rs1027965196 Chr.5: 179836405-179836405 on Build GRCh38
rs540123776 Chr.5: 179836401-179836401 on Build GRCh38
rs772788481 Chr.5: 179836404-179836404 on Build GRCh38
rs796259575 Chr.5: 179836738-179836738 on Build GRCh38
rs531862762 Chr.5: 179836381-179836381 on Build GRCh38
rs17409643 Chr.5: 179836807-179836807 on Build GRCh38
rs771743528 Chr.5: 179836549-179836549 on Build GRCh38
rs104893941 Chr.5: 179836445-179836445 on Build GRCh38
rs155790 Chr.5: 179836676-179836676 on Build GRCh38
rs796051869 Chr.5: 179836494-179836495 on Build GRCh38
rs777004439 Chr.5: 179836479-179836479 on Build GRCh38
rs755901084 Chr.5: 179836537-179836537 on Build GRCh38
rs534476029 Chr.5: 179836576-179836576 on Build GRCh38
rs777527845 Chr.5: 179836457-179836457 on Build GRCh38
rs199854262 Chr.5: 179836586-179836586 on Build GRCh38
rs760096900 Chr.5: 179836628-179836628 on Build GRCh38
rs748161241 Chr.5: 179836390-179836390 on Build GRCh38
rs770648641 Chr.5: 179836412-179836412 on Build GRCh38
rs17409649 Chr.5: 179836804-179836804 on Build GRCh38
rs369369096 Chr.5: 179836380-179836380 on Build GRCh38
rs777849713 Chr.5: 179836627-179836630 on Build GRCh38
rs771966860 Chr.5: 179836480-179836480 on Build GRCh38
rs759646319 Chr.5: 179836583-179836583 on Build GRCh38
rs747720686 Chr.5: 179836689-179836689 on Build GRCh38
rs376209849 Chr.5: 179836632-179836632 on Build GRCh38
rs1005652425 Chr.5: 179836737-179836737 on Build GRCh38
rs888099194 Chr.5: 179836705-179836705 on Build GRCh38
rs913918805 Chr.5: 179836780-179836781 on Build GRCh38
rs998103787 Chr.5: 179836753-179836753 on Build GRCh38
rs71714822 Chr.5: 179836777-179836778 on Build GRCh38
rs955301962 Chr.5: 179836761-179836761 on Build GRCh38
rs755261626 Chr.5: 179836504-179836504 on Build GRCh38
rs11249660 Chr.5: 179836785-179836785 on Build GRCh38
rs964933084 Chr.5: 179836749-179836749 on Build GRCh38
rs760432280 Chr.5: 179836625-179836625 on Build GRCh38
rs1041583907 Chr.5: 179836669-179836669 on Build GRCh38
rs762316663 Chr.5: 179836411-179836411 on Build GRCh38
rs945668579 Chr.5: 179836661-179836661 on Build GRCh38
rs770118706 Chr.5: 179836558-179836558 on Build GRCh38
rs757952273 Chr.5: 179836606-179836606 on Build GRCh38
rs773915429 Chr.5: 179836413-179836413 on Build GRCh38
rs140407570 Chr.5: 179836659-179836659 on Build GRCh38
rs148278350 Chr.5: 179836515-179836515 on Build GRCh38
rs926546897 Chr.5: 179836803-179836803 on Build GRCh38
rs781123506 Chr.5: 179836389-179836389 on Build GRCh38
rs201239306 Chr.5: 179836547-179836547 on Build GRCh38
rs752910741 Chr.5: 179836585-179836585 on Build GRCh38
rs763376504 Chr.5: 179836569-179836569 on Build GRCh38
rs765964997 Chr.5: 179836593-179836595 on Build GRCh38
rs751025643 Chr.5: 179836660-179836660 on Build GRCh38
rs763290503 Chr.5: 179836694-179836694 on Build GRCh38
rs529512338 Chr.5: 179836435-179836435 on Build GRCh38
rs746655349 Chr.5: 179836555-179836555 on Build GRCh38
rs757166561 Chr.5: 179836596-179836596 on Build GRCh38
rs752519655 Chr.5: 179836617-179836631 on Build GRCh38
rs751702490 Chr.5: 179836643-179836644 on Build GRCh38
rs11548636 Chr.5: 179836587-179836587 on Build GRCh38
rs987883372 Chr.5: 179836765-179836765 on Build GRCh38
rs374985304 Chr.5: 179836542-179836542 on Build GRCh38
rs775814512 Chr.5: 179836679-179836679 on Build GRCh38
rs753485135 Chr.5: 179836601-179836613 on Build GRCh38
rs138527258 Chr.5: 179836607-179836621 on Build GRCh38
rs759165538 Chr.5: 179836415-179836415 on Build GRCh38
rs772772504 Chr.5: 179836534-179836535 on Build GRCh38
rs967690981 Chr.5: 179836774-179836774 on Build GRCh38
rs750423230 Chr.5: 179836654-179836654 on Build GRCh38
rs763497372 Chr.5: 179836412-179836413 on Build GRCh38
rs764498902 Chr.5: 179836416-179836416 on Build GRCh38
rs774591916 Chr.5: 179836578-179836578 on Build GRCh38
rs773625766 Chr.5: 179836559-179836559 on Build GRCh38
rs369609665 Chr.5: 179836653-179836653 on Build GRCh38
rs1049449325 Chr.5: 179836817-179836817 on Build GRCh38
rs768290894 Chr.5: 179836617-179836617 on Build GRCh38
rs775033427 Chr.5: 179836627-179836627 on Build GRCh38
rs182058393 Chr.5: 179836584-179836584 on Build GRCh38
rs386418446 Chr.5: 179836768-179836769 on Build GRCh38
rs1017794004 Chr.5: 179836723-179836723 on Build GRCh38
rs369114803 Chr.5: 179836613-179836613 on Build GRCh38
rs937920044 Chr.5: 179836829-179836829 on Build GRCh38
rs758452325 Chr.5: 179836442-179836442 on Build GRCh38
rs933630561 Chr.5: 179836407-179836407 on Build GRCh38
rs778783549 Chr.5: 179836600-179836600 on Build GRCh38
rs754136945 Chr.5: 179836419-179836419 on Build GRCh38
rs763506589 Chr.5: 179836491-179836491 on Build GRCh38
rs201463748 Chr.5: 179836424-179836424 on Build GRCh38
rs778634431 Chr.5: 179836783-179836785 on Build GRCh38
rs573881131 Chr.5: 179836554-179836554 on Build GRCh38
rs773645716 Chr.5: 179836486-179836486 on Build GRCh38
rs760169726 Chr.5: 179836553-179836554 on Build GRCh38
rs371720013 Chr.5: 179836527-179836527 on Build GRCh38

View Full Product Details