Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 3258790-3259295 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSMG4
Entrez Gene ID
389362
Gene Name
proteasome assembly chaperone 4
Gene Aliases
C6orf86, PAC4, bA506K6.2
UniGene
Hs.207069
Gene Chromosome Location
Chr.6: 3254325-3308224 on Build GRCh38
Transcripts
NM_001128591.1 NM_001128592.1 NM_001135750.1 XM_011514591.2 XM_011514593.2 XM_011514594.2 XM_011514595.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6384021 c.23C>A p.A8D Chr.6: 3259045-3259045 on Build GRCh38
6385125 c.166A>G p.S56G Chr.6: 3259188-3259188 on Build GRCh38
7210610 c.84G>A p.W28* Chr.6: 3259143-3259143 on Build GRCh38
5423335 c.142C>T p.R48C Chr.6: 3259164-3259164 on Build GRCh38
5720529 c.6G>A p.E2E Chr.6: 3259028-3259028 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs957165322 Chr.6: 3258900-3258900 on Build GRCh38
rs966238276 Chr.6: 3259187-3259187 on Build GRCh38
rs9501962 Chr.6: 3259229-3259229 on Build GRCh38
rs752773059 Chr.6: 3258824-3258824 on Build GRCh38
rs897420276 Chr.6: 3258842-3258842 on Build GRCh38
rs905831483 Chr.6: 3258986-3258986 on Build GRCh38
rs768727783 Chr.6: 3259131-3259131 on Build GRCh38
rs547424149 Chr.6: 3258874-3258874 on Build GRCh38
rs916109075 Chr.6: 3259256-3259256 on Build GRCh38
rs761759612 Chr.6: 3259166-3259166 on Build GRCh38
rs527704382 Chr.6: 3258843-3258843 on Build GRCh38
rs759063079 Chr.6: 3259239-3259239 on Build GRCh38
rs746049177 Chr.6: 3259090-3259090 on Build GRCh38
rs1014956042 Chr.6: 3258978-3258978 on Build GRCh38
rs985860764 Chr.6: 3258869-3258870 on Build GRCh38
rs766109268 Chr.6: 3259210-3259210 on Build GRCh38
rs535627106 Chr.6: 3258976-3258976 on Build GRCh38
rs972186237 Chr.6: 3259250-3259250 on Build GRCh38
rs763515915 Chr.6: 3258995-3258995 on Build GRCh38
rs868665875 Chr.6: 3259141-3259141 on Build GRCh38
rs940465427 Chr.6: 3259099-3259099 on Build GRCh38
rs975759551 Chr.6: 3258912-3258912 on Build GRCh38
rs1045924845 Chr.6: 3259134-3259134 on Build GRCh38
rs899898819 Chr.6: 3259183-3259183 on Build GRCh38
rs976757943 Chr.6: 3258909-3258909 on Build GRCh38
rs779985403 Chr.6: 3259113-3259113 on Build GRCh38
rs762488081 Chr.6: 3259191-3259191 on Build GRCh38
rs953299385 Chr.6: 3259228-3259228 on Build GRCh38
rs527810564 Chr.6: 3259255-3259255 on Build GRCh38
rs1030930935 Chr.6: 3259020-3259020 on Build GRCh38
rs886187155 Chr.6: 3258822-3258822 on Build GRCh38
rs908913426 Chr.6: 3259231-3259231 on Build GRCh38
rs947831461 Chr.6: 3259116-3259116 on Build GRCh38
rs931747224 Chr.6: 3258820-3258820 on Build GRCh38
rs1054409221 Chr.6: 3259172-3259172 on Build GRCh38
rs989062872 Chr.6: 3258902-3258902 on Build GRCh38
rs1005731781 Chr.6: 3258832-3258832 on Build GRCh38
rs754414184 Chr.6: 3259070-3259070 on Build GRCh38
rs997372091 Chr.6: 3259193-3259193 on Build GRCh38
rs187112756 Chr.6: 3259040-3259040 on Build GRCh38
rs773096208 Chr.6: 3259182-3259182 on Build GRCh38
rs970110278 Chr.6: 3258873-3258873 on Build GRCh38
rs577429065 Chr.6: 3259055-3259055 on Build GRCh38
rs763712904 Chr.6: 3259116-3259117 on Build GRCh38
rs900229895 Chr.6: 3258814-3258814 on Build GRCh38
rs539791068 Chr.6: 3258899-3258899 on Build GRCh38
rs1001465794 Chr.6: 3259164-3259164 on Build GRCh38
rs941515884 Chr.6: 3258963-3258963 on Build GRCh38
rs747181103 Chr.6: 3259123-3259123 on Build GRCh38
rs749994023 Chr.6: 3259005-3259005 on Build GRCh38
rs1045730981 Chr.6: 3258985-3258985 on Build GRCh38
rs908975884 Chr.6: 3258952-3258952 on Build GRCh38
rs931639191 Chr.6: 3258939-3258939 on Build GRCh38
rs765773243 Chr.6: 3259046-3259046 on Build GRCh38
rs917681011 Chr.6: 3259251-3259251 on Build GRCh38
rs935030671 Chr.6: 3259148-3259148 on Build GRCh38
rs1024757356 Chr.6: 3258864-3258864 on Build GRCh38
rs989944980 Chr.6: 3258875-3258875 on Build GRCh38
rs947689407 Chr.6: 3258987-3258987 on Build GRCh38
rs570492431 Chr.6: 3258881-3258881 on Build GRCh38
rs999878787 Chr.6: 3259019-3259019 on Build GRCh38
rs950232465 Chr.6: 3259258-3259258 on Build GRCh38
rs191443505 Chr.6: 3259100-3259100 on Build GRCh38
rs1052745854 Chr.6: 3259152-3259152 on Build GRCh38
rs971552092 Chr.6: 3259264-3259264 on Build GRCh38
rs75909236 Chr.6: 3259073-3259073 on Build GRCh38
rs1011884472 Chr.6: 3258837-3258837 on Build GRCh38
rs1048810227 Chr.6: 3258948-3258948 on Build GRCh38
rs146321546 Chr.6: 3258907-3258907 on Build GRCh38
rs1010743932 Chr.6: 3259023-3259023 on Build GRCh38
rs1037632140 Chr.6: 3258965-3258965 on Build GRCh38
rs1020565524 Chr.6: 3259012-3259012 on Build GRCh38
rs970625717 Chr.6: 3258859-3258859 on Build GRCh38
rs114252161 Chr.6: 3259181-3259181 on Build GRCh38
rs964160513 Chr.6: 3259030-3259030 on Build GRCh38
rs951435378 Chr.6: 3259216-3259216 on Build GRCh38
rs1029762705 Chr.6: 3259190-3259190 on Build GRCh38
rs888578152 Chr.6: 3258825-3258825 on Build GRCh38
rs1033058670 Chr.6: 3258898-3258898 on Build GRCh38
rs751126684 Chr.6: 3259060-3259060 on Build GRCh38
rs1015819246 Chr.6: 3258838-3258838 on Build GRCh38
rs976365457 Chr.6: 3259028-3259028 on Build GRCh38
rs1028892015 Chr.6: 3259213-3259213 on Build GRCh38
rs916321712 Chr.6: 3259095-3259095 on Build GRCh38
rs976744920 Chr.6: 3259034-3259034 on Build GRCh38
rs750998014 Chr.6: 3259232-3259239 on Build GRCh38
rs570242105 Chr.6: 3258910-3258910 on Build GRCh38
rs957470074 Chr.6: 3259004-3259004 on Build GRCh38
rs944644135 Chr.6: 3258911-3258911 on Build GRCh38
rs998297817 Chr.6: 3259186-3259186 on Build GRCh38
rs779454348 Chr.6: 3259078-3259078 on Build GRCh38
rs201036341 Chr.6: 3259062-3259062 on Build GRCh38
rs941551803 Chr.6: 3258967-3258967 on Build GRCh38
rs1004219653 Chr.6: 3259222-3259229 on Build GRCh38
rs1006406614 Chr.6: 3258823-3258823 on Build GRCh38
rs999365429 Chr.6: 3258862-3258862 on Build GRCh38
rs910067891 Chr.6: 3258934-3258935 on Build GRCh38
rs572011294 Chr.6: 3259018-3259018 on Build GRCh38
rs759950024 Chr.6: 3259005-3259018 on Build GRCh38
rs866206808 Chr.6: 3259050-3259050 on Build GRCh38
rs893733061 Chr.6: 3259157-3259157 on Build GRCh38
rs184127704 Chr.6: 3259136-3259136 on Build GRCh38
rs982951574 Chr.6: 3258933-3258933 on Build GRCh38
rs921854513 Chr.6: 3259049-3259049 on Build GRCh38
rs984475530 Chr.6: 3259064-3259064 on Build GRCh38
rs893206420 Chr.6: 3259179-3259179 on Build GRCh38
rs921631908 Chr.6: 3258935-3258935 on Build GRCh38
rs906163000 Chr.6: 3259145-3259145 on Build GRCh38
rs563530189 Chr.6: 3259110-3259110 on Build GRCh38
rs555460327 Chr.6: 3258994-3258994 on Build GRCh38

View Full Product Details