Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 3286775-3287282 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSMG4
Entrez Gene ID
389362
Gene Name
proteasome assembly chaperone 4
Gene Aliases
C6orf86, PAC4, bA506K6.2
UniGene
Hs.207069
Gene Chromosome Location
Chr.6: 3254325-3308224 on Build GRCh38
Transcripts
XM_011514593.2 NM_001286455.1 NM_015482.1 NM_021945.6 NR_104448.1 XM_011514801.2 XM_017011180.1 XM_017011181.1 XM_017011182.1 XM_017011184.1 XM_017011186.1

Gene Symbol
SLC22A23
Entrez Gene ID
63027
Gene Name
solute carrier family 22 member 23
Gene Aliases
C6orf85
UniGene
Hs.713588
Gene Chromosome Location
Chr.6: 3268973-3456574 on Build GRCh38
Transcripts
XM_011514593.2 NM_001286455.1 NM_015482.1 NM_021945.6 NR_104448.1 XM_011514801.2 XM_017011180.1 XM_017011181.1 XM_017011182.1 XM_017011184.1 XM_017011186.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5405041 c.1472G>A p.G491E Chr.6: 3286933-3286933 on Build GRCh38
1226045 c.1436C>T p.S479F Chr.6: 3286969-3286969 on Build GRCh38
1287975 c.642G>A p.G214G Chr.6: 3286920-3286920 on Build GRCh38
6308027 c.1513C>A p.L505M Chr.6: 3286892-3286892 on Build GRCh38
4726708 c.667G>A p.A223T Chr.6: 3286895-3286895 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs536399766 Chr.6: 3286992-3286992 on Build GRCh38
rs761340954 Chr.6: 3286965-3286965 on Build GRCh38
rs756458794 Chr.6: 3286931-3286931 on Build GRCh38
rs4959796 Chr.6: 3287252-3287252 on Build GRCh38
rs889015746 Chr.6: 3287079-3287079 on Build GRCh38
rs964254654 Chr.6: 3287222-3287222 on Build GRCh38
rs760186741 Chr.6: 3286998-3286998 on Build GRCh38
rs763739645 Chr.6: 3286960-3286960 on Build GRCh38
rs764396067 Chr.6: 3287047-3287047 on Build GRCh38
rs748355311 Chr.6: 3286930-3286930 on Build GRCh38
rs147580105 Chr.6: 3286896-3286896 on Build GRCh38
rs749492480 Chr.6: 3286989-3286989 on Build GRCh38
rs777571864 Chr.6: 3287107-3287107 on Build GRCh38
rs772911297 Chr.6: 3287030-3287030 on Build GRCh38
rs929659195 Chr.6: 3287051-3287051 on Build GRCh38
rs973919504 Chr.6: 3287227-3287227 on Build GRCh38
rs765389281 Chr.6: 3287141-3287141 on Build GRCh38
rs184567316 Chr.6: 3287201-3287201 on Build GRCh38
rs138414914 Chr.6: 3287017-3287017 on Build GRCh38
rs142569200 Chr.6: 3286859-3286859 on Build GRCh38
rs767067685 Chr.6: 3287117-3287139 on Build GRCh38
rs550187337 Chr.6: 3286881-3286881 on Build GRCh38
rs781717357 Chr.6: 3286906-3286906 on Build GRCh38
rs776165428 Chr.6: 3286964-3286964 on Build GRCh38
rs1008224894 Chr.6: 3287208-3287208 on Build GRCh38
rs752031624 Chr.6: 3286895-3286895 on Build GRCh38
rs778910609 Chr.6: 3286994-3286994 on Build GRCh38
rs375953440 Chr.6: 3286848-3286848 on Build GRCh38
rs41301849 Chr.6: 3287164-3287164 on Build GRCh38
rs986575676 Chr.6: 3286807-3286807 on Build GRCh38
rs556499252 Chr.6: 3287146-3287146 on Build GRCh38
rs149321576 Chr.6: 3287025-3287025 on Build GRCh38
rs768069857 Chr.6: 3287032-3287032 on Build GRCh38
rs747820025 Chr.6: 3287098-3287098 on Build GRCh38
rs745847165 Chr.6: 3286949-3286949 on Build GRCh38
rs372168705 Chr.6: 3286816-3286816 on Build GRCh38
rs925691883 Chr.6: 3287238-3287238 on Build GRCh38
rs904094765 Chr.6: 3287142-3287142 on Build GRCh38
rs942590923 Chr.6: 3286823-3286823 on Build GRCh38
rs928386799 Chr.6: 3287096-3287096 on Build GRCh38
rs773003117 Chr.6: 3287102-3287102 on Build GRCh38
rs768469063 Chr.6: 3287094-3287094 on Build GRCh38
rs746982618 Chr.6: 3287086-3287086 on Build GRCh38
rs759265936 Chr.6: 3286860-3286860 on Build GRCh38
rs948244252 Chr.6: 3287132-3287132 on Build GRCh38
rs151086698 Chr.6: 3286858-3286858 on Build GRCh38
rs992388228 Chr.6: 3287186-3287186 on Build GRCh38
rs773389064 Chr.6: 3286853-3286853 on Build GRCh38
rs891216514 Chr.6: 3287195-3287195 on Build GRCh38
rs199733964 Chr.6: 3286974-3286974 on Build GRCh38
rs138000137 Chr.6: 3287040-3287040 on Build GRCh38
rs780896717 Chr.6: 3287097-3287097 on Build GRCh38
rs775414659 Chr.6: 3287008-3287008 on Build GRCh38
rs906938978 Chr.6: 3286894-3286894 on Build GRCh38
rs370232474 Chr.6: 3286935-3286935 on Build GRCh38
rs774294608 Chr.6: 3286947-3286947 on Build GRCh38
rs752318692 Chr.6: 3286878-3286878 on Build GRCh38
rs142078855 Chr.6: 3286950-3286950 on Build GRCh38
rs1030929769 Chr.6: 3287183-3287183 on Build GRCh38
rs770928245 Chr.6: 3286938-3286938 on Build GRCh38
rs750435931 Chr.6: 3287083-3287083 on Build GRCh38
rs150075690 Chr.6: 3286990-3286990 on Build GRCh38
rs757480302 Chr.6: 3287072-3287072 on Build GRCh38
rs764508070 Chr.6: 3287126-3287126 on Build GRCh38
rs140418737 Chr.6: 3287026-3287026 on Build GRCh38
rs370351292 Chr.6: 3286958-3286958 on Build GRCh38
rs201752643 Chr.6: 3287110-3287110 on Build GRCh38
rs776013893 Chr.6: 3286821-3286821 on Build GRCh38
rs778961106 Chr.6: 3287078-3287078 on Build GRCh38
rs760316368 Chr.6: 3286956-3286956 on Build GRCh38
rs745793578 Chr.6: 3287004-3287004 on Build GRCh38
rs761418308 Chr.6: 3286823-3286829 on Build GRCh38
rs895278319 Chr.6: 3287115-3287115 on Build GRCh38
rs777709967 Chr.6: 3286932-3286932 on Build GRCh38
rs774319638 Chr.6: 3287138-3287138 on Build GRCh38
rs766855461 Chr.6: 3286893-3286893 on Build GRCh38
rs373927826 Chr.6: 3286980-3286980 on Build GRCh38
rs754223734 Chr.6: 3287127-3287127 on Build GRCh38
rs1027339082 Chr.6: 3287235-3287235 on Build GRCh38
rs772064434 Chr.6: 3287006-3287006 on Build GRCh38
rs754070162 Chr.6: 3287069-3287069 on Build GRCh38
rs763094340 Chr.6: 3286952-3286952 on Build GRCh38
rs761050533 Chr.6: 3287046-3287046 on Build GRCh38
rs111449554 Chr.6: 3287133-3287133 on Build GRCh38
rs992733578 Chr.6: 3286995-3286995 on Build GRCh38
rs755587708 Chr.6: 3286977-3286977 on Build GRCh38
rs761137126 Chr.6: 3287123-3287123 on Build GRCh38
rs752078303 Chr.6: 3286973-3286973 on Build GRCh38
rs756727462 Chr.6: 3286888-3286888 on Build GRCh38
rs979584169 Chr.6: 3287229-3287229 on Build GRCh38
rs977102370 Chr.6: 3287039-3287039 on Build GRCh38
rs962437281 Chr.6: 3286942-3286942 on Build GRCh38
rs1021626570 Chr.6: 3287197-3287197 on Build GRCh38
rs767333215 Chr.6: 3286872-3286872 on Build GRCh38
rs748410717 Chr.6: 3286847-3286847 on Build GRCh38
rs371359993 Chr.6: 3287100-3287100 on Build GRCh38
rs573601720 Chr.6: 3287158-3287158 on Build GRCh38
rs887887960 Chr.6: 3287139-3287139 on Build GRCh38
rs766205435 Chr.6: 3286957-3286957 on Build GRCh38
rs1018311341 Chr.6: 3287209-3287209 on Build GRCh38
rs1012880731 Chr.6: 3286943-3286943 on Build GRCh38
rs760373448 Chr.6: 3286880-3286880 on Build GRCh38
rs200186759 Chr.6: 3286966-3286966 on Build GRCh38
rs374488596 Chr.6: 3287085-3287085 on Build GRCh38
rs6910086 Chr.6: 3286884-3286884 on Build GRCh38
rs951306255 Chr.6: 3287236-3287236 on Build GRCh38
rs764687839 Chr.6: 3286967-3286967 on Build GRCh38
rs767975985 Chr.6: 3286978-3286978 on Build GRCh38
rs577661046 Chr.6: 3287111-3287111 on Build GRCh38
rs761039686 Chr.6: 3286979-3286979 on Build GRCh38

View Full Product Details