Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 16143561-16144100 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYLIP
Entrez Gene ID
29116
Gene Name
myosin regulatory light chain interacting protein
Gene Aliases
IDOL, MIR
UniGene
Hs.484738
Gene Chromosome Location
Chr.6: 16129086-16151015 on Build GRCh38
Transcripts
NM_013262.3 XM_005249032.2 XM_005249033.2 XM_017010789.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4878589 c.663-1G>T p.? Chr.6: 16143698-16143698 on Build GRCh38
266983 c.819A>G p.A273A Chr.6: 16143855-16143855 on Build GRCh38
6547104 c.720G>T p.T240T Chr.6: 16143756-16143756 on Build GRCh38
7266276 c.813G>A p.T271T Chr.6: 16143849-16143849 on Build GRCh38
121263 c.705G>A p.K235K Chr.6: 16143741-16143741 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377214864 Chr.6: 16143687-16143687 on Build GRCh38
rs759282321 Chr.6: 16143947-16143947 on Build GRCh38
rs897843533 Chr.6: 16144028-16144028 on Build GRCh38
rs374072017 Chr.6: 16143783-16143783 on Build GRCh38
rs117833494 Chr.6: 16143619-16143619 on Build GRCh38
rs748156535 Chr.6: 16143859-16143859 on Build GRCh38
rs868222035 Chr.6: 16143865-16143865 on Build GRCh38
rs969126814 Chr.6: 16143925-16143925 on Build GRCh38
rs1020345790 Chr.6: 16143995-16143995 on Build GRCh38
rs954324042 Chr.6: 16143600-16143600 on Build GRCh38
rs112531050 Chr.6: 16143756-16143756 on Build GRCh38
rs533248503 Chr.6: 16144056-16144056 on Build GRCh38
rs142137048 Chr.6: 16144054-16144054 on Build GRCh38
rs201036784 Chr.6: 16143875-16143875 on Build GRCh38
rs773676911 Chr.6: 16143809-16143809 on Build GRCh38
rs2072783 Chr.6: 16143666-16143666 on Build GRCh38
rs199943196 Chr.6: 16143808-16143808 on Build GRCh38
rs777230333 Chr.6: 16143819-16143819 on Build GRCh38
rs779983827 Chr.6: 16143852-16143852 on Build GRCh38
rs185799393 Chr.6: 16143970-16143970 on Build GRCh38
rs1056972642 Chr.6: 16143636-16143636 on Build GRCh38
rs751026013 Chr.6: 16143836-16143836 on Build GRCh38
rs762101742 Chr.6: 16143699-16143699 on Build GRCh38
rs34627146 Chr.6: 16143849-16143849 on Build GRCh38
rs112824443 Chr.6: 16143757-16143757 on Build GRCh38
rs924550725 Chr.6: 16143934-16143934 on Build GRCh38
rs202197437 Chr.6: 16143779-16143779 on Build GRCh38
rs937910671 Chr.6: 16144016-16144016 on Build GRCh38
rs978684518 Chr.6: 16143622-16143622 on Build GRCh38
rs975465422 Chr.6: 16143611-16143611 on Build GRCh38
rs749160939 Chr.6: 16143853-16143853 on Build GRCh38
rs376620126 Chr.6: 16143889-16143889 on Build GRCh38
rs371230170 Chr.6: 16143823-16143823 on Build GRCh38
rs940276828 Chr.6: 16144048-16144048 on Build GRCh38
rs189866709 Chr.6: 16144027-16144027 on Build GRCh38
rs948545093 Chr.6: 16143983-16143983 on Build GRCh38
rs371593218 Chr.6: 16143688-16143688 on Build GRCh38
rs992699508 Chr.6: 16143957-16143957 on Build GRCh38
rs573401763 Chr.6: 16144009-16144009 on Build GRCh38
rs138175799 Chr.6: 16143755-16143755 on Build GRCh38
rs772380564 Chr.6: 16143861-16143861 on Build GRCh38
rs761323849 Chr.6: 16143721-16143721 on Build GRCh38
rs947402420 Chr.6: 16143645-16143645 on Build GRCh38
rs757959611 Chr.6: 16144029-16144029 on Build GRCh38
rs760153641 Chr.6: 16143898-16143898 on Build GRCh38
rs140483917 Chr.6: 16143708-16143708 on Build GRCh38
rs1027618230 Chr.6: 16143590-16143590 on Build GRCh38
rs767024725 Chr.6: 16143816-16143816 on Build GRCh38
rs777281734 Chr.6: 16143897-16143897 on Build GRCh38
rs767052483 Chr.6: 16143844-16143844 on Build GRCh38
rs748472377 Chr.6: 16143791-16143791 on Build GRCh38
rs368981890 Chr.6: 16143660-16143660 on Build GRCh38
rs562047496 Chr.6: 16144037-16144037 on Build GRCh38
rs770710256 Chr.6: 16143653-16143653 on Build GRCh38
rs774588479 Chr.6: 16143696-16143696 on Build GRCh38
rs917059051 Chr.6: 16143968-16143968 on Build GRCh38
rs553822291 Chr.6: 16143935-16143935 on Build GRCh38
rs756090946 Chr.6: 16143768-16143768 on Build GRCh38
rs564252332 Chr.6: 16143624-16143624 on Build GRCh38
rs149059329 Chr.6: 16144052-16144052 on Build GRCh38
rs965834103 Chr.6: 16143588-16143588 on Build GRCh38
rs781005169 Chr.6: 16143657-16143657 on Build GRCh38
rs2072784 Chr.6: 16143633-16143633 on Build GRCh38
rs754869754 Chr.6: 16143855-16143855 on Build GRCh38
rs1004368023 Chr.6: 16143761-16143761 on Build GRCh38
rs566688451 Chr.6: 16143743-16143743 on Build GRCh38
rs749397478 Chr.6: 16143781-16143781 on Build GRCh38
rs544194779 Chr.6: 16144053-16144053 on Build GRCh38
rs76272508 Chr.6: 16144003-16144003 on Build GRCh38
rs921261226 Chr.6: 16143616-16143616 on Build GRCh38
rs751165545 Chr.6: 16143744-16143744 on Build GRCh38
rs978735595 Chr.6: 16143933-16143933 on Build GRCh38
rs774445093 Chr.6: 16143784-16143784 on Build GRCh38
rs988886720 Chr.6: 16143975-16143975 on Build GRCh38
rs767859655 Chr.6: 16143703-16143703 on Build GRCh38
rs887385455 Chr.6: 16144033-16144033 on Build GRCh38
rs752296595 Chr.6: 16143732-16143732 on Build GRCh38
rs750254052 Chr.6: 16143848-16143848 on Build GRCh38
rs201781624 Chr.6: 16143815-16143815 on Build GRCh38
rs1048523287 Chr.6: 16144057-16144057 on Build GRCh38
rs150429938 Chr.6: 16143722-16143722 on Build GRCh38
rs763592472 Chr.6: 16143832-16143832 on Build GRCh38
rs771362963 Chr.6: 16143890-16143890 on Build GRCh38
rs549317876 Chr.6: 16143774-16143774 on Build GRCh38
rs769712209 Chr.6: 16143684-16143684 on Build GRCh38
rs773528802 Chr.6: 16143867-16143867 on Build GRCh38
rs779109727 Chr.6: 16143858-16143858 on Build GRCh38
rs572389874 Chr.6: 16143936-16143936 on Build GRCh38
rs369174288 Chr.6: 16143837-16143837 on Build GRCh38
rs768901842 Chr.6: 16143690-16143690 on Build GRCh38
rs770715660 Chr.6: 16143822-16143822 on Build GRCh38
rs968791781 Chr.6: 16144002-16144002 on Build GRCh38

View Full Product Details