Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 43059553-43060052 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLC4
Entrez Gene ID
89953
Gene Name
kinesin light chain 4
Gene Aliases
KNSL8, bA387M24.3
UniGene
Hs.655123
Gene Chromosome Location
Chr.6: 43059594-43075099 on Build GRCh38
Transcripts
NM_001289034.1 NM_001289035.1 NM_138343.3 NM_201521.2 NM_201522.2 NM_201523.2 XM_011514989.1 XM_017011457.1 XM_005249161.4

Gene Symbol
MRPL2
Entrez Gene ID
51069
Gene Name
mitochondrial ribosomal protein L2
Gene Aliases
CGI-22, MRP-L14, RPML14
UniGene
Hs.55041
Gene Chromosome Location
Chr.6: 43054029-43059863 on Build GRCh38
Transcripts
NM_001289034.1 NM_001289035.1 NM_138343.3 NM_201521.2 NM_201522.2 NM_201523.2 XM_011514989.1 XM_017011457.1 XM_005249161.4

SNP Detail

SNP ID Chromosome Location
rs904357182 Chr.6: 43059678-43059678 on Build GRCh38
rs910225245 Chr.6: 43059895-43059895 on Build GRCh38
rs551198086 Chr.6: 43059928-43059928 on Build GRCh38
rs371929554 Chr.6: 43059852-43059852 on Build GRCh38
rs201875814 Chr.6: 43059587-43059587 on Build GRCh38
rs772413100 Chr.6: 43059597-43059597 on Build GRCh38
rs560275367 Chr.6: 43059838-43059838 on Build GRCh38
rs1037721519 Chr.6: 43059982-43059982 on Build GRCh38
rs759722034 Chr.6: 43059920-43059920 on Build GRCh38
rs918982718 Chr.6: 43059822-43059822 on Build GRCh38
rs992663150 Chr.6: 43059906-43059906 on Build GRCh38
rs533640685 Chr.6: 43059976-43059976 on Build GRCh38
rs1016660769 Chr.6: 43059744-43059744 on Build GRCh38
rs547497838 Chr.6: 43059587-43059588 on Build GRCh38
rs181359651 Chr.6: 43059704-43059704 on Build GRCh38
rs998914587 Chr.6: 43059683-43059683 on Build GRCh38
rs886515223 Chr.6: 43059720-43059720 on Build GRCh38
rs1005767494 Chr.6: 43059768-43059768 on Build GRCh38
rs1042876380 Chr.6: 43059670-43059670 on Build GRCh38
rs886548820 Chr.6: 43059940-43059940 on Build GRCh38
rs557531263 Chr.6: 43059849-43059852 on Build GRCh38
rs905876658 Chr.6: 43060023-43060023 on Build GRCh38
rs910467728 Chr.6: 43059661-43059661 on Build GRCh38
rs36070052 Chr.6: 43059835-43059836 on Build GRCh38
rs961542781 Chr.6: 43059796-43059796 on Build GRCh38
rs1014653916 Chr.6: 43059700-43059700 on Build GRCh38
rs900479289 Chr.6: 43060001-43060001 on Build GRCh38
rs1044342205 Chr.6: 43060013-43060013 on Build GRCh38
rs1043011471 Chr.6: 43059901-43059901 on Build GRCh38
rs138820331 Chr.6: 43059922-43059922 on Build GRCh38
rs35272167 Chr.6: 43059889-43059890 on Build GRCh38
rs571274971 Chr.6: 43059974-43059974 on Build GRCh38
rs972953921 Chr.6: 43059819-43059819 on Build GRCh38
rs751551973 Chr.6: 43060012-43060012 on Build GRCh38
rs773589744 Chr.6: 43059612-43059612 on Build GRCh38
rs987534852 Chr.6: 43059881-43059881 on Build GRCh38
rs369401741 Chr.6: 43059696-43059696 on Build GRCh38
rs548843407 Chr.6: 43059875-43059875 on Build GRCh38
rs113446316 Chr.6: 43060005-43060005 on Build GRCh38
rs544404060 Chr.6: 43059596-43059596 on Build GRCh38
rs186024408 Chr.6: 43060028-43060028 on Build GRCh38
rs929844693 Chr.6: 43059923-43059923 on Build GRCh38
rs540558021 Chr.6: 43059909-43059909 on Build GRCh38
rs965328306 Chr.6: 43059584-43059584 on Build GRCh38
rs766596885 Chr.6: 43059702-43059702 on Build GRCh38
rs564560591 Chr.6: 43059671-43059671 on Build GRCh38
rs372770513 Chr.6: 43059665-43059665 on Build GRCh38
rs1051461483 Chr.6: 43059646-43059646 on Build GRCh38
rs988538022 Chr.6: 43059599-43059599 on Build GRCh38
rs747481808 Chr.6: 43059753-43059753 on Build GRCh38
rs947041966 Chr.6: 43059652-43059652 on Build GRCh38
rs926920217 Chr.6: 43059886-43059886 on Build GRCh38
rs758772404 Chr.6: 43059778-43059778 on Build GRCh38
rs978477708 Chr.6: 43059853-43059856 on Build GRCh38
rs918906519 Chr.6: 43059580-43059580 on Build GRCh38
rs943226492 Chr.6: 43059897-43059897 on Build GRCh38
rs988569087 Chr.6: 43059844-43059847 on Build GRCh38
rs1053628176 Chr.6: 43059694-43059694 on Build GRCh38
rs114646323 Chr.6: 43059732-43059732 on Build GRCh38
rs866718898 Chr.6: 43059673-43059673 on Build GRCh38
rs942097606 Chr.6: 43059942-43059942 on Build GRCh38
rs996804901 Chr.6: 43059752-43059752 on Build GRCh38
rs895887247 Chr.6: 43059993-43059993 on Build GRCh38
rs776023717 Chr.6: 43059621-43059621 on Build GRCh38

View Full Product Details