Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 43528737-43529237 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLR1C
Entrez Gene ID
9533
Gene Name
RNA polymerase I subunit C
Gene Aliases
AC40, HLD11, RPA39, RPA40, RPA5, RPAC1, RPC40, TCS3
UniGene
Hs.743509
Gene Chromosome Location
Chr.6: 43517024-43562404 on Build GRCh38
Transcripts
NM_001318876.1 XM_011515000.2 NM_020750.2

Gene Symbol
XPO5
Entrez Gene ID
57510
Gene Name
exportin 5
Gene Aliases
exp5
UniGene
Hs.203206
Gene Chromosome Location
Chr.6: 43522330-43576075 on Build GRCh38
Transcripts
NM_001318876.1 XM_011515000.2 NM_020750.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3396650 c.2706C>A p.F902L Chr.6: 43528897-43528897 on Build GRCh38
6853517 c.2774T>C p.M925T Chr.6: 43528829-43528829 on Build GRCh38
7079185 c.2725G>A p.E909K Chr.6: 43528878-43528878 on Build GRCh38
1444742 c.2679T>G p.R893R Chr.6: 43528924-43528924 on Build GRCh38
22090 c.2678G>A p.R893H Chr.6: 43528925-43528925 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1111786 Chr.6: 43529131-43529131 on Build GRCh38
rs771432017 Chr.6: 43528806-43528806 on Build GRCh38
rs751500353 Chr.6: 43528858-43528858 on Build GRCh38
rs1041376709 Chr.6: 43529120-43529120 on Build GRCh38
rs909998814 Chr.6: 43529163-43529163 on Build GRCh38
rs921367110 Chr.6: 43529058-43529058 on Build GRCh38
rs1022026749 Chr.6: 43528777-43528777 on Build GRCh38
rs965479107 Chr.6: 43529044-43529044 on Build GRCh38
rs534453356 Chr.6: 43528795-43528795 on Build GRCh38
rs774365742 Chr.6: 43529200-43529200 on Build GRCh38
rs914451551 Chr.6: 43529101-43529101 on Build GRCh38
rs775634114 Chr.6: 43528958-43528958 on Build GRCh38
rs776922426 Chr.6: 43528965-43528966 on Build GRCh38
rs200634355 Chr.6: 43528761-43528761 on Build GRCh38
rs765427694 Chr.6: 43528819-43528819 on Build GRCh38
rs888529858 Chr.6: 43528997-43528997 on Build GRCh38
rs373586741 Chr.6: 43528928-43528928 on Build GRCh38
rs561324245 Chr.6: 43529105-43529105 on Build GRCh38
rs532631004 Chr.6: 43529184-43529184 on Build GRCh38
rs778502782 Chr.6: 43528892-43528892 on Build GRCh38
rs764384609 Chr.6: 43529153-43529153 on Build GRCh38
rs777099513 Chr.6: 43528808-43528808 on Build GRCh38
rs770192857 Chr.6: 43528957-43528957 on Build GRCh38
rs771519947 Chr.6: 43528937-43528937 on Build GRCh38
rs759827821 Chr.6: 43528814-43528814 on Build GRCh38
rs759465779 Chr.6: 43528967-43528968 on Build GRCh38
rs146489790 Chr.6: 43529064-43529064 on Build GRCh38
rs370480839 Chr.6: 43528854-43528854 on Build GRCh38
rs748786340 Chr.6: 43528790-43528790 on Build GRCh38
rs768205007 Chr.6: 43528794-43528794 on Build GRCh38
rs764175153 Chr.6: 43528855-43528855 on Build GRCh38
rs899786465 Chr.6: 43529011-43529011 on Build GRCh38
rs376338326 Chr.6: 43528891-43528891 on Build GRCh38
rs779731474 Chr.6: 43528789-43528789 on Build GRCh38
rs750264133 Chr.6: 43528869-43528869 on Build GRCh38
rs200844834 Chr.6: 43529213-43529213 on Build GRCh38
rs575564678 Chr.6: 43529063-43529063 on Build GRCh38
rs1022471708 Chr.6: 43529099-43529099 on Build GRCh38
rs761209381 Chr.6: 43528798-43528798 on Build GRCh38
rs892145138 Chr.6: 43529071-43529071 on Build GRCh38
rs929977773 Chr.6: 43529155-43529156 on Build GRCh38
rs144114311 Chr.6: 43528970-43528970 on Build GRCh38
rs746258057 Chr.6: 43528955-43528955 on Build GRCh38
rs556992692 Chr.6: 43529045-43529045 on Build GRCh38
rs780986096 Chr.6: 43528868-43528868 on Build GRCh38
rs777256955 Chr.6: 43528953-43528953 on Build GRCh38
rs755903379 Chr.6: 43528872-43528872 on Build GRCh38
rs917482354 Chr.6: 43529151-43529151 on Build GRCh38
rs781727509 Chr.6: 43529043-43529043 on Build GRCh38
rs186651686 Chr.6: 43529148-43529148 on Build GRCh38
rs941228411 Chr.6: 43529000-43529000 on Build GRCh38
rs1106841 Chr.6: 43528924-43528924 on Build GRCh38
rs752933412 Chr.6: 43528821-43528821 on Build GRCh38
rs937556059 Chr.6: 43528887-43528887 on Build GRCh38
rs538910409 Chr.6: 43528972-43528972 on Build GRCh38
rs1003072850 Chr.6: 43528769-43528769 on Build GRCh38
rs779637378 Chr.6: 43528879-43528879 on Build GRCh38
rs550172239 Chr.6: 43528925-43528925 on Build GRCh38
rs540775608 Chr.6: 43529142-43529142 on Build GRCh38
rs373330630 Chr.6: 43528880-43528880 on Build GRCh38
rs1016044762 Chr.6: 43529005-43529005 on Build GRCh38
rs982766590 Chr.6: 43529162-43529162 on Build GRCh38
rs1009207440 Chr.6: 43529072-43529072 on Build GRCh38
rs757146568 Chr.6: 43528861-43528861 on Build GRCh38
rs774838633 Chr.6: 43529191-43529191 on Build GRCh38

View Full Product Details