Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154945891-154946386 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PBXIP1
Entrez Gene ID
57326
Gene Name
PBX homeobox interacting protein 1
Gene Aliases
HPIP
UniGene
Hs.505806
Gene Chromosome Location
Chr.1: 154944080-154956163 on Build GRCh38
Transcripts
NM_001317734.1 NM_001317735.1 NM_020524.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
463070 c.1553A>C p.K518T Chr.1: 154946121-154946121 on Build GRCh38
6738487 c.1304G>A p.R435K Chr.1: 154946370-154946370 on Build GRCh38
5798831 c.1766C>G p.S589* Chr.1: 154945908-154945908 on Build GRCh38
6738483 c.1454G>A p.R485Q Chr.1: 154946220-154946220 on Build GRCh38
2209403 c.1335C>A p.N445K Chr.1: 154946339-154946339 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780861473 Chr.1: 154946044-154946044 on Build GRCh38
rs372327280 Chr.1: 154945959-154945959 on Build GRCh38
rs549911070 Chr.1: 154946021-154946021 on Build GRCh38
rs144596543 Chr.1: 154946085-154946085 on Build GRCh38
rs746249442 Chr.1: 154946195-154946195 on Build GRCh38
rs766278866 Chr.1: 154946127-154946127 on Build GRCh38
rs775576270 Chr.1: 154945971-154945971 on Build GRCh38
rs1036989166 Chr.1: 154946356-154946356 on Build GRCh38
rs770976660 Chr.1: 154946255-154946255 on Build GRCh38
rs750831105 Chr.1: 154946327-154946327 on Build GRCh38
rs763216826 Chr.1: 154946363-154946363 on Build GRCh38
rs558780475 Chr.1: 154946222-154946222 on Build GRCh38
rs752416542 Chr.1: 154946294-154946294 on Build GRCh38
rs537557788 Chr.1: 154946048-154946048 on Build GRCh38
rs564342820 Chr.1: 154945927-154945927 on Build GRCh38
rs750255589 Chr.1: 154945998-154945998 on Build GRCh38
rs890641219 Chr.1: 154946076-154946076 on Build GRCh38
rs12760148 Chr.1: 154946268-154946268 on Build GRCh38
rs368629922 Chr.1: 154945946-154945946 on Build GRCh38
rs199952236 Chr.1: 154946174-154946174 on Build GRCh38
rs771104168 Chr.1: 154946138-154946138 on Build GRCh38
rs758049081 Chr.1: 154945911-154945911 on Build GRCh38
rs771183280 Chr.1: 154945936-154945936 on Build GRCh38
rs529260163 Chr.1: 154946083-154946083 on Build GRCh38
rs373071090 Chr.1: 154946312-154946312 on Build GRCh38
rs760808622 Chr.1: 154946106-154946106 on Build GRCh38
rs375178576 Chr.1: 154945919-154945919 on Build GRCh38
rs751055768 Chr.1: 154946223-154946224 on Build GRCh38
rs758240858 Chr.1: 154945999-154945999 on Build GRCh38
rs138826372 Chr.1: 154946046-154946046 on Build GRCh38
rs772350583 Chr.1: 154946199-154946199 on Build GRCh38
rs983795779 Chr.1: 154946281-154946281 on Build GRCh38
rs747146952 Chr.1: 154946212-154946212 on Build GRCh38
rs915627261 Chr.1: 154945967-154945967 on Build GRCh38
rs762940530 Chr.1: 154946124-154946124 on Build GRCh38
rs763666133 Chr.1: 154946320-154946320 on Build GRCh38
rs751340150 Chr.1: 154946128-154946128 on Build GRCh38
rs759479530 Chr.1: 154946237-154946239 on Build GRCh38
rs780296483 Chr.1: 154946206-154946206 on Build GRCh38
rs775493267 Chr.1: 154946094-154946094 on Build GRCh38
rs192315620 Chr.1: 154946279-154946279 on Build GRCh38
rs1037319940 Chr.1: 154946060-154946060 on Build GRCh38
rs200974199 Chr.1: 154946071-154946071 on Build GRCh38
rs758808885 Chr.1: 154946340-154946340 on Build GRCh38
rs374693871 Chr.1: 154945997-154945997 on Build GRCh38
rs761672762 Chr.1: 154945990-154945990 on Build GRCh38
rs762522814 Chr.1: 154946116-154946118 on Build GRCh38
rs747077154 Chr.1: 154946078-154946078 on Build GRCh38
rs551771123 Chr.1: 154946186-154946186 on Build GRCh38
rs759372832 Chr.1: 154946278-154946278 on Build GRCh38
rs377754535 Chr.1: 154946343-154946343 on Build GRCh38
rs377123006 Chr.1: 154946168-154946168 on Build GRCh38
rs547352737 Chr.1: 154946116-154946116 on Build GRCh38
rs113959089 Chr.1: 154945993-154945993 on Build GRCh38
rs768808417 Chr.1: 154946220-154946220 on Build GRCh38
rs141826563 Chr.1: 154946149-154946149 on Build GRCh38
rs778312189 Chr.1: 154946066-154946066 on Build GRCh38
rs148829554 Chr.1: 154946322-154946322 on Build GRCh38
rs79076478 Chr.1: 154946201-154946201 on Build GRCh38
rs767193192 Chr.1: 154946160-154946160 on Build GRCh38
rs760205696 Chr.1: 154946310-154946310 on Build GRCh38
rs529270147 Chr.1: 154946263-154946263 on Build GRCh38
rs774354445 Chr.1: 154946264-154946264 on Build GRCh38
rs150642338 Chr.1: 154946178-154946178 on Build GRCh38
rs754608491 Chr.1: 154946030-154946030 on Build GRCh38
rs779266116 Chr.1: 154946191-154946191 on Build GRCh38
rs772226029 Chr.1: 154946093-154946093 on Build GRCh38
rs754375858 Chr.1: 154946182-154946182 on Build GRCh38
rs939507097 Chr.1: 154945943-154945943 on Build GRCh38
rs754555288 Chr.1: 154945923-154945923 on Build GRCh38
rs779391134 Chr.1: 154946325-154946325 on Build GRCh38
rs146377687 Chr.1: 154946179-154946179 on Build GRCh38
rs765132144 Chr.1: 154946110-154946110 on Build GRCh38
rs368909325 Chr.1: 154946061-154946061 on Build GRCh38
rs150327322 Chr.1: 154945942-154945942 on Build GRCh38
rs370421303 Chr.1: 154946172-154946172 on Build GRCh38
rs765210465 Chr.1: 154945992-154945992 on Build GRCh38
rs749660304 Chr.1: 154945932-154945932 on Build GRCh38
rs752188899 Chr.1: 154946163-154946163 on Build GRCh38
rs765844541 Chr.1: 154946163-154946165 on Build GRCh38
rs767617186 Chr.1: 154945970-154945970 on Build GRCh38
rs199769816 Chr.1: 154945926-154945926 on Build GRCh38
rs749664136 Chr.1: 154946070-154946070 on Build GRCh38
rs137982788 Chr.1: 154946012-154946012 on Build GRCh38
rs1028429983 Chr.1: 154946111-154946111 on Build GRCh38
rs770215901 Chr.1: 154946221-154946221 on Build GRCh38
rs762995182 Chr.1: 154946229-154946229 on Build GRCh38
rs901037795 Chr.1: 154946065-154946065 on Build GRCh38

View Full Product Details