Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 126340029-126340536 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPW
Entrez Gene ID
387103
Gene Name
centromere protein W
Gene Aliases
C6orf173, CENP-W, CUG2
UniGene
Hs.486401
Gene Chromosome Location
Chr.6: 126339696-126483320 on Build GRCh38
Transcripts
NM_001012507.3 NM_001286524.1 NM_001286525.1 NR_104462.1 XM_017010845.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4933577 c.22T>C p.S8P Chr.6: 126340295-126340295 on Build GRCh38
3374032 c.71G>A p.R24Q Chr.6: 126340344-126340344 on Build GRCh38
6424202 c.164C>T p.T55M Chr.6: 126340437-126340437 on Build GRCh38
4890035 c.16A>G p.I6V Chr.6: 126340289-126340289 on Build GRCh38
739324 c.36G>C p.Q12H Chr.6: 126340309-126340309 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750981982 Chr.6: 126340396-126340396 on Build GRCh38
rs938904787 Chr.6: 126340067-126340067 on Build GRCh38
rs759230009 Chr.6: 126340363-126340363 on Build GRCh38
rs766480500 Chr.6: 126340412-126340412 on Build GRCh38
rs544986292 Chr.6: 126340062-126340062 on Build GRCh38
rs972300110 Chr.6: 126340051-126340051 on Build GRCh38
rs528679207 Chr.6: 126340195-126340195 on Build GRCh38
rs764549124 Chr.6: 126340248-126340248 on Build GRCh38
rs762147073 Chr.6: 126340384-126340384 on Build GRCh38
rs1055985078 Chr.6: 126340107-126340107 on Build GRCh38
rs1004188680 Chr.6: 126340079-126340079 on Build GRCh38
rs769333541 Chr.6: 126340177-126340177 on Build GRCh38
rs777804656 Chr.6: 126340440-126340440 on Build GRCh38
rs1037027979 Chr.6: 126340324-126340324 on Build GRCh38
rs551526114 Chr.6: 126340329-126340329 on Build GRCh38
rs750494370 Chr.6: 126340289-126340289 on Build GRCh38
rs759992998 Chr.6: 126340451-126340451 on Build GRCh38
rs775042733 Chr.6: 126340477-126340477 on Build GRCh38
rs947433842 Chr.6: 126340141-126340141 on Build GRCh38
rs769794204 Chr.6: 126340430-126340431 on Build GRCh38
rs569626276 Chr.6: 126340332-126340332 on Build GRCh38
rs755321237 Chr.6: 126340064-126340064 on Build GRCh38
rs866043423 Chr.6: 126340383-126340383 on Build GRCh38
rs934732760 Chr.6: 126340486-126340486 on Build GRCh38
rs760899266 Chr.6: 126340238-126340238 on Build GRCh38
rs762610865 Chr.6: 126340226-126340226 on Build GRCh38
rs781329059 Chr.6: 126340420-126340420 on Build GRCh38
rs766390386 Chr.6: 126340494-126340494 on Build GRCh38
rs971339104 Chr.6: 126340207-126340207 on Build GRCh38
rs79157385 Chr.6: 126340129-126340129 on Build GRCh38
rs541914710 Chr.6: 126340437-126340437 on Build GRCh38
rs916907796 Chr.6: 126340202-126340202 on Build GRCh38
rs773673369 Chr.6: 126340491-126340491 on Build GRCh38
rs182439161 Chr.6: 126340478-126340478 on Build GRCh38
rs901462146 Chr.6: 126340100-126340100 on Build GRCh38
rs996741500 Chr.6: 126340208-126340208 on Build GRCh38
rs533023379 Chr.6: 126340318-126340318 on Build GRCh38
rs79621298 Chr.6: 126340482-126340482 on Build GRCh38
rs747281256 Chr.6: 126340305-126340305 on Build GRCh38
rs1041166529 Chr.6: 126340507-126340507 on Build GRCh38
rs754229837 Chr.6: 126340256-126340256 on Build GRCh38
rs866312787 Chr.6: 126340144-126340144 on Build GRCh38
rs375772295 Chr.6: 126340446-126340446 on Build GRCh38
rs781048571 Chr.6: 126340328-126340328 on Build GRCh38
rs930460737 Chr.6: 126340271-126340271 on Build GRCh38
rs1028486958 Chr.6: 126340143-126340143 on Build GRCh38
rs1028265884 Chr.6: 126340210-126340210 on Build GRCh38
rs771748592 Chr.6: 126340471-126340471 on Build GRCh38
rs1015991925 Chr.6: 126340094-126340094 on Build GRCh38
rs745787203 Chr.6: 126340267-126340268 on Build GRCh38
rs34550377 Chr.6: 126340463-126340464 on Build GRCh38
rs758508637 Chr.6: 126340291-126340291 on Build GRCh38
rs765698258 Chr.6: 126340391-126340391 on Build GRCh38
rs745424998 Chr.6: 126340463-126340463 on Build GRCh38
rs372192318 Chr.6: 126340147-126340147 on Build GRCh38
rs909945926 Chr.6: 126340316-126340316 on Build GRCh38
rs772863586 Chr.6: 126340224-126340224 on Build GRCh38
rs763458606 Chr.6: 126340493-126340493 on Build GRCh38
rs567495845 Chr.6: 126340366-126340366 on Build GRCh38
rs756014160 Chr.6: 126340438-126340438 on Build GRCh38
rs142786698 Chr.6: 126340296-126340296 on Build GRCh38
rs758893982 Chr.6: 126340223-126340223 on Build GRCh38
rs764306077 Chr.6: 126340365-126340365 on Build GRCh38
rs367748361 Chr.6: 126340149-126340149 on Build GRCh38
rs754218294 Chr.6: 126340088-126340088 on Build GRCh38
rs758426987 Chr.6: 126340397-126340397 on Build GRCh38
rs982278973 Chr.6: 126340374-126340374 on Build GRCh38
rs41285260 Chr.6: 126340356-126340356 on Build GRCh38
rs186040085 Chr.6: 126340179-126340179 on Build GRCh38
rs190907582 Chr.6: 126340215-126340215 on Build GRCh38
rs372798798 Chr.6: 126340444-126340444 on Build GRCh38
rs201673480 Chr.6: 126340350-126340350 on Build GRCh38
rs367746527 Chr.6: 126340159-126340159 on Build GRCh38
rs375635063 Chr.6: 126340419-126340419 on Build GRCh38
rs773350989 Chr.6: 126340345-126340345 on Build GRCh38
rs757793929 Chr.6: 126340269-126340269 on Build GRCh38
rs201770676 Chr.6: 126340230-126340230 on Build GRCh38
rs971014146 Chr.6: 126340510-126340510 on Build GRCh38
rs751305549 Chr.6: 126340236-126340236 on Build GRCh38

View Full Product Details