Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 132327922-132328432 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MOXD1
Entrez Gene ID
26002
Gene Name
monooxygenase DBH like 1
Gene Aliases
MOX, PRO5780, dJ248E1.1
UniGene
Hs.6909
Gene Chromosome Location
Chr.6: 132296055-132401534 on Build GRCh38
Transcripts
NM_015529.3 XM_017010714.1 XM_017010715.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5766787 c.843+7C>A p.? Chr.6: 132328408-132328408 on Build GRCh38
382431 c.876C>A p.S292S Chr.6: 132328083-132328083 on Build GRCh38
4912502 c.835G>T p.G279C Chr.6: 132328423-132328423 on Build GRCh38
5766788 c.639+7C>A p.? Chr.6: 132328408-132328408 on Build GRCh38
1073363 c.697T>G p.Y233D Chr.6: 132328058-132328058 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755002490 Chr.6: 132327968-132327968 on Build GRCh38
rs964004747 Chr.6: 132328247-132328247 on Build GRCh38
rs775322297 Chr.6: 132328404-132328404 on Build GRCh38
rs1007400013 Chr.6: 132328144-132328144 on Build GRCh38
rs750501782 Chr.6: 132328042-132328042 on Build GRCh38
rs924846024 Chr.6: 132328043-132328043 on Build GRCh38
rs560879034 Chr.6: 132328099-132328099 on Build GRCh38
rs374751992 Chr.6: 132328063-132328063 on Build GRCh38
rs760457577 Chr.6: 132328409-132328409 on Build GRCh38
rs774381497 Chr.6: 132328116-132328116 on Build GRCh38
rs887706416 Chr.6: 132328359-132328359 on Build GRCh38
rs748514692 Chr.6: 132328165-132328165 on Build GRCh38
rs762938062 Chr.6: 132328103-132328103 on Build GRCh38
rs767702851 Chr.6: 132328400-132328400 on Build GRCh38
rs924909467 Chr.6: 132328294-132328294 on Build GRCh38
rs370593936 Chr.6: 132328048-132328048 on Build GRCh38
rs576772098 Chr.6: 132328353-132328353 on Build GRCh38
rs534022785 Chr.6: 132328271-132328271 on Build GRCh38
rs373611028 Chr.6: 132328164-132328164 on Build GRCh38
rs139692804 Chr.6: 132328036-132328036 on Build GRCh38
rs868402008 Chr.6: 132328258-132328258 on Build GRCh38
rs748724921 Chr.6: 132328064-132328064 on Build GRCh38
rs778639166 Chr.6: 132328158-132328158 on Build GRCh38
rs1017195029 Chr.6: 132328221-132328221 on Build GRCh38
rs756651392 Chr.6: 132328157-132328157 on Build GRCh38
rs769236976 Chr.6: 132327987-132327987 on Build GRCh38
rs371015456 Chr.6: 132327971-132327971 on Build GRCh38
rs762452148 Chr.6: 132328097-132328097 on Build GRCh38
rs777207963 Chr.6: 132328038-132328038 on Build GRCh38
rs554604197 Chr.6: 132328386-132328386 on Build GRCh38
rs775970970 Chr.6: 132328032-132328032 on Build GRCh38
rs754663140 Chr.6: 132328053-132328053 on Build GRCh38
rs773403183 Chr.6: 132328102-132328102 on Build GRCh38
rs760932034 Chr.6: 132328034-132328034 on Build GRCh38
rs758095663 Chr.6: 132328163-132328163 on Build GRCh38
rs770869108 Chr.6: 132328370-132328370 on Build GRCh38
rs146625320 Chr.6: 132328065-132328065 on Build GRCh38
rs977966133 Chr.6: 132328282-132328282 on Build GRCh38
rs751439225 Chr.6: 132328052-132328052 on Build GRCh38
rs745458279 Chr.6: 132328161-132328161 on Build GRCh38
rs187976345 Chr.6: 132328235-132328235 on Build GRCh38
rs777312330 Chr.6: 132328096-132328096 on Build GRCh38
rs764005730 Chr.6: 132328146-132328146 on Build GRCh38
rs200008764 Chr.6: 132328013-132328013 on Build GRCh38
rs563471641 Chr.6: 132328348-132328348 on Build GRCh38
rs199511460 Chr.6: 132328123-132328123 on Build GRCh38
rs773525452 Chr.6: 132328366-132328366 on Build GRCh38
rs777424313 Chr.6: 132328012-132328012 on Build GRCh38
rs778422135 Chr.6: 132328071-132328071 on Build GRCh38
rs993316267 Chr.6: 132328132-132328132 on Build GRCh38
rs1047980304 Chr.6: 132328351-132328351 on Build GRCh38
rs749003675 Chr.6: 132328364-132328364 on Build GRCh38
rs542546100 Chr.6: 132328363-132328363 on Build GRCh38
rs955569158 Chr.6: 132328278-132328278 on Build GRCh38
rs144409218 Chr.6: 132328062-132328062 on Build GRCh38
rs183408441 Chr.6: 132328332-132328332 on Build GRCh38
rs759793102 Chr.6: 132328392-132328392 on Build GRCh38
rs560081252 Chr.6: 132328149-132328149 on Build GRCh38
rs1045500725 Chr.6: 132328118-132328118 on Build GRCh38
rs767259255 Chr.6: 132328130-132328130 on Build GRCh38
rs766499776 Chr.6: 132328049-132328049 on Build GRCh38
rs769081184 Chr.6: 132328079-132328079 on Build GRCh38
rs141413110 Chr.6: 132328319-132328319 on Build GRCh38
rs6938195 Chr.6: 132328172-132328172 on Build GRCh38
rs3736698 Chr.6: 132328184-132328184 on Build GRCh38
rs936211987 Chr.6: 132328296-132328296 on Build GRCh38
rs749399897 Chr.6: 132328367-132328367 on Build GRCh38
rs537757336 Chr.6: 132328211-132328211 on Build GRCh38
rs527981136 Chr.6: 132327952-132327952 on Build GRCh38
rs758533142 Chr.6: 132328220-132328220 on Build GRCh38
rs960850314 Chr.6: 132328272-132328272 on Build GRCh38
rs747661667 Chr.6: 132327975-132327975 on Build GRCh38
rs764092444 Chr.6: 132328410-132328410 on Build GRCh38
rs377425439 Chr.6: 132328072-132328072 on Build GRCh38
rs192506527 Chr.6: 132328255-132328255 on Build GRCh38
rs770458019 Chr.6: 132328014-132328014 on Build GRCh38
rs752455928 Chr.6: 132328139-132328139 on Build GRCh38
rs796930954 Chr.6: 132328281-132328281 on Build GRCh38
rs765370417 Chr.6: 132328041-132328041 on Build GRCh38
rs866276317 Chr.6: 132328078-132328078 on Build GRCh38
rs989840668 Chr.6: 132328304-132328304 on Build GRCh38
rs1038845848 Chr.6: 132328147-132328147 on Build GRCh38
rs774530723 Chr.6: 132328273-132328276 on Build GRCh38
rs753658082 Chr.6: 132328411-132328411 on Build GRCh38
rs906922648 Chr.6: 132328334-132328334 on Build GRCh38
rs756092604 Chr.6: 132328141-132328141 on Build GRCh38
rs781122762 Chr.6: 132328057-132328057 on Build GRCh38
rs761994258 Chr.6: 132328039-132328039 on Build GRCh38
rs529711671 Chr.6: 132328110-132328110 on Build GRCh38
rs915614546 Chr.6: 132328305-132328305 on Build GRCh38
rs554272573 Chr.6: 132328318-132328318 on Build GRCh38

View Full Product Details