Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 138218113-138218611 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARFGEF3
Entrez Gene ID
57221
Gene Name
ARFGEF family member 3
Gene Aliases
A7322, BIG3, C6orf92, KIAA1244, PPP1R33, dJ171N11.1
UniGene
Hs.194408
Gene Chromosome Location
Chr.6: 138161690-138344663 on Build GRCh38
Transcripts
NM_020340.4 NM_021635.2

Gene Symbol
PBOV1
Entrez Gene ID
59351
Gene Name
prostate and breast cancer overexpressed 1
Gene Aliases
UC28, UROC28, dJ171N11.2
UniGene
Hs.302016
Gene Chromosome Location
Chr.6: 138215986-138218490 on Build GRCh38
Transcripts
NM_020340.4 NM_021635.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7000334 c.196A>G p.T66A Chr.6: 138218200-138218200 on Build GRCh38
7163844 c.271_279delAAAGGTTCC p.K91_S93delKGS Chr.6: 138218117-138218125 on Build GRCh38
7382523 c.197C>T p.T66I Chr.6: 138218199-138218199 on Build GRCh38
4778186 c.281C>A p.S94* Chr.6: 138218115-138218115 on Build GRCh38
6313447 c.232C>A p.H78N Chr.6: 138218164-138218164 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753140592 Chr.6: 138218226-138218226 on Build GRCh38
rs757860303 Chr.6: 138218199-138218199 on Build GRCh38
rs80104385 Chr.6: 138218298-138218298 on Build GRCh38
rs371672323 Chr.6: 138218583-138218583 on Build GRCh38
rs990881080 Chr.6: 138218552-138218552 on Build GRCh38
rs370519608 Chr.6: 138218411-138218411 on Build GRCh38
rs755450557 Chr.6: 138218139-138218139 on Build GRCh38
rs9494968 Chr.6: 138218190-138218190 on Build GRCh38
rs779230396 Chr.6: 138218255-138218255 on Build GRCh38
rs930895176 Chr.6: 138218361-138218361 on Build GRCh38
rs370570610 Chr.6: 138218403-138218403 on Build GRCh38
rs139990493 Chr.6: 138218476-138218476 on Build GRCh38
rs764555788 Chr.6: 138218370-138218370 on Build GRCh38
rs375150636 Chr.6: 138218546-138218546 on Build GRCh38
rs761564524 Chr.6: 138218289-138218289 on Build GRCh38
rs995904398 Chr.6: 138218480-138218480 on Build GRCh38
rs753935616 Chr.6: 138218352-138218352 on Build GRCh38
rs187792104 Chr.6: 138218575-138218575 on Build GRCh38
rs373451602 Chr.6: 138218253-138218253 on Build GRCh38
rs376330598 Chr.6: 138218182-138218182 on Build GRCh38
rs559691424 Chr.6: 138218263-138218263 on Build GRCh38
rs747102219 Chr.6: 138218415-138218415 on Build GRCh38
rs577940950 Chr.6: 138218148-138218148 on Build GRCh38
rs778010258 Chr.6: 138218149-138218149 on Build GRCh38
rs377205904 Chr.6: 138218163-138218163 on Build GRCh38
rs573153047 Chr.6: 138218421-138218421 on Build GRCh38
rs144182876 Chr.6: 138218281-138218281 on Build GRCh38
rs73774688 Chr.6: 138218235-138218235 on Build GRCh38
rs146062869 Chr.6: 138218303-138218303 on Build GRCh38
rs781625357 Chr.6: 138218145-138218145 on Build GRCh38
rs761283885 Chr.6: 138218328-138218328 on Build GRCh38
rs11758517 Chr.6: 138218419-138218419 on Build GRCh38
rs774538801 Chr.6: 138218155-138218155 on Build GRCh38
rs933363346 Chr.6: 138218469-138218469 on Build GRCh38
rs868438087 Chr.6: 138218227-138218227 on Build GRCh38
rs773566105 Chr.6: 138218266-138218266 on Build GRCh38
rs6927706 Chr.6: 138218178-138218178 on Build GRCh38
rs749512320 Chr.6: 138218150-138218150 on Build GRCh38
rs775726606 Chr.6: 138218265-138218265 on Build GRCh38
rs1050951698 Chr.6: 138218488-138218488 on Build GRCh38
rs377720782 Chr.6: 138218293-138218293 on Build GRCh38
rs868278553 Chr.6: 138218374-138218374 on Build GRCh38
rs1008548320 Chr.6: 138218443-138218443 on Build GRCh38
rs938053981 Chr.6: 138218321-138218321 on Build GRCh38
rs772541252 Chr.6: 138218222-138218222 on Build GRCh38
rs183062454 Chr.6: 138218354-138218354 on Build GRCh38
rs757523859 Chr.6: 138218389-138218389 on Build GRCh38
rs1055714686 Chr.6: 138218348-138218348 on Build GRCh38
rs749916055 Chr.6: 138218198-138218198 on Build GRCh38
rs966733370 Chr.6: 138218256-138218256 on Build GRCh38
rs752567742 Chr.6: 138218457-138218457 on Build GRCh38
rs754390771 Chr.6: 138218388-138218388 on Build GRCh38
rs377341385 Chr.6: 138218393-138218393 on Build GRCh38
rs1039991194 Chr.6: 138218459-138218459 on Build GRCh38
rs868326655 Chr.6: 138218180-138218180 on Build GRCh38
rs962751373 Chr.6: 138218221-138218221 on Build GRCh38
rs959452400 Chr.6: 138218550-138218550 on Build GRCh38
rs758753095 Chr.6: 138218407-138218407 on Build GRCh38
rs547254963 Chr.6: 138218195-138218195 on Build GRCh38
rs920676070 Chr.6: 138218570-138218570 on Build GRCh38
rs767211326 Chr.6: 138218395-138218401 on Build GRCh38
rs898335431 Chr.6: 138218516-138218516 on Build GRCh38
rs891072019 Chr.6: 138218492-138218492 on Build GRCh38
rs900271614 Chr.6: 138218477-138218477 on Build GRCh38
rs765857849 Chr.6: 138218329-138218329 on Build GRCh38
rs368997138 Chr.6: 138218300-138218300 on Build GRCh38
rs149844213 Chr.6: 138218502-138218502 on Build GRCh38
rs373323732 Chr.6: 138218236-138218236 on Build GRCh38
rs779541822 Chr.6: 138218217-138218217 on Build GRCh38
rs971285051 Chr.6: 138218533-138218533 on Build GRCh38
rs761527000 Chr.6: 138218186-138218186 on Build GRCh38
rs751096429 Chr.6: 138218346-138218346 on Build GRCh38
rs1018484436 Chr.6: 138218505-138218505 on Build GRCh38
rs369010164 Chr.6: 138218160-138218160 on Build GRCh38
rs762077228 Chr.6: 138218214-138218215 on Build GRCh38
rs768547108 Chr.6: 138218279-138218279 on Build GRCh38
rs760652024 Chr.6: 138218277-138218277 on Build GRCh38
rs574974734 Chr.6: 138218350-138218352 on Build GRCh38

View Full Product Details