Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 159727328-159727857 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SOD2
Entrez Gene ID
6648
Gene Name
superoxide dismutase 2, mitochondrial
Gene Aliases
IPO-B, IPOB, MNSOD, MVCD6, Mn-SOD
UniGene
Hs.487046
Gene Chromosome Location
Chr.6: 159679064-159762529 on Build GRCh38
Transcripts
NM_001322817.1 NM_001322819.1 NM_001270531.1 NM_001270532.1 NM_001270533.1 NM_004906.4 NM_152857.2 NM_152858.2 XM_017011514.1 XM_017011516.1

Gene Symbol
WTAP
Entrez Gene ID
9589
Gene Name
Wilms tumor 1 associated protein
Gene Aliases
Mum2
UniGene
Hs.446091
Gene Chromosome Location
Chr.6: 159726680-159756319 on Build GRCh38
Transcripts
NM_001322817.1 NM_001322819.1 NM_001270531.1 NM_001270532.1 NM_001270533.1 NM_004906.4 NM_152857.2 NM_152858.2 XM_017011514.1 XM_017011516.1

SNP Detail

SNP ID Chromosome Location
rs980020916 Chr.6: 159727630-159727630 on Build GRCh38
rs975300432 Chr.6: 159727509-159727509 on Build GRCh38
rs191151761 Chr.6: 159727441-159727441 on Build GRCh38
rs772525335 Chr.6: 159727401-159727402 on Build GRCh38
rs890717274 Chr.6: 159727791-159727791 on Build GRCh38
rs988329870 Chr.6: 159727505-159727505 on Build GRCh38
rs984454208 Chr.6: 159727796-159727796 on Build GRCh38
rs1057126473 Chr.6: 159727350-159727350 on Build GRCh38
rs182631265 Chr.6: 159727486-159727486 on Build GRCh38
rs972543999 Chr.6: 159727477-159727477 on Build GRCh38
rs908408109 Chr.6: 159727559-159727559 on Build GRCh38
rs747518702 Chr.6: 159727403-159727404 on Build GRCh38
rs748944442 Chr.6: 159727387-159727388 on Build GRCh38
rs962161314 Chr.6: 159727582-159727582 on Build GRCh38
rs939889112 Chr.6: 159727476-159727476 on Build GRCh38
rs1044402167 Chr.6: 159727588-159727588 on Build GRCh38
rs755675555 Chr.6: 159727383-159727384 on Build GRCh38
rs1029019784 Chr.6: 159727425-159727425 on Build GRCh38
rs779482252 Chr.6: 159727386-159727387 on Build GRCh38
rs111664512 Chr.6: 159727378-159727378 on Build GRCh38
rs766462112 Chr.6: 159727373-159727374 on Build GRCh38
rs1048257210 Chr.6: 159727553-159727553 on Build GRCh38
rs931243311 Chr.6: 159727528-159727528 on Build GRCh38
rs1044271947 Chr.6: 159727492-159727492 on Build GRCh38
rs921170610 Chr.6: 159727512-159727512 on Build GRCh38
rs1043701181 Chr.6: 159727470-159727487 on Build GRCh38
rs773728704 Chr.6: 159727402-159727403 on Build GRCh38
rs559838648 Chr.6: 159727415-159727415 on Build GRCh38
rs779186891 Chr.6: 159727391-159727392 on Build GRCh38
rs781413171 Chr.6: 159727352-159727352 on Build GRCh38
rs917306660 Chr.6: 159727816-159727816 on Build GRCh38
rs867150630 Chr.6: 159727556-159727556 on Build GRCh38
rs1056805779 Chr.6: 159727669-159727669 on Build GRCh38
rs919783630 Chr.6: 159727608-159727608 on Build GRCh38
rs905553549 Chr.6: 159727501-159727501 on Build GRCh38
rs749516659 Chr.6: 159727377-159727378 on Build GRCh38
rs946224165 Chr.6: 159727572-159727572 on Build GRCh38
rs913491366 Chr.6: 159727502-159727502 on Build GRCh38
rs912191726 Chr.6: 159727506-159727506 on Build GRCh38
rs1009953556 Chr.6: 159727537-159727537 on Build GRCh38
rs749863358 Chr.6: 159727376-159727377 on Build GRCh38
rs1043258697 Chr.6: 159727544-159727544 on Build GRCh38
rs13198551 Chr.6: 159727407-159727407 on Build GRCh38
rs961999314 Chr.6: 159727811-159727811 on Build GRCh38
rs968827534 Chr.6: 159727399-159727399 on Build GRCh38
rs939865930 Chr.6: 159727803-159727803 on Build GRCh38
rs2475566 Chr.6: 159727709-159727709 on Build GRCh38
rs1000443296 Chr.6: 159727695-159727695 on Build GRCh38
rs776642727 Chr.6: 159727405-159727406 on Build GRCh38
rs774224221 Chr.6: 159727407-159727408 on Build GRCh38
rs975057870 Chr.6: 159727772-159727772 on Build GRCh38
rs1057462817 Chr.6: 159727564-159727564 on Build GRCh38
rs1021761618 Chr.6: 159727405-159727405 on Build GRCh38
rs968965869 Chr.6: 159727409-159727409 on Build GRCh38
rs904670470 Chr.6: 159727548-159727548 on Build GRCh38
rs965436014 Chr.6: 159727508-159727508 on Build GRCh38
rs920972585 Chr.6: 159727785-159727785 on Build GRCh38
rs895790310 Chr.6: 159727573-159727573 on Build GRCh38
rs956412406 Chr.6: 159727742-159727742 on Build GRCh38
rs902495148 Chr.6: 159727618-159727619 on Build GRCh38
rs965154088 Chr.6: 159727765-159727765 on Build GRCh38
rs891991971 Chr.6: 159727373-159727373 on Build GRCh38
rs990387028 Chr.6: 159727489-159727489 on Build GRCh38
rs564156087 Chr.6: 159727665-159727665 on Build GRCh38
rs932017006 Chr.6: 159727784-159727784 on Build GRCh38
rs748513847 Chr.6: 159727398-159727399 on Build GRCh38
rs1019796526 Chr.6: 159727755-159727755 on Build GRCh38
rs1027686886 Chr.6: 159727834-159727835 on Build GRCh38
rs188148278 Chr.6: 159727792-159727792 on Build GRCh38
rs938556538 Chr.6: 159727656-159727656 on Build GRCh38
rs765364134 Chr.6: 159727742-159727744 on Build GRCh38
rs940088923 Chr.6: 159727561-159727561 on Build GRCh38
rs1008897935 Chr.6: 159727560-159727560 on Build GRCh38
rs761734374 Chr.6: 159727406-159727407 on Build GRCh38
rs543268414 Chr.6: 159727406-159727406 on Build GRCh38
rs776889635 Chr.6: 159727374-159727375 on Build GRCh38
rs956480322 Chr.6: 159727498-159727498 on Build GRCh38
rs71033554 Chr.6: 159727399-159727405 on Build GRCh38
rs1013080548 Chr.6: 159727380-159727380 on Build GRCh38
rs948828306 Chr.6: 159727827-159727827 on Build GRCh38
rs796803487 Chr.6: 159727689-159727689 on Build GRCh38
rs776887728 Chr.6: 159727375-159727381 on Build GRCh38
rs1014939303 Chr.6: 159727802-159727802 on Build GRCh38
rs1029235056 Chr.6: 159727557-159727557 on Build GRCh38
rs771012467 Chr.6: 159727404-159727405 on Build GRCh38
rs927229874 Chr.6: 159727648-159727649 on Build GRCh38
rs954717951 Chr.6: 159727432-159727432 on Build GRCh38
rs904764580 Chr.6: 159727628-159727628 on Build GRCh38
rs923350178 Chr.6: 159727741-159727741 on Build GRCh38
rs376992337 Chr.6: 159727757-159727757 on Build GRCh38
rs987990774 Chr.6: 159727463-159727463 on Build GRCh38
rs551715992 Chr.6: 159727487-159727487 on Build GRCh38
rs1051159294 Chr.6: 159727752-159727752 on Build GRCh38
rs908334536 Chr.6: 159727800-159727800 on Build GRCh38
rs745637479 Chr.6: 159727387-159727387 on Build GRCh38
rs1012881855 Chr.6: 159727587-159727587 on Build GRCh38
rs549922853 Chr.6: 159727775-159727775 on Build GRCh38
rs1009295710 Chr.6: 159727749-159727749 on Build GRCh38
rs1057187588 Chr.6: 159727804-159727804 on Build GRCh38
rs945795599 Chr.6: 159727703-159727703 on Build GRCh38

View Full Product Details