Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 19145068-19145610 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FERD3L
Entrez Gene ID
222894
Gene Name
Fer3-like bHLH transcription factor
Gene Aliases
N-TWIST, NATO3, NTWIST, bHLHa31
UniGene
Hs.592168
Gene Chromosome Location
Chr.7: 19144782-19145421 on Build GRCh38
Transcripts
NM_152898.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3832457 c.229G>T p.E77* Chr.7: 19145134-19145134 on Build GRCh38
6548037 c.88C>A p.L30I Chr.7: 19145275-19145275 on Build GRCh38
5000257 c.289G>T p.V97L Chr.7: 19145074-19145074 on Build GRCh38
1635226 c.234G>A p.E78E Chr.7: 19145129-19145129 on Build GRCh38
6663114 c.49G>A p.V17I Chr.7: 19145314-19145314 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144380902 Chr.7: 19145517-19145517 on Build GRCh38
rs556470561 Chr.7: 19145482-19145482 on Build GRCh38
rs772421874 Chr.7: 19145414-19145414 on Build GRCh38
rs201207258 Chr.7: 19145350-19145350 on Build GRCh38
rs17140723 Chr.7: 19145554-19145554 on Build GRCh38
rs977633636 Chr.7: 19145144-19145144 on Build GRCh38
rs267601446 Chr.7: 19145225-19145225 on Build GRCh38
rs751983847 Chr.7: 19145269-19145269 on Build GRCh38
rs578068028 Chr.7: 19145302-19145302 on Build GRCh38
rs904455045 Chr.7: 19145328-19145328 on Build GRCh38
rs939961770 Chr.7: 19145521-19145521 on Build GRCh38
rs749621995 Chr.7: 19145126-19145127 on Build GRCh38
rs758952578 Chr.7: 19145191-19145191 on Build GRCh38
rs371532641 Chr.7: 19145168-19145168 on Build GRCh38
rs149671993 Chr.7: 19145182-19145182 on Build GRCh38
rs753124786 Chr.7: 19145121-19145121 on Build GRCh38
rs774277416 Chr.7: 19145410-19145410 on Build GRCh38
rs778323274 Chr.7: 19145196-19145196 on Build GRCh38
rs139603599 Chr.7: 19145150-19145150 on Build GRCh38
rs149352998 Chr.7: 19145297-19145297 on Build GRCh38
rs758900719 Chr.7: 19145122-19145122 on Build GRCh38
rs977058081 Chr.7: 19145525-19145525 on Build GRCh38
rs138944204 Chr.7: 19145124-19145125 on Build GRCh38
rs769635262 Chr.7: 19145135-19145135 on Build GRCh38
rs749513486 Chr.7: 19145398-19145398 on Build GRCh38
rs1054944807 Chr.7: 19145547-19145547 on Build GRCh38
rs576241857 Chr.7: 19145431-19145431 on Build GRCh38
rs768552326 Chr.7: 19145229-19145229 on Build GRCh38
rs770587723 Chr.7: 19145131-19145131 on Build GRCh38
rs755824880 Chr.7: 19145337-19145337 on Build GRCh38
rs538301661 Chr.7: 19145255-19145255 on Build GRCh38
rs531963091 Chr.7: 19145543-19145543 on Build GRCh38
rs933723984 Chr.7: 19145474-19145474 on Build GRCh38
rs376088902 Chr.7: 19145527-19145527 on Build GRCh38
rs113824308 Chr.7: 19145356-19145356 on Build GRCh38
rs776846708 Chr.7: 19145247-19145247 on Build GRCh38
rs760925330 Chr.7: 19145152-19145152 on Build GRCh38
rs71017023 Chr.7: 19145128-19145129 on Build GRCh38
rs765667013 Chr.7: 19145176-19145176 on Build GRCh38
rs756884639 Chr.7: 19145206-19145206 on Build GRCh38
rs1056049781 Chr.7: 19145369-19145369 on Build GRCh38
rs145174585 Chr.7: 19145349-19145349 on Build GRCh38
rs774165450 Chr.7: 19145235-19145235 on Build GRCh38
rs959722211 Chr.7: 19145146-19145146 on Build GRCh38
rs747929132 Chr.7: 19145236-19145236 on Build GRCh38
rs771240754 Chr.7: 19145248-19145248 on Build GRCh38
rs759602135 Chr.7: 19145173-19145173 on Build GRCh38
rs767978345 Chr.7: 19145331-19145331 on Build GRCh38
rs74366495 Chr.7: 19145109-19145109 on Build GRCh38
rs764953205 Chr.7: 19145279-19145279 on Build GRCh38
rs767735592 Chr.7: 19145092-19145092 on Build GRCh38
rs770152463 Chr.7: 19145380-19145380 on Build GRCh38
rs970737177 Chr.7: 19145172-19145172 on Build GRCh38
rs17140725 Chr.7: 19145585-19145585 on Build GRCh38
rs866406481 Chr.7: 19145263-19145263 on Build GRCh38
rs762209961 Chr.7: 19145330-19145330 on Build GRCh38
rs753361746 Chr.7: 19145345-19145345 on Build GRCh38
rs762425944 Chr.7: 19145411-19145411 on Build GRCh38
rs73079402 Chr.7: 19145129-19145129 on Build GRCh38
rs978041295 Chr.7: 19145406-19145406 on Build GRCh38
rs72568438 Chr.7: 19145127-19145128 on Build GRCh38
rs745377944 Chr.7: 19145218-19145218 on Build GRCh38
rs762869329 Chr.7: 19145143-19145143 on Build GRCh38
rs926082819 Chr.7: 19145149-19145149 on Build GRCh38
rs957572564 Chr.7: 19145429-19145429 on Build GRCh38
rs986500885 Chr.7: 19145508-19145508 on Build GRCh38
rs751934968 Chr.7: 19145199-19145199 on Build GRCh38
rs894297628 Chr.7: 19145384-19145384 on Build GRCh38
rs770099494 Chr.7: 19145314-19145314 on Build GRCh38
rs752444969 Chr.7: 19145284-19145284 on Build GRCh38
rs925596158 Chr.7: 19145533-19145533 on Build GRCh38
rs775679607 Chr.7: 19145257-19145257 on Build GRCh38
rs569834973 Chr.7: 19145223-19145223 on Build GRCh38
rs931736445 Chr.7: 19145270-19145270 on Build GRCh38
rs1024314430 Chr.7: 19145586-19145586 on Build GRCh38
rs199985178 Chr.7: 19145123-19145123 on Build GRCh38
rs757836312 Chr.7: 19145329-19145329 on Build GRCh38
rs775344707 Chr.7: 19145138-19145138 on Build GRCh38
rs779493637 Chr.7: 19145363-19145363 on Build GRCh38
rs539063986 Chr.7: 19145304-19145304 on Build GRCh38
rs763583427 Chr.7: 19145258-19145258 on Build GRCh38
rs865802256 Chr.7: 19145185-19145185 on Build GRCh38
rs749275441 Chr.7: 19145224-19145224 on Build GRCh38
rs780842452 Chr.7: 19145215-19145215 on Build GRCh38
rs750152542 Chr.7: 19145289-19145289 on Build GRCh38
rs746140479 Chr.7: 19145312-19145312 on Build GRCh38
rs755704450 Chr.7: 19145290-19145290 on Build GRCh38
rs750706710 Chr.7: 19145099-19145099 on Build GRCh38
rs536561206 Chr.7: 19145110-19145110 on Build GRCh38
rs768494350 Chr.7: 19145145-19145145 on Build GRCh38
rs373401542 Chr.7: 19145440-19145440 on Build GRCh38
rs764699745 Chr.7: 19145327-19145327 on Build GRCh38
rs762682650 Chr.7: 19145091-19145091 on Build GRCh38
rs77376837 Chr.7: 19145422-19145422 on Build GRCh38
rs750726032 Chr.7: 19145332-19145332 on Build GRCh38
rs771073410 Chr.7: 19145244-19145244 on Build GRCh38
rs771944436 Chr.7: 19145348-19145348 on Build GRCh38
rs144084584 Chr.7: 19145113-19145113 on Build GRCh38
rs554438974 Chr.7: 19145132-19145132 on Build GRCh38
rs987224231 Chr.7: 19145288-19145288 on Build GRCh38
rs764644677 Chr.7: 19145260-19145260 on Build GRCh38
rs757571462 Chr.7: 19145272-19145272 on Build GRCh38
rs746730187 Chr.7: 19145378-19145378 on Build GRCh38
rs746300527 Chr.7: 19145546-19145546 on Build GRCh38
rs200268805 Chr.7: 19145390-19145390 on Build GRCh38
rs887053735 Chr.7: 19145303-19145303 on Build GRCh38
rs777653239 Chr.7: 19145377-19145377 on Build GRCh38
rs374794480 Chr.7: 19145306-19145306 on Build GRCh38
rs999975859 Chr.7: 19145117-19145117 on Build GRCh38
rs539046082 Chr.7: 19145434-19145434 on Build GRCh38
rs369136977 Chr.7: 19145324-19145324 on Build GRCh38
rs760870327 Chr.7: 19145103-19145103 on Build GRCh38
rs766581757 Chr.7: 19145161-19145161 on Build GRCh38
rs906567304 Chr.7: 19145412-19145412 on Build GRCh38
rs779426266 Chr.7: 19145119-19145119 on Build GRCh38
rs745337460 Chr.7: 19145134-19145134 on Build GRCh38
rs572618270 Chr.7: 19145357-19145357 on Build GRCh38
rs987665472 Chr.7: 19145493-19145493 on Build GRCh38
rs748759205 Chr.7: 19145296-19145296 on Build GRCh38
rs967665991 Chr.7: 19145386-19145386 on Build GRCh38
rs34966908 Chr.7: 19145123-19145124 on Build GRCh38
rs768911386 Chr.7: 19145408-19145408 on Build GRCh38
rs564664145 Chr.7: 19145541-19145541 on Build GRCh38
rs771969370 Chr.7: 19145305-19145305 on Build GRCh38
rs377664801 Chr.7: 19145250-19145250 on Build GRCh38
rs868787231 Chr.7: 19145205-19145205 on Build GRCh38
rs1025426514 Chr.7: 19145170-19145170 on Build GRCh38

View Full Product Details