Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 26193490-26193998 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNRNPA2B1
Entrez Gene ID
3181
Gene Name
heterogeneous nuclear ribonucleoprotein A2/B1
Gene Aliases
HNRNPA2, HNRNPB1, HNRPA2, HNRPA2B1, HNRPB1, IBMPFD2, RNPA2, SNRPB1
UniGene
Hs.487774
Gene Chromosome Location
Chr.7: 26189927-26200793 on Build GRCh38
Transcripts
NM_002137.3 NM_031243.2 XM_005249729.1 XM_017012109.1 XM_017012110.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1088796 c.813C>T p.Y271Y Chr.7: 26193603-26193603 on Build GRCh38
3637680 c.757G>A p.G253R Chr.7: 26193659-26193659 on Build GRCh38
5676911 c.758-2A>G p.? Chr.7: 26193696-26193696 on Build GRCh38
452900 c.769G>T p.G257* Chr.7: 26193683-26193683 on Build GRCh38
6177270 c.763A>T p.N255Y Chr.7: 26193689-26193689 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs914768834 Chr.7: 26193790-26193790 on Build GRCh38
rs560386975 Chr.7: 26193842-26193842 on Build GRCh38
rs546944823 Chr.7: 26193833-26193833 on Build GRCh38
rs748970314 Chr.7: 26193846-26193846 on Build GRCh38
rs367958673 Chr.7: 26193586-26193586 on Build GRCh38
rs887322400 Chr.7: 26193911-26193911 on Build GRCh38
rs911752782 Chr.7: 26193518-26193518 on Build GRCh38
rs763653211 Chr.7: 26193562-26193562 on Build GRCh38
rs753907415 Chr.7: 26193685-26193685 on Build GRCh38
rs757011566 Chr.7: 26193705-26193705 on Build GRCh38
rs928784293 Chr.7: 26193524-26193524 on Build GRCh38
rs374010523 Chr.7: 26193788-26193788 on Build GRCh38
rs755028208 Chr.7: 26193687-26193687 on Build GRCh38
rs758179103 Chr.7: 26193709-26193709 on Build GRCh38
rs907405573 Chr.7: 26193752-26193752 on Build GRCh38
rs767834377 Chr.7: 26193644-26193646 on Build GRCh38
rs747054718 Chr.7: 26193909-26193909 on Build GRCh38
rs76558212 Chr.7: 26193971-26193971 on Build GRCh38
rs776163204 Chr.7: 26193606-26193606 on Build GRCh38
rs921738357 Chr.7: 26193825-26193825 on Build GRCh38
rs1005073257 Chr.7: 26193801-26193801 on Build GRCh38
rs748997181 Chr.7: 26193740-26193740 on Build GRCh38
rs371101422 Chr.7: 26193715-26193715 on Build GRCh38
rs781001460 Chr.7: 26193717-26193717 on Build GRCh38
rs879250482 Chr.7: 26193819-26193819 on Build GRCh38
rs951099598 Chr.7: 26193755-26193755 on Build GRCh38
rs995805049 Chr.7: 26193878-26193878 on Build GRCh38
rs533907485 Chr.7: 26193634-26193634 on Build GRCh38
rs141885504 Chr.7: 26193672-26193672 on Build GRCh38
rs372103090 Chr.7: 26193678-26193678 on Build GRCh38
rs777686952 Chr.7: 26193591-26193591 on Build GRCh38
rs946062120 Chr.7: 26193868-26193868 on Build GRCh38
rs1018539468 Chr.7: 26193958-26193958 on Build GRCh38
rs377457065 Chr.7: 26193756-26193756 on Build GRCh38
rs931595761 Chr.7: 26193901-26193901 on Build GRCh38
rs889199852 Chr.7: 26193856-26193856 on Build GRCh38
rs768067916 Chr.7: 26193544-26193544 on Build GRCh38
rs548947084 Chr.7: 26193867-26193867 on Build GRCh38
rs981817271 Chr.7: 26193875-26193875 on Build GRCh38
rs765436421 Chr.7: 26193704-26193704 on Build GRCh38
rs199909637 Chr.7: 26193660-26193660 on Build GRCh38
rs997984275 Chr.7: 26193607-26193607 on Build GRCh38
rs577689101 Chr.7: 26193599-26193599 on Build GRCh38
rs774914044 Chr.7: 26193633-26193635 on Build GRCh38
rs376850826 Chr.7: 26193564-26193564 on Build GRCh38
rs375157988 Chr.7: 26193671-26193671 on Build GRCh38
rs762262068 Chr.7: 26193666-26193666 on Build GRCh38
rs368483139 Chr.7: 26193673-26193673 on Build GRCh38
rs575580732 Chr.7: 26193764-26193764 on Build GRCh38
rs769252415 Chr.7: 26193729-26193729 on Build GRCh38
rs914715188 Chr.7: 26193923-26193923 on Build GRCh38
rs751035830 Chr.7: 26193552-26193552 on Build GRCh38
rs1055935184 Chr.7: 26193765-26193765 on Build GRCh38
rs764968841 Chr.7: 26193711-26193711 on Build GRCh38
rs750237493 Chr.7: 26193728-26193728 on Build GRCh38
rs780330366 Chr.7: 26193554-26193554 on Build GRCh38
rs1055939327 Chr.7: 26193559-26193559 on Build GRCh38
rs770392533 Chr.7: 26193601-26193601 on Build GRCh38
rs755143390 Chr.7: 26193953-26193953 on Build GRCh38
rs990067815 Chr.7: 26193854-26193854 on Build GRCh38
rs116376035 Chr.7: 26193888-26193888 on Build GRCh38
rs544483738 Chr.7: 26193782-26193782 on Build GRCh38
rs1004554080 Chr.7: 26193946-26193946 on Build GRCh38
rs751899839 Chr.7: 26193542-26193542 on Build GRCh38
rs144309126 Chr.7: 26193569-26193569 on Build GRCh38
rs981065957 Chr.7: 26193894-26193894 on Build GRCh38
rs1047302178 Chr.7: 26193902-26193902 on Build GRCh38
rs995745544 Chr.7: 26193770-26193770 on Build GRCh38
rs1011873350 Chr.7: 26193577-26193577 on Build GRCh38
rs375449398 Chr.7: 26193741-26193741 on Build GRCh38
rs745389971 Chr.7: 26193625-26193625 on Build GRCh38
rs533171854 Chr.7: 26193817-26193817 on Build GRCh38
rs775193659 Chr.7: 26193739-26193739 on Build GRCh38
rs947615443 Chr.7: 26193798-26193798 on Build GRCh38
rs189453698 Chr.7: 26193848-26193848 on Build GRCh38
rs898767721 Chr.7: 26193767-26193767 on Build GRCh38
rs1051797158 Chr.7: 26193855-26193855 on Build GRCh38
rs745306574 Chr.7: 26193721-26193721 on Build GRCh38
rs780043993 Chr.7: 26193887-26193887 on Build GRCh38
rs573649692 Chr.7: 26193716-26193716 on Build GRCh38
rs753977811 Chr.7: 26193555-26193555 on Build GRCh38
rs898651674 Chr.7: 26193889-26193889 on Build GRCh38
rs1054874872 Chr.7: 26193695-26193695 on Build GRCh38
rs111827134 Chr.7: 26193966-26193966 on Build GRCh38
rs756250163 Chr.7: 26193553-26193553 on Build GRCh38
rs762095626 Chr.7: 26193543-26193543 on Build GRCh38
rs762284994 Chr.7: 26193568-26193570 on Build GRCh38
rs962586145 Chr.7: 26193872-26193872 on Build GRCh38
rs112962868 Chr.7: 26193573-26193573 on Build GRCh38
rs564175935 Chr.7: 26193800-26193800 on Build GRCh38
rs181836797 Chr.7: 26193899-26193899 on Build GRCh38
rs561875273 Chr.7: 26193784-26193784 on Build GRCh38
rs768495573 Chr.7: 26193885-26193885 on Build GRCh38
rs777597130 Chr.7: 26193710-26193710 on Build GRCh38
rs974564918 Chr.7: 26193822-26193822 on Build GRCh38
rs769315163 Chr.7: 26193549-26193550 on Build GRCh38
rs186651396 Chr.7: 26193747-26193747 on Build GRCh38
rs112685785 Chr.7: 26193697-26193697 on Build GRCh38

View Full Product Details