Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 157683028-157683573 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCRL3
Entrez Gene ID
115352
Gene Name
Fc receptor like 3
Gene Aliases
CD307c, FCRH3, IFGP3, IRTA3, SPAP2
UniGene
Hs.292449
Gene Chromosome Location
Chr.1: 157676481-157700985 on Build GRCh38
Transcripts
NM_001320333.1 NM_052939.3 NR_135214.1 NR_135215.1 NR_135216.1 NR_135217.1 XM_006711145.1

SNP Detail

SNP ID Chromosome Location
rs990580053 Chr.1: 157683437-157683437 on Build GRCh38
rs978055480 Chr.1: 157683496-157683496 on Build GRCh38
rs144059362 Chr.1: 157683291-157683291 on Build GRCh38
rs143094480 Chr.1: 157683246-157683246 on Build GRCh38
rs139081042 Chr.1: 157683244-157683244 on Build GRCh38
rs1038708517 Chr.1: 157683312-157683313 on Build GRCh38
rs990866284 Chr.1: 157683165-157683165 on Build GRCh38
rs182189448 Chr.1: 157683363-157683363 on Build GRCh38
rs898789291 Chr.1: 157683332-157683332 on Build GRCh38
rs1000504315 Chr.1: 157683381-157683381 on Build GRCh38
rs886575457 Chr.1: 157683146-157683146 on Build GRCh38
rs537981621 Chr.1: 157683162-157683162 on Build GRCh38
rs374184544 Chr.1: 157683173-157683173 on Build GRCh38
rs911830426 Chr.1: 157683258-157683261 on Build GRCh38
rs768311751 Chr.1: 157683384-157683384 on Build GRCh38
rs1012888659 Chr.1: 157683422-157683422 on Build GRCh38
rs968419281 Chr.1: 157683472-157683472 on Build GRCh38
rs12075547 Chr.1: 157683107-157683107 on Build GRCh38
rs917690310 Chr.1: 157683461-157683461 on Build GRCh38
rs761324132 Chr.1: 157683245-157683245 on Build GRCh38
rs571337633 Chr.1: 157683150-157683150 on Build GRCh38
rs374599000 Chr.1: 157683181-157683181 on Build GRCh38
rs754784192 Chr.1: 157683265-157683265 on Build GRCh38
rs925000082 Chr.1: 157683155-157683155 on Build GRCh38
rs966354339 Chr.1: 157683141-157683141 on Build GRCh38
rs546469462 Chr.1: 157683342-157683342 on Build GRCh38
rs67102218 Chr.1: 157683397-157683397 on Build GRCh38
rs374361037 Chr.1: 157683541-157683541 on Build GRCh38
rs370521737 Chr.1: 157683167-157683167 on Build GRCh38
rs187156318 Chr.1: 157683538-157683538 on Build GRCh38
rs201405001 Chr.1: 157683208-157683208 on Build GRCh38
rs779303583 Chr.1: 157683187-157683187 on Build GRCh38
rs539742014 Chr.1: 157683120-157683120 on Build GRCh38
rs955437418 Chr.1: 157683542-157683542 on Build GRCh38
rs989074266 Chr.1: 157683139-157683139 on Build GRCh38
rs537059644 Chr.1: 157683406-157683406 on Build GRCh38
rs528949405 Chr.1: 157683447-157683447 on Build GRCh38
rs943266808 Chr.1: 157683271-157683271 on Build GRCh38
rs368380385 Chr.1: 157683179-157683179 on Build GRCh38
rs959872600 Chr.1: 157683161-157683161 on Build GRCh38
rs77506973 Chr.1: 157683257-157683257 on Build GRCh38
rs370751094 Chr.1: 157683094-157683094 on Build GRCh38
rs113547739 Chr.1: 157683529-157683529 on Build GRCh38
rs762984348 Chr.1: 157683256-157683256 on Build GRCh38
rs1047548143 Chr.1: 157683366-157683366 on Build GRCh38
rs879223618 Chr.1: 157683163-157683163 on Build GRCh38
rs1035311517 Chr.1: 157683158-157683158 on Build GRCh38
rs141977401 Chr.1: 157683140-157683140 on Build GRCh38
rs746446767 Chr.1: 157683193-157683193 on Build GRCh38
rs576322592 Chr.1: 157683419-157683419 on Build GRCh38
rs990618229 Chr.1: 157683164-157683164 on Build GRCh38
rs770009843 Chr.1: 157683371-157683371 on Build GRCh38
rs373001148 Chr.1: 157683140-157683140 on Build GRCh38
rs781304049 Chr.1: 157683197-157683197 on Build GRCh38
rs970572203 Chr.1: 157683498-157683498 on Build GRCh38
rs373927500 Chr.1: 157683169-157683169 on Build GRCh38
rs1022296770 Chr.1: 157683426-157683426 on Build GRCh38
rs890621965 Chr.1: 157683321-157683321 on Build GRCh38
rs775962948 Chr.1: 157683205-157683205 on Build GRCh38
rs867390976 Chr.1: 157683353-157683353 on Build GRCh38
rs886382740 Chr.1: 157683192-157683192 on Build GRCh38
rs547581269 Chr.1: 157683106-157683106 on Build GRCh38
rs751276568 Chr.1: 157683168-157683168 on Build GRCh38
rs563962458 Chr.1: 157683352-157683352 on Build GRCh38
rs749893936 Chr.1: 157683248-157683248 on Build GRCh38
rs768327107 Chr.1: 157683235-157683235 on Build GRCh38

View Full Product Details