Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 42924551-42925059 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSMA2
Entrez Gene ID
5683
Gene Name
proteasome subunit alpha 2
Gene Aliases
HC3, MU, PMSA2, PSC2
UniGene
Hs.333786
Gene Chromosome Location
Chr.7: 42916861-42932206 on Build GRCh38
Transcripts
NM_002787.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7107645 c.369G>A p.Q123Q Chr.7: 42924680-42924680 on Build GRCh38
1089731 c.307G>T p.E103* Chr.7: 42924742-42924742 on Build GRCh38
32399 c.328C>G p.L110V Chr.7: 42924721-42924721 on Build GRCh38
6012676 c.252-6T>G p.? Chr.7: 42924803-42924803 on Build GRCh38
80404 c.261G>A p.V87V Chr.7: 42924788-42924788 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775538034 Chr.7: 42924751-42924751 on Build GRCh38
rs765913483 Chr.7: 42924689-42924689 on Build GRCh38
rs747406381 Chr.7: 42924715-42924715 on Build GRCh38
rs778355024 Chr.7: 42924703-42924703 on Build GRCh38
rs35856051 Chr.7: 42924804-42924805 on Build GRCh38
rs894165941 Chr.7: 42924582-42924582 on Build GRCh38
rs755740459 Chr.7: 42924669-42924669 on Build GRCh38
rs866363627 Chr.7: 42924793-42924793 on Build GRCh38
rs2303586 Chr.7: 42924624-42924624 on Build GRCh38
rs757758580 Chr.7: 42924732-42924732 on Build GRCh38
rs753417947 Chr.7: 42924690-42924690 on Build GRCh38
rs919807879 Chr.7: 42924608-42924608 on Build GRCh38
rs767232925 Chr.7: 42924665-42924665 on Build GRCh38
rs757819854 Chr.7: 42924824-42924824 on Build GRCh38
rs770113879 Chr.7: 42924750-42924750 on Build GRCh38
rs1015256446 Chr.7: 42924961-42924961 on Build GRCh38
rs200851330 Chr.7: 42924761-42924761 on Build GRCh38
rs910648761 Chr.7: 42924659-42924659 on Build GRCh38
rs577677150 Chr.7: 42924942-42924942 on Build GRCh38
rs755090116 Chr.7: 42924959-42924959 on Build GRCh38
rs904098170 Chr.7: 42924878-42924878 on Build GRCh38
rs915547339 Chr.7: 42924953-42924953 on Build GRCh38
rs780874804 Chr.7: 42925031-42925031 on Build GRCh38
rs112347813 Chr.7: 42924589-42924589 on Build GRCh38
rs746145368 Chr.7: 42924744-42924744 on Build GRCh38
rs1012634200 Chr.7: 42924587-42924587 on Build GRCh38
rs920043847 Chr.7: 42924884-42924884 on Build GRCh38
rs754395581 Chr.7: 42924694-42924694 on Build GRCh38
rs1057172395 Chr.7: 42924581-42924581 on Build GRCh38
rs112713667 Chr.7: 42924979-42924979 on Build GRCh38
rs957208188 Chr.7: 42924764-42924764 on Build GRCh38
rs878907887 Chr.7: 42924935-42924935 on Build GRCh38
rs764643823 Chr.7: 42924812-42924812 on Build GRCh38
rs761684191 Chr.7: 42924632-42924632 on Build GRCh38
rs562466961 Chr.7: 42924920-42924920 on Build GRCh38
rs760422656 Chr.7: 42924790-42924790 on Build GRCh38
rs1040454858 Chr.7: 42924698-42924698 on Build GRCh38
rs779177524 Chr.7: 42924985-42924985 on Build GRCh38
rs534124076 Chr.7: 42924604-42924604 on Build GRCh38
rs768563334 Chr.7: 42924756-42924756 on Build GRCh38
rs961201899 Chr.7: 42924968-42924968 on Build GRCh38
rs752162856 Chr.7: 42924817-42924817 on Build GRCh38
rs749412007 Chr.7: 42924754-42924754 on Build GRCh38
rs995291240 Chr.7: 42924969-42924969 on Build GRCh38
rs373907182 Chr.7: 42924874-42924874 on Build GRCh38
rs112816021 Chr.7: 42924773-42924773 on Build GRCh38
rs975432507 Chr.7: 42924883-42924883 on Build GRCh38
rs781721172 Chr.7: 42924828-42924828 on Build GRCh38
rs761735044 Chr.7: 42924769-42924769 on Build GRCh38
rs1054780499 Chr.7: 42924905-42924905 on Build GRCh38
rs766040241 Chr.7: 42924795-42924795 on Build GRCh38
rs151168516 Chr.7: 42924777-42924777 on Build GRCh38
rs781677685 Chr.7: 42924743-42924743 on Build GRCh38
rs766501758 Chr.7: 42924894-42924894 on Build GRCh38
rs754167028 Chr.7: 42924901-42924901 on Build GRCh38
rs908441601 Chr.7: 42924900-42924900 on Build GRCh38
rs760881945 Chr.7: 42924658-42924658 on Build GRCh38
rs1057050780 Chr.7: 42924924-42924924 on Build GRCh38
rs754518957 Chr.7: 42924806-42924806 on Build GRCh38
rs79709250 Chr.7: 42924668-42924668 on Build GRCh38
rs952543883 Chr.7: 42924890-42924890 on Build GRCh38
rs1031236461 Chr.7: 42924737-42924737 on Build GRCh38
rs573709469 Chr.7: 42924801-42924801 on Build GRCh38

View Full Product Details