Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 57470479-57470987 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF716
Entrez Gene ID
441234
Gene Name
zinc finger protein 716
Gene Aliases
-
UniGene
Hs.533121
Gene Chromosome Location
Chr.7: 57450177-57473559 on Build GRCh38
Transcripts
NM_001159279.1

SNP Detail

SNP ID Chromosome Location
rs201733972 Chr.7: 57470719-57470719 on Build GRCh38
rs748453241 Chr.7: 57470815-57470815 on Build GRCh38
rs782595152 Chr.7: 57470962-57470962 on Build GRCh38
rs782553910 Chr.7: 57470720-57470720 on Build GRCh38
rs185864592 Chr.7: 57470611-57470611 on Build GRCh38
rs782542537 Chr.7: 57470561-57470561 on Build GRCh38
rs4870665 Chr.7: 57470921-57470921 on Build GRCh38
rs543068955 Chr.7: 57470659-57470659 on Build GRCh38
rs200633243 Chr.7: 57470718-57470718 on Build GRCh38
rs60945087 Chr.7: 57470718-57470720 on Build GRCh38
rs10572321 Chr.7: 57470718-57470718 on Build GRCh38
rs372452787 Chr.7: 57470813-57470813 on Build GRCh38
rs80018518 Chr.7: 57470731-57470731 on Build GRCh38
rs535858758 Chr.7: 57470539-57470539 on Build GRCh38
rs555453045 Chr.7: 57470708-57470708 on Build GRCh38
rs534387325 Chr.7: 57470730-57470732 on Build GRCh38
rs553913123 Chr.7: 57470607-57470607 on Build GRCh38
rs576240020 Chr.7: 57470658-57470658 on Build GRCh38
rs138526109 Chr.7: 57470623-57470623 on Build GRCh38
rs548187704 Chr.7: 57470621-57470621 on Build GRCh38
rs369736931 Chr.7: 57470807-57470807 on Build GRCh38
rs534574843 Chr.7: 57470570-57470570 on Build GRCh38
rs573687397 Chr.7: 57470780-57470780 on Build GRCh38
rs577158775 Chr.7: 57470808-57470809 on Build GRCh38
rs559129505 Chr.7: 57470853-57470853 on Build GRCh38
rs539793173 Chr.7: 57470613-57470613 on Build GRCh38

View Full Product Details