Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 64078172-64078683 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF727
Entrez Gene ID
442319
Gene Name
zinc finger protein 727
Gene Aliases
ZNF727P
UniGene
Hs.640774
Gene Chromosome Location
Chr.7: 64044806-64085184 on Build GRCh38
Transcripts
NM_001159522.1 XM_017012225.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4830992 c.1226C>T p.S409L Chr.7: 64078275-64078275 on Build GRCh38
6012721 c.1338C>T p.H446H Chr.7: 64078387-64078387 on Build GRCh38
6198043 c.1261G>T p.V421L Chr.7: 64078310-64078310 on Build GRCh38
4451844 c.1432A>T p.R478* Chr.7: 64078481-64078481 on Build GRCh38
5632445 c.1190A>T p.K397I Chr.7: 64078239-64078239 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751118239 Chr.7: 64078468-64078468 on Build GRCh38
rs373807490 Chr.7: 64078472-64078472 on Build GRCh38
rs749517977 Chr.7: 64078398-64078398 on Build GRCh38
rs762715064 Chr.7: 64078458-64078458 on Build GRCh38
rs770098921 Chr.7: 64078260-64078260 on Build GRCh38
rs781058984 Chr.7: 64078245-64078245 on Build GRCh38
rs772956206 Chr.7: 64078452-64078452 on Build GRCh38
rs376865671 Chr.7: 64078234-64078234 on Build GRCh38
rs879240230 Chr.7: 64078219-64078219 on Build GRCh38
rs774935528 Chr.7: 64078266-64078266 on Build GRCh38
rs61734882 Chr.7: 64078400-64078400 on Build GRCh38
rs534222609 Chr.7: 64078352-64078352 on Build GRCh38
rs753868803 Chr.7: 64078201-64078201 on Build GRCh38
rs879111220 Chr.7: 64078239-64078239 on Build GRCh38
rs992042274 Chr.7: 64078602-64078602 on Build GRCh38
rs775972972 Chr.7: 64078296-64078296 on Build GRCh38
rs750261277 Chr.7: 64078424-64078424 on Build GRCh38
rs184621223 Chr.7: 64078414-64078414 on Build GRCh38
rs902039743 Chr.7: 64078342-64078342 on Build GRCh38
rs878918311 Chr.7: 64078204-64078204 on Build GRCh38
rs529969293 Chr.7: 64078483-64078483 on Build GRCh38
rs765019374 Chr.7: 64078335-64078335 on Build GRCh38
rs201927068 Chr.7: 64078292-64078294 on Build GRCh38
rs376378694 Chr.7: 64078286-64078286 on Build GRCh38
rs770960497 Chr.7: 64078399-64078399 on Build GRCh38
rs757524752 Chr.7: 64078495-64078495 on Build GRCh38
rs766014320 Chr.7: 64078334-64078334 on Build GRCh38
rs773725426 Chr.7: 64078567-64078567 on Build GRCh38
rs778254750 Chr.7: 64078206-64078206 on Build GRCh38
rs758166849 Chr.7: 64078405-64078405 on Build GRCh38
rs755842172 Chr.7: 64078476-64078476 on Build GRCh38
rs754012290 Chr.7: 64078491-64078491 on Build GRCh38
rs759624396 Chr.7: 64078593-64078593 on Build GRCh38
rs747500552 Chr.7: 64078535-64078535 on Build GRCh38
rs769089513 Chr.7: 64078541-64078541 on Build GRCh38
rs761040590 Chr.7: 64078428-64078428 on Build GRCh38
rs780080862 Chr.7: 64078232-64078232 on Build GRCh38
rs757082916 Chr.7: 64078235-64078235 on Build GRCh38
rs554580552 Chr.7: 64078293-64078293 on Build GRCh38
rs774774271 Chr.7: 64078284-64078284 on Build GRCh38
rs1036461313 Chr.7: 64078267-64078267 on Build GRCh38
rs777918107 Chr.7: 64078394-64078394 on Build GRCh38
rs749430205 Chr.7: 64078280-64078280 on Build GRCh38
rs192642994 Chr.7: 64078255-64078255 on Build GRCh38
rs997686103 Chr.7: 64078355-64078355 on Build GRCh38
rs1027927909 Chr.7: 64078242-64078242 on Build GRCh38
rs145191276 Chr.7: 64078309-64078309 on Build GRCh38
rs369383353 Chr.7: 64078510-64078510 on Build GRCh38
rs554031557 Chr.7: 64078361-64078361 on Build GRCh38
rs773280386 Chr.7: 64078275-64078275 on Build GRCh38
rs376951484 Chr.7: 64078473-64078473 on Build GRCh38
rs750021112 Chr.7: 64078210-64078210 on Build GRCh38
rs772058522 Chr.7: 64078526-64078526 on Build GRCh38
rs775703854 Chr.7: 64078415-64078415 on Build GRCh38
rs761366126 Chr.7: 64078301-64078301 on Build GRCh38
rs750111321 Chr.7: 64078310-64078310 on Build GRCh38
rs577382731 Chr.7: 64078393-64078393 on Build GRCh38
rs532360624 Chr.7: 64078611-64078611 on Build GRCh38
rs766944088 Chr.7: 64078590-64078590 on Build GRCh38
rs759031228 Chr.7: 64078573-64078573 on Build GRCh38
rs780541826 Chr.7: 64078531-64078531 on Build GRCh38
rs569132093 Chr.7: 64078227-64078227 on Build GRCh38
rs375140618 Chr.7: 64078552-64078552 on Build GRCh38
rs754988114 Chr.7: 64078358-64078358 on Build GRCh38
rs575445622 Chr.7: 64078429-64078429 on Build GRCh38
rs752641307 Chr.7: 64078594-64078594 on Build GRCh38
rs756279984 Chr.7: 64078368-64078368 on Build GRCh38
rs750178277 Chr.7: 64078366-64078366 on Build GRCh38
rs754871508 Chr.7: 64078470-64078470 on Build GRCh38
rs544216590 Chr.7: 64078459-64078459 on Build GRCh38
rs771015568 Chr.7: 64078283-64078283 on Build GRCh38
rs746017225 Chr.7: 64078518-64078518 on Build GRCh38
rs530096457 Chr.7: 64078272-64078272 on Build GRCh38
rs756277490 Chr.7: 64078619-64078619 on Build GRCh38
rs757913629 Chr.7: 64078314-64078314 on Build GRCh38
rs368318480 Chr.7: 64078553-64078553 on Build GRCh38
rs28360698 Chr.7: 64078546-64078546 on Build GRCh38
rs969441847 Chr.7: 64078633-64078633 on Build GRCh38

View Full Product Details