Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 99444691-99445233 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP5J2-PTCD1
Entrez Gene ID
100526740
Gene Name
ATP5J2-PTCD1 readthrough
Gene Aliases
-
UniGene
Hs.632313
Gene Chromosome Location
Chr.7: 99416739-99466201 on Build GRCh38
Transcripts
NM_001198879.1 NM_001081559.2 NM_001318160.1 NM_001318161.1 NM_001318162.1 NM_006693.3 XM_011515755.2 XM_011515756.2 XM_011515757.2 XM_017011700.1 XM_017011701.1 XM_017011702.1 XM_017011703.1

Gene Symbol
CPSF4
Entrez Gene ID
10898
Gene Name
cleavage and polyadenylation specific factor 4
Gene Aliases
CPSF30, NAR, NEB-1, NEB1
UniGene
Hs.489287
Gene Chromosome Location
Chr.7: 99438920-99457377 on Build GRCh38
Transcripts
NM_001198879.1 NM_001081559.2 NM_001318160.1 NM_001318161.1 NM_001318162.1 NM_006693.3 XM_011515755.2 XM_011515756.2 XM_011515757.2 XM_017011700.1 XM_017011701.1 XM_017011702.1 XM_017011703.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3883834 c.119T>C p.V40A Chr.7: 99444804-99444804 on Build GRCh38
3642871 c.129C>T p.F43F Chr.7: 99444814-99444814 on Build GRCh38
6440619 c.107C>T p.S36L Chr.7: 99444792-99444792 on Build GRCh38
748295 c.129C>G p.F43L Chr.7: 99444814-99444814 on Build GRCh38
6634884 c.111C>T p.G37G Chr.7: 99444796-99444796 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770988515 Chr.7: 99444814-99444814 on Build GRCh38
rs765093215 Chr.7: 99444915-99444915 on Build GRCh38
rs966433737 Chr.7: 99445115-99445115 on Build GRCh38
rs888659868 Chr.7: 99445158-99445158 on Build GRCh38
rs200888443 Chr.7: 99444852-99444852 on Build GRCh38
rs777549046 Chr.7: 99444880-99444880 on Build GRCh38
rs925214566 Chr.7: 99444954-99444954 on Build GRCh38
rs780601360 Chr.7: 99444767-99444767 on Build GRCh38
rs200441692 Chr.7: 99444877-99444877 on Build GRCh38
rs142674841 Chr.7: 99444811-99444811 on Build GRCh38
rs536096832 Chr.7: 99444892-99444892 on Build GRCh38
rs149573029 Chr.7: 99445069-99445069 on Build GRCh38
rs1041465488 Chr.7: 99445110-99445110 on Build GRCh38
rs778861442 Chr.7: 99444736-99444736 on Build GRCh38
rs146279758 Chr.7: 99445022-99445022 on Build GRCh38
rs945059577 Chr.7: 99444934-99444934 on Build GRCh38
rs544344658 Chr.7: 99444996-99444996 on Build GRCh38
rs531792455 Chr.7: 99445116-99445116 on Build GRCh38
rs561206674 Chr.7: 99445104-99445104 on Build GRCh38
rs997090095 Chr.7: 99444900-99444900 on Build GRCh38
rs750880958 Chr.7: 99444737-99444737 on Build GRCh38
rs769057045 Chr.7: 99444796-99444796 on Build GRCh38
rs926519443 Chr.7: 99445075-99445075 on Build GRCh38
rs141709534 Chr.7: 99444879-99444879 on Build GRCh38
rs375665139 Chr.7: 99445058-99445058 on Build GRCh38
rs56845845 Chr.7: 99445023-99445023 on Build GRCh38
rs901460226 Chr.7: 99444894-99444894 on Build GRCh38
rs1042074728 Chr.7: 99444935-99444935 on Build GRCh38
rs370789330 Chr.7: 99444739-99444739 on Build GRCh38
rs997223538 Chr.7: 99445125-99445125 on Build GRCh38
rs752081731 Chr.7: 99444865-99444865 on Build GRCh38
rs901160424 Chr.7: 99445114-99445114 on Build GRCh38
rs747779253 Chr.7: 99444878-99444878 on Build GRCh38
rs181776049 Chr.7: 99444763-99444763 on Build GRCh38
rs541709199 Chr.7: 99445127-99445127 on Build GRCh38
rs747343935 Chr.7: 99444793-99444793 on Build GRCh38
rs374208884 Chr.7: 99444860-99444860 on Build GRCh38
rs573890533 Chr.7: 99444979-99444979 on Build GRCh38
rs777931686 Chr.7: 99445139-99445139 on Build GRCh38
rs372838501 Chr.7: 99444872-99444872 on Build GRCh38
rs778623393 Chr.7: 99444785-99444786 on Build GRCh38
rs45610838 Chr.7: 99445071-99445071 on Build GRCh38
rs759412959 Chr.7: 99444832-99444832 on Build GRCh38
rs936511178 Chr.7: 99445089-99445089 on Build GRCh38
rs965584657 Chr.7: 99445107-99445107 on Build GRCh38
rs747785959 Chr.7: 99444799-99444799 on Build GRCh38
rs773406962 Chr.7: 99444888-99444889 on Build GRCh38
rs539085839 Chr.7: 99444834-99444834 on Build GRCh38
rs936588942 Chr.7: 99444965-99444965 on Build GRCh38
rs768278236 Chr.7: 99444780-99444780 on Build GRCh38
rs370696647 Chr.7: 99444861-99444861 on Build GRCh38
rs373953303 Chr.7: 99445146-99445146 on Build GRCh38
rs961755642 Chr.7: 99445046-99445046 on Build GRCh38
rs1028938062 Chr.7: 99444910-99444910 on Build GRCh38
rs545104600 Chr.7: 99445064-99445064 on Build GRCh38
rs750461703 Chr.7: 99444851-99444851 on Build GRCh38
rs550113069 Chr.7: 99444771-99444771 on Build GRCh38
rs772613412 Chr.7: 99444805-99444805 on Build GRCh38
rs949360086 Chr.7: 99445070-99445070 on Build GRCh38
rs769282679 Chr.7: 99444802-99444802 on Build GRCh38
rs1046498841 Chr.7: 99445044-99445044 on Build GRCh38
rs908930328 Chr.7: 99444728-99444728 on Build GRCh38
rs765549723 Chr.7: 99444848-99444848 on Build GRCh38
rs774475029 Chr.7: 99444819-99444819 on Build GRCh38
rs1055058399 Chr.7: 99445204-99445204 on Build GRCh38
rs374735353 Chr.7: 99444871-99444871 on Build GRCh38
rs542907283 Chr.7: 99445095-99445095 on Build GRCh38
rs201226770 Chr.7: 99444833-99444833 on Build GRCh38
rs757707645 Chr.7: 99445091-99445091 on Build GRCh38
rs772673697 Chr.7: 99444870-99444870 on Build GRCh38
rs748581621 Chr.7: 99444790-99444802 on Build GRCh38
rs748741546 Chr.7: 99444764-99444773 on Build GRCh38
rs759386832 Chr.7: 99444827-99444827 on Build GRCh38
rs555981886 Chr.7: 99444907-99444907 on Build GRCh38
rs202063300 Chr.7: 99444765-99444765 on Build GRCh38
rs936295396 Chr.7: 99445197-99445197 on Build GRCh38
rs757039577 Chr.7: 99445096-99445096 on Build GRCh38

View Full Product Details