Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 103355366-103355872 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSMC2
Entrez Gene ID
5701
Gene Name
proteasome 26S subunit, ATPase 2
Gene Aliases
MSS1, Nbla10058, S7
UniGene
Hs.437366
Gene Chromosome Location
Chr.7: 103344405-103369395 on Build GRCh38
Transcripts
NM_001204453.1 NM_002803.3 XM_005250505.1 NM_001321787.1 NM_206883.2 NM_206884.2 NM_206885.2 NR_135802.1

Gene Symbol
SLC26A5
Entrez Gene ID
375611
Gene Name
solute carrier family 26 member 5
Gene Aliases
DFNB61, PRES
UniGene
Hs.585146
Gene Chromosome Location
Chr.7: 103352598-103446177 on Build GRCh38
Transcripts
NM_001204453.1 NM_002803.3 XM_005250505.1 NM_001321787.1 NM_206883.2 NM_206884.2 NM_206885.2 NR_135802.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3662822 c.268G>T p.E90* Chr.7: 103355771-103355771 on Build GRCh38
1083569 c.212G>A p.G71D Chr.7: 103355715-103355715 on Build GRCh38
1635029 c.287C>T p.A96V Chr.7: 103355790-103355790 on Build GRCh38
326259 c.217G>T p.A73S Chr.7: 103355720-103355720 on Build GRCh38
3777969 c.264G>C p.Q88H Chr.7: 103355767-103355767 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs954949967 Chr.7: 103355486-103355486 on Build GRCh38
rs374572482 Chr.7: 103355797-103355797 on Build GRCh38
rs752538032 Chr.7: 103355522-103355522 on Build GRCh38
rs570032609 Chr.7: 103355589-103355589 on Build GRCh38
rs750732605 Chr.7: 103355710-103355710 on Build GRCh38
rs943647795 Chr.7: 103355528-103355528 on Build GRCh38
rs1034420575 Chr.7: 103355539-103355539 on Build GRCh38
rs751491617 Chr.7: 103355726-103355726 on Build GRCh38
rs566178115 Chr.7: 103355409-103355409 on Build GRCh38
rs769299088 Chr.7: 103355643-103355643 on Build GRCh38
rs765356840 Chr.7: 103355689-103355689 on Build GRCh38
rs143876338 Chr.7: 103355608-103355608 on Build GRCh38
rs1012569087 Chr.7: 103355845-103355845 on Build GRCh38
rs112392059 Chr.7: 103355467-103355467 on Build GRCh38
rs777028783 Chr.7: 103355791-103355791 on Build GRCh38
rs929517646 Chr.7: 103355567-103355567 on Build GRCh38
rs1013225411 Chr.7: 103355427-103355427 on Build GRCh38
rs185393845 Chr.7: 103355455-103355455 on Build GRCh38
rs958686465 Chr.7: 103355601-103355601 on Build GRCh38
rs372193601 Chr.7: 103355807-103355807 on Build GRCh38
rs777282950 Chr.7: 103355800-103355800 on Build GRCh38
rs548394882 Chr.7: 103355465-103355465 on Build GRCh38
rs762280906 Chr.7: 103355681-103355681 on Build GRCh38
rs1040184224 Chr.7: 103355404-103355404 on Build GRCh38
rs775905068 Chr.7: 103355669-103355669 on Build GRCh38
rs577267093 Chr.7: 103355806-103355806 on Build GRCh38
rs74998067 Chr.7: 103355773-103355773 on Build GRCh38
rs780287209 Chr.7: 103355716-103355716 on Build GRCh38
rs536929747 Chr.7: 103355452-103355452 on Build GRCh38
rs776502618 Chr.7: 103355580-103355582 on Build GRCh38
rs878927065 Chr.7: 103355714-103355714 on Build GRCh38
rs1047879815 Chr.7: 103355582-103355582 on Build GRCh38
rs987496955 Chr.7: 103355491-103355491 on Build GRCh38
rs765701551 Chr.7: 103355844-103355844 on Build GRCh38
rs764475256 Chr.7: 103355678-103355678 on Build GRCh38
rs761187255 Chr.7: 103355675-103355675 on Build GRCh38
rs781229277 Chr.7: 103355738-103355738 on Build GRCh38
rs894396094 Chr.7: 103355402-103355402 on Build GRCh38
rs377307873 Chr.7: 103355801-103355801 on Build GRCh38
rs974093499 Chr.7: 103355544-103355544 on Build GRCh38
rs201584289 Chr.7: 103355731-103355731 on Build GRCh38
rs907413866 Chr.7: 103355493-103355493 on Build GRCh38
rs374887564 Chr.7: 103355817-103355817 on Build GRCh38
rs770490547 Chr.7: 103355651-103355651 on Build GRCh38
rs758549858 Chr.7: 103355715-103355715 on Build GRCh38
rs762071287 Chr.7: 103355827-103355827 on Build GRCh38
rs762434450 Chr.7: 103355650-103355650 on Build GRCh38
rs920828272 Chr.7: 103355558-103355558 on Build GRCh38
rs771149027 Chr.7: 103355644-103355644 on Build GRCh38
rs769017116 Chr.7: 103355808-103355808 on Build GRCh38

View Full Product Details