Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 152135607-152136114 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KMT2C
Entrez Gene ID
58508
Gene Name
lysine methyltransferase 2C
Gene Aliases
HALR, MLL3
UniGene
Hs.647120
Gene Chromosome Location
Chr.7: 152134925-152436642 on Build GRCh38
Transcripts
NM_170606.2 XM_005250025.4 XM_005250026.3 XM_005250027.4 XM_005250028.4 XM_005250031.4 XM_006716077.3 XM_006716078.3 XM_006716079.3 XM_011516450.2 XM_011516451.2 XM_011516452.2 XM_011516453.2 XM_011516454.2 XM_011516456.2 XM_017012480.1 XM_017012481.1 XM_017012482.1 XM_017012483.1 XM_017012484.1 XM_017012485.1 XM_017012486.1 XM_017012487.1 XM_017012488.1 XM_017012489.1 XM_017012490.1

SNP Detail

SNP ID Chromosome Location
rs947539750 Chr.7: 152135747-152135747 on Build GRCh38
rs1053548446 Chr.7: 152135723-152135723 on Build GRCh38
rs781749494 Chr.7: 152135825-152135825 on Build GRCh38
rs34337634 Chr.7: 152136090-152136091 on Build GRCh38
rs1016308495 Chr.7: 152135875-152135875 on Build GRCh38
rs776658382 Chr.7: 152136065-152136065 on Build GRCh38
rs987147144 Chr.7: 152135636-152135636 on Build GRCh38
rs923603245 Chr.7: 152135981-152135981 on Build GRCh38
rs191408875 Chr.7: 152136023-152136023 on Build GRCh38
rs909323238 Chr.7: 152135845-152135845 on Build GRCh38
rs893274910 Chr.7: 152135729-152135729 on Build GRCh38
rs1034022351 Chr.7: 152135992-152135992 on Build GRCh38
rs544479179 Chr.7: 152135964-152135964 on Build GRCh38
rs1010099183 Chr.7: 152135899-152135899 on Build GRCh38
rs914992192 Chr.7: 152136027-152136027 on Build GRCh38
rs762300434 Chr.7: 152135960-152135960 on Build GRCh38
rs926946197 Chr.7: 152135736-152135736 on Build GRCh38
rs947821307 Chr.7: 152136045-152136045 on Build GRCh38
rs115150706 Chr.7: 152136068-152136068 on Build GRCh38
rs139097987 Chr.7: 152135876-152135876 on Build GRCh38
rs992782079 Chr.7: 152135811-152135811 on Build GRCh38
rs757839907 Chr.7: 152135818-152135818 on Build GRCh38
rs368122505 Chr.7: 152135831-152135831 on Build GRCh38
rs886458535 Chr.7: 152135838-152135838 on Build GRCh38
rs555243884 Chr.7: 152135821-152135821 on Build GRCh38
rs938317852 Chr.7: 152135756-152135756 on Build GRCh38
rs756148754 Chr.7: 152135971-152135971 on Build GRCh38
rs751922863 Chr.7: 152135875-152135875 on Build GRCh38
rs989190662 Chr.7: 152135995-152135995 on Build GRCh38
rs935046008 Chr.7: 152135715-152135715 on Build GRCh38
rs542548967 Chr.7: 152136093-152136093 on Build GRCh38
rs891654566 Chr.7: 152135888-152135888 on Build GRCh38
rs566168122 Chr.7: 152135706-152135706 on Build GRCh38
rs557984122 Chr.7: 152135856-152135856 on Build GRCh38
rs556980905 Chr.7: 152136026-152136026 on Build GRCh38
rs1003567644 Chr.7: 152135794-152135794 on Build GRCh38
rs906050524 Chr.7: 152136087-152136087 on Build GRCh38
rs1039066929 Chr.7: 152135866-152135866 on Build GRCh38
rs570014017 Chr.7: 152135867-152135867 on Build GRCh38
rs535188953 Chr.7: 152135789-152135789 on Build GRCh38
rs954635690 Chr.7: 152135896-152135896 on Build GRCh38
rs1021457035 Chr.7: 152135919-152135919 on Build GRCh38
rs918093436 Chr.7: 152135812-152135812 on Build GRCh38
rs768505538 Chr.7: 152135777-152135777 on Build GRCh38
rs992437254 Chr.7: 152136085-152136085 on Build GRCh38
rs573794942 Chr.7: 152135874-152135874 on Build GRCh38
rs773818306 Chr.7: 152135940-152135940 on Build GRCh38
rs968440874 Chr.7: 152135634-152135634 on Build GRCh38
rs549680792 Chr.7: 152135693-152135693 on Build GRCh38
rs1021363709 Chr.7: 152135633-152135633 on Build GRCh38

View Full Product Details