Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 155084071-155084561 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HTR5A
Entrez Gene ID
3361
Gene Name
5-hydroxytryptamine receptor 5A
Gene Aliases
5-HT5A
UniGene
Hs.65791
Gene Chromosome Location
Chr.7: 155070324-155087392 on Build GRCh38
Transcripts
NM_024012.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3636667 c.882C>T p.F294F Chr.7: 155084295-155084295 on Build GRCh38
7371652 c.1009A>T p.I337F Chr.7: 155084422-155084422 on Build GRCh38
3636669 c.925C>T p.P309S Chr.7: 155084338-155084338 on Build GRCh38
3636671 c.948C>T p.P316P Chr.7: 155084361-155084361 on Build GRCh38
5721130 c.747G>A p.K249K Chr.7: 155084160-155084160 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750273210 Chr.7: 155084146-155084146 on Build GRCh38
rs753686247 Chr.7: 155084263-155084263 on Build GRCh38
rs577792110 Chr.7: 155084374-155084374 on Build GRCh38
rs774229710 Chr.7: 155084522-155084522 on Build GRCh38
rs750349786 Chr.7: 155084214-155084214 on Build GRCh38
rs773870920 Chr.7: 155084254-155084254 on Build GRCh38
rs377046718 Chr.7: 155084521-155084521 on Build GRCh38
rs750571143 Chr.7: 155084349-155084349 on Build GRCh38
rs775484626 Chr.7: 155084503-155084503 on Build GRCh38
rs759634832 Chr.7: 155084531-155084531 on Build GRCh38
rs113935396 Chr.7: 155084262-155084262 on Build GRCh38
rs780600560 Chr.7: 155084466-155084466 on Build GRCh38
rs990870904 Chr.7: 155084517-155084517 on Build GRCh38
rs755467797 Chr.7: 155084444-155084444 on Build GRCh38
rs995870942 Chr.7: 155084167-155084167 on Build GRCh38
rs556407933 Chr.7: 155084361-155084361 on Build GRCh38
rs199543736 Chr.7: 155084124-155084124 on Build GRCh38
rs753593458 Chr.7: 155084230-155084230 on Build GRCh38
rs746576122 Chr.7: 155084392-155084392 on Build GRCh38
rs776018823 Chr.7: 155084244-155084244 on Build GRCh38
rs764943544 Chr.7: 155084338-155084338 on Build GRCh38
rs755900282 Chr.7: 155084161-155084161 on Build GRCh38
rs576598432 Chr.7: 155084276-155084276 on Build GRCh38
rs376821448 Chr.7: 155084253-155084253 on Build GRCh38
rs770639507 Chr.7: 155084337-155084337 on Build GRCh38
rs761345698 Chr.7: 155084239-155084239 on Build GRCh38
rs868590290 Chr.7: 155084388-155084388 on Build GRCh38
rs747763482 Chr.7: 155084313-155084313 on Build GRCh38
rs776253608 Chr.7: 155084344-155084345 on Build GRCh38
rs202028626 Chr.7: 155084258-155084258 on Build GRCh38
rs748385154 Chr.7: 155084173-155084173 on Build GRCh38
rs766169807 Chr.7: 155084229-155084229 on Build GRCh38
rs745821144 Chr.7: 155084417-155084417 on Build GRCh38
rs745353794 Chr.7: 155084483-155084483 on Build GRCh38
rs758551610 Chr.7: 155084177-155084177 on Build GRCh38
rs779179069 Chr.7: 155084364-155084364 on Build GRCh38
rs186966602 Chr.7: 155084343-155084343 on Build GRCh38
rs367704188 Chr.7: 155084257-155084257 on Build GRCh38
rs370190833 Chr.7: 155084122-155084122 on Build GRCh38
rs779166177 Chr.7: 155084172-155084172 on Build GRCh38
rs755448249 Chr.7: 155084363-155084363 on Build GRCh38
rs572459769 Chr.7: 155084235-155084235 on Build GRCh38
rs150067714 Chr.7: 155084203-155084203 on Build GRCh38
rs202114711 Chr.7: 155084340-155084340 on Build GRCh38
rs759457068 Chr.7: 155084332-155084332 on Build GRCh38
rs758761162 Chr.7: 155084455-155084455 on Build GRCh38
rs756856532 Chr.7: 155084461-155084461 on Build GRCh38
rs138756848 Chr.7: 155084309-155084309 on Build GRCh38
rs775897660 Chr.7: 155084512-155084512 on Build GRCh38
rs748499187 Chr.7: 155084366-155084366 on Build GRCh38
rs771718011 Chr.7: 155084319-155084319 on Build GRCh38
rs538768507 Chr.7: 155084209-155084209 on Build GRCh38
rs760998533 Chr.7: 155084529-155084529 on Build GRCh38
rs772768835 Chr.7: 155084322-155084322 on Build GRCh38
rs769720876 Chr.7: 155084419-155084419 on Build GRCh38
rs754315935 Chr.7: 155084362-155084362 on Build GRCh38
rs764432346 Chr.7: 155084270-155084270 on Build GRCh38
rs140437792 Chr.7: 155084292-155084292 on Build GRCh38
rs746782576 Chr.7: 155084287-155084287 on Build GRCh38
rs1009027342 Chr.7: 155084119-155084119 on Build GRCh38
rs776610389 Chr.7: 155084530-155084530 on Build GRCh38
rs777944194 Chr.7: 155084187-155084187 on Build GRCh38
rs769282243 Chr.7: 155084506-155084506 on Build GRCh38
rs770437433 Chr.7: 155084404-155084404 on Build GRCh38
rs749161920 Chr.7: 155084519-155084519 on Build GRCh38
rs756258628 Chr.7: 155084360-155084360 on Build GRCh38
rs753047638 Chr.7: 155084454-155084454 on Build GRCh38
rs757241496 Chr.7: 155084144-155084144 on Build GRCh38
rs776050067 Chr.7: 155084409-155084409 on Build GRCh38
rs1031605672 Chr.7: 155084198-155084198 on Build GRCh38
rs771505360 Chr.7: 155084193-155084193 on Build GRCh38
rs745395980 Chr.7: 155084492-155084493 on Build GRCh38
rs995856401 Chr.7: 155084496-155084496 on Build GRCh38
rs767334665 Chr.7: 155084145-155084145 on Build GRCh38
rs559915059 Chr.7: 155084099-155084099 on Build GRCh38
rs114114647 Chr.7: 155084118-155084118 on Build GRCh38
rs750481084 Chr.7: 155084290-155084290 on Build GRCh38
rs769669704 Chr.7: 155084501-155084501 on Build GRCh38
rs1044277747 Chr.7: 155084168-155084168 on Build GRCh38
rs760159283 Chr.7: 155084326-155084326 on Build GRCh38
rs112433847 Chr.7: 155084140-155084140 on Build GRCh38
rs767824283 Chr.7: 155084348-155084348 on Build GRCh38
rs765740227 Chr.7: 155084445-155084445 on Build GRCh38
rs776532195 Chr.7: 155084329-155084329 on Build GRCh38
rs140450324 Chr.7: 155084325-155084325 on Build GRCh38
rs151277627 Chr.7: 155084289-155084289 on Build GRCh38
rs368982359 Chr.7: 155084125-155084125 on Build GRCh38
rs771306601 Chr.7: 155084117-155084117 on Build GRCh38
rs149191890 Chr.7: 155084208-155084208 on Build GRCh38
rs778049475 Chr.7: 155084456-155084456 on Build GRCh38
rs770383539 Chr.7: 155084240-155084240 on Build GRCh38
rs145572673 Chr.7: 155084460-155084460 on Build GRCh38
rs777378523 Chr.7: 155084375-155084375 on Build GRCh38
rs750993537 Chr.7: 155084137-155084137 on Build GRCh38
rs766540353 Chr.7: 155084130-155084130 on Build GRCh38
rs763663863 Chr.7: 155084128-155084128 on Build GRCh38
rs373904053 Chr.7: 155084430-155084430 on Build GRCh38
rs757266979 Chr.7: 155084236-155084236 on Build GRCh38
rs376598745 Chr.7: 155084328-155084328 on Build GRCh38
rs776996961 Chr.7: 155084196-155084196 on Build GRCh38
rs768890468 Chr.7: 155084305-155084305 on Build GRCh38
rs747015978 Chr.7: 155084192-155084192 on Build GRCh38
rs145552975 Chr.7: 155084202-155084202 on Build GRCh38
rs773969198 Chr.7: 155084234-155084234 on Build GRCh38
rs755990394 Chr.7: 155084226-155084226 on Build GRCh38
rs775196416 Chr.7: 155084426-155084426 on Build GRCh38
rs773555483 Chr.7: 155084429-155084429 on Build GRCh38
rs900278431 Chr.7: 155084355-155084355 on Build GRCh38
rs770293896 Chr.7: 155084201-155084201 on Build GRCh38
rs900184640 Chr.7: 155084440-155084440 on Build GRCh38
rs758673974 Chr.7: 155084231-155084231 on Build GRCh38
rs753903465 Chr.7: 155084431-155084431 on Build GRCh38
rs371458123 Chr.7: 155084296-155084296 on Build GRCh38
rs746290830 Chr.7: 155084197-155084197 on Build GRCh38
rs145653373 Chr.7: 155084295-155084295 on Build GRCh38

View Full Product Details