Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161076266-161076768 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NECTIN4
Entrez Gene ID
81607
Gene Name
nectin cell adhesion molecule 4
Gene Aliases
EDSS1, LNIR, PRR4, PVRL4, nectin-4
UniGene
Hs.492490
Gene Chromosome Location
Chr.1: 161070991-161089595 on Build GRCh38
Transcripts
NM_030916.2 XM_005245508.3 XM_011510021.2 XM_011510022.2 XM_011510023.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4024304 c.850C>T p.R284W Chr.1: 161076356-161076356 on Build GRCh38
3477156 c.772C>T p.L258L Chr.1: 161076434-161076434 on Build GRCh38
6760513 c.731-1G>A p.? Chr.1: 161076476-161076476 on Build GRCh38
5767584 c.851+8C>T p.? Chr.1: 161076347-161076347 on Build GRCh38
5583984 c.743C>T p.A248V Chr.1: 161076463-161076463 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs938205653 Chr.1: 161076626-161076647 on Build GRCh38
rs201593617 Chr.1: 161076345-161076345 on Build GRCh38
rs766114872 Chr.1: 161076482-161076482 on Build GRCh38
rs972841233 Chr.1: 161076384-161076384 on Build GRCh38
rs764630945 Chr.1: 161076651-161076651 on Build GRCh38
rs199581159 Chr.1: 161076412-161076412 on Build GRCh38
rs758862899 Chr.1: 161076306-161076306 on Build GRCh38
rs376208610 Chr.1: 161076519-161076519 on Build GRCh38
rs772998747 Chr.1: 161076383-161076383 on Build GRCh38
rs902402754 Chr.1: 161076529-161076529 on Build GRCh38
rs995094652 Chr.1: 161076741-161076741 on Build GRCh38
rs961044807 Chr.1: 161076348-161076348 on Build GRCh38
rs772302133 Chr.1: 161076407-161076407 on Build GRCh38
rs778073888 Chr.1: 161076437-161076437 on Build GRCh38
rs111398053 Chr.1: 161076525-161076525 on Build GRCh38
rs550295998 Chr.1: 161076647-161076647 on Build GRCh38
rs1012755009 Chr.1: 161076629-161076629 on Build GRCh38
rs776577224 Chr.1: 161076425-161076425 on Build GRCh38
rs375484596 Chr.1: 161076356-161076356 on Build GRCh38
rs376047872 Chr.1: 161076344-161076344 on Build GRCh38
rs1009093097 Chr.1: 161076553-161076553 on Build GRCh38
rs532581380 Chr.1: 161076399-161076399 on Build GRCh38
rs1054235728 Chr.1: 161076592-161076592 on Build GRCh38
rs371803246 Chr.1: 161076322-161076322 on Build GRCh38
rs141330389 Chr.1: 161076569-161076569 on Build GRCh38
rs758176682 Chr.1: 161076334-161076334 on Build GRCh38
rs998578491 Chr.1: 161076395-161076395 on Build GRCh38
rs781120252 Chr.1: 161076495-161076495 on Build GRCh38
rs750621376 Chr.1: 161076443-161076443 on Build GRCh38
rs983559291 Chr.1: 161076700-161076700 on Build GRCh38
rs950839160 Chr.1: 161076649-161076649 on Build GRCh38
rs754882119 Chr.1: 161076488-161076488 on Build GRCh38
rs368147524 Chr.1: 161076664-161076664 on Build GRCh38
rs117323434 Chr.1: 161076325-161076325 on Build GRCh38
rs564639302 Chr.1: 161076601-161076601 on Build GRCh38
rs765500135 Chr.1: 161076438-161076438 on Build GRCh38
rs183203372 Chr.1: 161076607-161076607 on Build GRCh38
rs187839290 Chr.1: 161076727-161076727 on Build GRCh38
rs943812162 Chr.1: 161076471-161076471 on Build GRCh38
rs769578135 Chr.1: 161076375-161076375 on Build GRCh38
rs931092593 Chr.1: 161076424-161076424 on Build GRCh38
rs747731302 Chr.1: 161076504-161076504 on Build GRCh38
rs267606991 Chr.1: 161076355-161076355 on Build GRCh38
rs962295926 Chr.1: 161076737-161076737 on Build GRCh38
rs751316786 Chr.1: 161076483-161076483 on Build GRCh38
rs374676117 Chr.1: 161076408-161076408 on Build GRCh38
rs145417930 Chr.1: 161076522-161076522 on Build GRCh38
rs145688995 Chr.1: 161076428-161076428 on Build GRCh38
rs1003639526 Chr.1: 161076674-161076674 on Build GRCh38
rs764614701 Chr.1: 161076415-161076415 on Build GRCh38
rs911064660 Chr.1: 161076464-161076464 on Build GRCh38
rs1012330017 Chr.1: 161076302-161076302 on Build GRCh38
rs866769481 Chr.1: 161076554-161076554 on Build GRCh38
rs1024258977 Chr.1: 161076305-161076305 on Build GRCh38
rs935222072 Chr.1: 161076553-161076553 on Build GRCh38
rs758225856 Chr.1: 161076449-161076449 on Build GRCh38
rs995379880 Chr.1: 161076633-161076633 on Build GRCh38

View Full Product Details