Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35101813-35102322 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STOML2
Entrez Gene ID
30968
Gene Name
stomatin like 2
Gene Aliases
HSPC108, SLP-2
UniGene
Hs.3439
Gene Chromosome Location
Chr.9: 35099776-35103195 on Build GRCh38
Transcripts
NM_001287031.1 NM_001287032.1 NM_001287033.1 NM_013442.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3907094 c.323G>A p.R108H Chr.9: 35101923-35101923 on Build GRCh38
6792189 c.232_233delAG p.L79fs*9 Chr.9: 35102145-35102146 on Build GRCh38
4972038 c.216_220delGATCC p.R74fs*13 Chr.9: 35102158-35102162 on Build GRCh38
4618857 c.245T>C p.I82T Chr.9: 35102133-35102133 on Build GRCh38
1108611 c.272C>T p.A91V Chr.9: 35102106-35102106 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764493595 Chr.9: 35102245-35102246 on Build GRCh38
rs763766258 Chr.9: 35102185-35102185 on Build GRCh38
rs374440519 Chr.9: 35101977-35101977 on Build GRCh38
rs532967341 Chr.9: 35102123-35102123 on Build GRCh38
rs374435906 Chr.9: 35101845-35101845 on Build GRCh38
rs749144448 Chr.9: 35101905-35101905 on Build GRCh38
rs866231774 Chr.9: 35101902-35101902 on Build GRCh38
rs531585494 Chr.9: 35102267-35102267 on Build GRCh38
rs763483882 Chr.9: 35102163-35102163 on Build GRCh38
rs979840018 Chr.9: 35102120-35102120 on Build GRCh38
rs772219629 Chr.9: 35101854-35101854 on Build GRCh38
rs34203068 Chr.9: 35102129-35102129 on Build GRCh38
rs942918065 Chr.9: 35102293-35102293 on Build GRCh38
rs113748657 Chr.9: 35101964-35101964 on Build GRCh38
rs11548407 Chr.9: 35101912-35101912 on Build GRCh38
rs374968568 Chr.9: 35101907-35101907 on Build GRCh38
rs368433324 Chr.9: 35102016-35102016 on Build GRCh38
rs76112676 Chr.9: 35102252-35102252 on Build GRCh38
rs888518578 Chr.9: 35101981-35101981 on Build GRCh38
rs371699957 Chr.9: 35102192-35102192 on Build GRCh38
rs571038997 Chr.9: 35101878-35101878 on Build GRCh38
rs768863648 Chr.9: 35102111-35102111 on Build GRCh38
rs752186895 Chr.9: 35102012-35102012 on Build GRCh38
rs755768193 Chr.9: 35101895-35101895 on Build GRCh38
rs77199230 Chr.9: 35101871-35101871 on Build GRCh38
rs367783139 Chr.9: 35101973-35101973 on Build GRCh38
rs745910436 Chr.9: 35101924-35101924 on Build GRCh38
rs761453854 Chr.9: 35101853-35101853 on Build GRCh38
rs552286803 Chr.9: 35102212-35102212 on Build GRCh38
rs779870674 Chr.9: 35102221-35102221 on Build GRCh38
rs763393232 Chr.9: 35101950-35101951 on Build GRCh38
rs897414448 Chr.9: 35101885-35101885 on Build GRCh38
rs930241266 Chr.9: 35102249-35102249 on Build GRCh38
rs796452972 Chr.9: 35101871-35101873 on Build GRCh38
rs374236012 Chr.9: 35102109-35102109 on Build GRCh38
rs956161557 Chr.9: 35102044-35102044 on Build GRCh38
rs998142177 Chr.9: 35101987-35101987 on Build GRCh38
rs750286745 Chr.9: 35102208-35102208 on Build GRCh38
rs755042355 Chr.9: 35102238-35102238 on Build GRCh38
rs774406660 Chr.9: 35101839-35101839 on Build GRCh38
rs1005042027 Chr.9: 35102194-35102194 on Build GRCh38
rs761505910 Chr.9: 35102198-35102198 on Build GRCh38
rs201279905 Chr.9: 35101879-35101879 on Build GRCh38
rs767029262 Chr.9: 35101991-35101991 on Build GRCh38
rs144693505 Chr.9: 35102165-35102165 on Build GRCh38
rs901175778 Chr.9: 35101986-35101986 on Build GRCh38
rs200654595 Chr.9: 35101866-35101866 on Build GRCh38
rs751589163 Chr.9: 35102222-35102222 on Build GRCh38
rs530749219 Chr.9: 35102191-35102191 on Build GRCh38
rs1042312558 Chr.9: 35102037-35102037 on Build GRCh38
rs759066984 Chr.9: 35101988-35101988 on Build GRCh38
rs773757788 Chr.9: 35102161-35102161 on Build GRCh38
rs750341682 Chr.9: 35102095-35102095 on Build GRCh38
rs753441091 Chr.9: 35102213-35102213 on Build GRCh38
rs113358588 Chr.9: 35102094-35102094 on Build GRCh38
rs777589644 Chr.9: 35101898-35101898 on Build GRCh38
rs757079345 Chr.9: 35102074-35102074 on Build GRCh38
rs973617565 Chr.9: 35102225-35102225 on Build GRCh38
rs764986636 Chr.9: 35102199-35102199 on Build GRCh38
rs199826401 Chr.9: 35101943-35101943 on Build GRCh38
rs771404483 Chr.9: 35102164-35102164 on Build GRCh38
rs996303075 Chr.9: 35101841-35101841 on Build GRCh38
rs777041148 Chr.9: 35101966-35101966 on Build GRCh38
rs139221530 Chr.9: 35102108-35102108 on Build GRCh38
rs1000712162 Chr.9: 35102070-35102070 on Build GRCh38
rs368707588 Chr.9: 35102083-35102083 on Build GRCh38
rs368615584 Chr.9: 35102145-35102145 on Build GRCh38
rs693325 Chr.9: 35101873-35101873 on Build GRCh38
rs775815527 Chr.9: 35101858-35101858 on Build GRCh38
rs752414148 Chr.9: 35101893-35101893 on Build GRCh38
rs760901687 Chr.9: 35101859-35101859 on Build GRCh38
rs773884667 Chr.9: 35101985-35101985 on Build GRCh38
rs763708469 Chr.9: 35102048-35102048 on Build GRCh38
rs370437732 Chr.9: 35101889-35101889 on Build GRCh38
rs563889169 Chr.9: 35102096-35102096 on Build GRCh38
rs770358893 Chr.9: 35101984-35101984 on Build GRCh38
rs371826848 Chr.9: 35102097-35102097 on Build GRCh38
rs772218681 Chr.9: 35101940-35101940 on Build GRCh38
rs377389253 Chr.9: 35101882-35101882 on Build GRCh38
rs760111495 Chr.9: 35102175-35102175 on Build GRCh38
rs200065914 Chr.9: 35102068-35102068 on Build GRCh38
rs768827289 Chr.9: 35101945-35101945 on Build GRCh38
rs775482860 Chr.9: 35101942-35101942 on Build GRCh38
rs781568932 Chr.9: 35102122-35102122 on Build GRCh38
rs548596894 Chr.9: 35102269-35102269 on Build GRCh38
rs777603377 Chr.9: 35102044-35102045 on Build GRCh38
rs781515715 Chr.9: 35102240-35102240 on Build GRCh38

View Full Product Details