Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35699099-35699606 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TLN1
Entrez Gene ID
7094
Gene Name
talin 1
Gene Aliases
ILWEQ, TLN
UniGene
Hs.471014
Gene Chromosome Location
Chr.9: 35697337-35732395 on Build GRCh38
Transcripts
NM_006289.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6115953 c.6802C>T p.Q2268* Chr.9: 35699428-35699428 on Build GRCh38
3716177 c.6862G>C p.E2288Q Chr.9: 35699368-35699368 on Build GRCh38
1108714 c.6781C>T p.P2261S Chr.9: 35699449-35699449 on Build GRCh38
3657195 c.6842C>T p.T2281I Chr.9: 35699388-35699388 on Build GRCh38
3926770 c.6836C>T p.S2279F Chr.9: 35699394-35699394 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs41313454 Chr.9: 35699549-35699549 on Build GRCh38
rs564802746 Chr.9: 35699513-35699513 on Build GRCh38
rs756816174 Chr.9: 35699167-35699170 on Build GRCh38
rs768307016 Chr.9: 35699374-35699374 on Build GRCh38
rs373132892 Chr.9: 35699205-35699205 on Build GRCh38
rs757836976 Chr.9: 35699336-35699336 on Build GRCh38
rs780140944 Chr.9: 35699156-35699156 on Build GRCh38
rs780547670 Chr.9: 35699170-35699170 on Build GRCh38
rs771447783 Chr.9: 35699121-35699121 on Build GRCh38
rs866716729 Chr.9: 35699247-35699247 on Build GRCh38
rs777162223 Chr.9: 35699230-35699230 on Build GRCh38
rs754293192 Chr.9: 35699335-35699335 on Build GRCh38
rs779678019 Chr.9: 35699143-35699143 on Build GRCh38
rs920318969 Chr.9: 35699275-35699275 on Build GRCh38
rs557591653 Chr.9: 35699236-35699236 on Build GRCh38
rs761654683 Chr.9: 35699173-35699173 on Build GRCh38
rs1041805294 Chr.9: 35699536-35699536 on Build GRCh38
rs763015066 Chr.9: 35699197-35699197 on Build GRCh38
rs991685405 Chr.9: 35699198-35699198 on Build GRCh38
rs759561962 Chr.9: 35699420-35699420 on Build GRCh38
rs749052015 Chr.9: 35699415-35699415 on Build GRCh38
rs912176258 Chr.9: 35699217-35699217 on Build GRCh38
rs751968163 Chr.9: 35699154-35699156 on Build GRCh38
rs908249637 Chr.9: 35699277-35699277 on Build GRCh38
rs764349081 Chr.9: 35699475-35699475 on Build GRCh38
rs769765240 Chr.9: 35699175-35699175 on Build GRCh38
rs11541907 Chr.9: 35699435-35699435 on Build GRCh38
rs1055602869 Chr.9: 35699305-35699305 on Build GRCh38
rs759541152 Chr.9: 35699307-35699307 on Build GRCh38
rs971722710 Chr.9: 35699564-35699564 on Build GRCh38
rs544639009 Chr.9: 35699483-35699483 on Build GRCh38
rs776457797 Chr.9: 35699167-35699167 on Build GRCh38
rs953535617 Chr.9: 35699541-35699541 on Build GRCh38
rs746459933 Chr.9: 35699349-35699349 on Build GRCh38
rs766356106 Chr.9: 35699204-35699204 on Build GRCh38
rs377341072 Chr.9: 35699330-35699330 on Build GRCh38
rs767633292 Chr.9: 35699425-35699425 on Build GRCh38
rs773118269 Chr.9: 35699407-35699407 on Build GRCh38
rs747846158 Chr.9: 35699396-35699396 on Build GRCh38
rs73438721 Chr.9: 35699406-35699406 on Build GRCh38
rs550311375 Chr.9: 35699556-35699556 on Build GRCh38
rs374711808 Chr.9: 35699237-35699237 on Build GRCh38
rs774185391 Chr.9: 35699418-35699418 on Build GRCh38
rs903230014 Chr.9: 35699538-35699538 on Build GRCh38
rs369048701 Chr.9: 35699331-35699331 on Build GRCh38
rs770836745 Chr.9: 35699416-35699416 on Build GRCh38
rs767686341 Chr.9: 35699308-35699308 on Build GRCh38
rs779299435 Chr.9: 35699348-35699348 on Build GRCh38
rs745560223 Chr.9: 35699314-35699315 on Build GRCh38
rs775657730 Chr.9: 35699458-35699458 on Build GRCh38
rs760950226 Chr.9: 35699474-35699474 on Build GRCh38
rs530832559 Chr.9: 35699572-35699572 on Build GRCh38
rs752936309 Chr.9: 35699320-35699320 on Build GRCh38
rs746625724 Chr.9: 35699154-35699154 on Build GRCh38
rs367889462 Chr.9: 35699267-35699267 on Build GRCh38
rs768362216 Chr.9: 35699158-35699158 on Build GRCh38
rs772995514 Chr.9: 35699190-35699190 on Build GRCh38
rs780974070 Chr.9: 35699392-35699392 on Build GRCh38
rs1055059387 Chr.9: 35699559-35699559 on Build GRCh38
rs757564484 Chr.9: 35699512-35699512 on Build GRCh38
rs201432200 Chr.9: 35699188-35699188 on Build GRCh38
rs561143929 Chr.9: 35699574-35699574 on Build GRCh38
rs377350733 Chr.9: 35699491-35699491 on Build GRCh38
rs962582713 Chr.9: 35699287-35699287 on Build GRCh38

View Full Product Details