Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 120604942-120605449 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MEGF9
Entrez Gene ID
1955
Gene Name
multiple EGF like domains 9
Gene Aliases
EGFL5
UniGene
Hs.744903
Gene Chromosome Location
Chr.9: 120600918-120714487 on Build GRCh38
Transcripts
NM_001080497.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1459696 c.1802G>T p.S601I Chr.9: 120605308-120605308 on Build GRCh38
3653737 c.1686G>A p.V562V Chr.9: 120605424-120605424 on Build GRCh38
4945748 c.1789A>G p.I597V Chr.9: 120605321-120605321 on Build GRCh38
396793 c.1776G>A p.L592L Chr.9: 120605334-120605334 on Build GRCh38
1257606 c.1910A>G p.Y637C Chr.9: 120605200-120605200 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775513446 Chr.9: 120605110-120605110 on Build GRCh38
rs970821555 Chr.9: 120605045-120605045 on Build GRCh38
rs377070491 Chr.9: 120605206-120605206 on Build GRCh38
rs568179279 Chr.9: 120605056-120605056 on Build GRCh38
rs767203285 Chr.9: 120605419-120605419 on Build GRCh38
rs185776336 Chr.9: 120605368-120605368 on Build GRCh38
rs771209570 Chr.9: 120605286-120605286 on Build GRCh38
rs914166767 Chr.9: 120605136-120605136 on Build GRCh38
rs566862903 Chr.9: 120605000-120605000 on Build GRCh38
rs888301484 Chr.9: 120605260-120605260 on Build GRCh38
rs368513662 Chr.9: 120605337-120605337 on Build GRCh38
rs372166478 Chr.9: 120605359-120605359 on Build GRCh38
rs370355752 Chr.9: 120605215-120605215 on Build GRCh38
rs778345020 Chr.9: 120605384-120605384 on Build GRCh38
rs756856340 Chr.9: 120605214-120605214 on Build GRCh38
rs749871238 Chr.9: 120605235-120605235 on Build GRCh38
rs779468777 Chr.9: 120605398-120605398 on Build GRCh38
rs16909942 Chr.9: 120605106-120605106 on Build GRCh38
rs534962699 Chr.9: 120605007-120605007 on Build GRCh38
rs1049484974 Chr.9: 120605220-120605220 on Build GRCh38
rs926796095 Chr.9: 120605088-120605088 on Build GRCh38
rs1037329070 Chr.9: 120605081-120605081 on Build GRCh38
rs375532572 Chr.9: 120605183-120605183 on Build GRCh38
rs766265557 Chr.9: 120605382-120605382 on Build GRCh38
rs753519832 Chr.9: 120605356-120605356 on Build GRCh38
rs749876021 Chr.9: 120605388-120605388 on Build GRCh38
rs766727535 Chr.9: 120605161-120605161 on Build GRCh38
rs764070750 Chr.9: 120605244-120605244 on Build GRCh38
rs551759248 Chr.9: 120604999-120604999 on Build GRCh38
rs151265929 Chr.9: 120604998-120604998 on Build GRCh38
rs367917074 Chr.9: 120605240-120605240 on Build GRCh38
rs972499291 Chr.9: 120605027-120605027 on Build GRCh38
rs774708360 Chr.9: 120605302-120605302 on Build GRCh38
rs771407528 Chr.9: 120605390-120605390 on Build GRCh38
rs761604118 Chr.9: 120605357-120605357 on Build GRCh38
rs374966823 Chr.9: 120605217-120605217 on Build GRCh38
rs118083394 Chr.9: 120605274-120605274 on Build GRCh38
rs56019987 Chr.9: 120605403-120605403 on Build GRCh38
rs774511906 Chr.9: 120605173-120605173 on Build GRCh38
rs961307308 Chr.9: 120605013-120605013 on Build GRCh38
rs764115864 Chr.9: 120605355-120605355 on Build GRCh38
rs375532860 Chr.9: 120605323-120605323 on Build GRCh38
rs569304313 Chr.9: 120604992-120604992 on Build GRCh38
rs982251735 Chr.9: 120605378-120605378 on Build GRCh38
rs949554753 Chr.9: 120604991-120604991 on Build GRCh38
rs751098164 Chr.9: 120605369-120605369 on Build GRCh38
rs150442642 Chr.9: 120605153-120605153 on Build GRCh38
rs1036849858 Chr.9: 120605336-120605336 on Build GRCh38
rs770360660 Chr.9: 120605251-120605251 on Build GRCh38
rs756124436 Chr.9: 120605198-120605198 on Build GRCh38
rs773662821 Chr.9: 120605145-120605145 on Build GRCh38
rs897104981 Chr.9: 120605343-120605343 on Build GRCh38
rs958423357 Chr.9: 120605099-120605099 on Build GRCh38
rs775680586 Chr.9: 120605340-120605340 on Build GRCh38
rs777949260 Chr.9: 120605265-120605265 on Build GRCh38
rs900378751 Chr.9: 120605285-120605285 on Build GRCh38
rs759691534 Chr.9: 120605176-120605176 on Build GRCh38
rs140511263 Chr.9: 120605048-120605048 on Build GRCh38
rs1005488431 Chr.9: 120604981-120604981 on Build GRCh38
rs753700161 Chr.9: 120605207-120605207 on Build GRCh38
rs988204367 Chr.9: 120605352-120605352 on Build GRCh38
rs751379482 Chr.9: 120605392-120605392 on Build GRCh38
rs556783992 Chr.9: 120605127-120605127 on Build GRCh38
rs550809995 Chr.9: 120604985-120604985 on Build GRCh38
rs772371789 Chr.9: 120605418-120605418 on Build GRCh38
rs758099186 Chr.9: 120605364-120605364 on Build GRCh38
rs779676330 Chr.9: 120605242-120605242 on Build GRCh38
rs768599041 Chr.9: 120605078-120605078 on Build GRCh38
rs772441151 Chr.9: 120605044-120605044 on Build GRCh38
rs752563123 Chr.9: 120605188-120605188 on Build GRCh38

View Full Product Details