Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 38674134-38674635 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSPAN7
Entrez Gene ID
7102
Gene Name
tetraspanin 7
Gene Aliases
A15, CCG-B7, CD231, DXS1692E, MRX58, MXS1, TALLA-1, TM4SF2, TM4SF2b
UniGene
Hs.441664
Gene Chromosome Location
Chr.X: 38561478-38688918 on Build GRCh38
Transcripts
NM_004615.3

SNP Detail

SNP ID Chromosome Location
rs763236751 Chr.X: 38674191-38674191 on Build GRCh38
rs757108282 Chr.X: 38674278-38674278 on Build GRCh38
rs745763721 Chr.X: 38674284-38674284 on Build GRCh38
rs954512701 Chr.X: 38674367-38674367 on Build GRCh38
rs17851593 Chr.X: 38674254-38674254 on Build GRCh38
rs998805598 Chr.X: 38674301-38674301 on Build GRCh38
rs758763402 Chr.X: 38674394-38674394 on Build GRCh38
rs138427413 Chr.X: 38674270-38674270 on Build GRCh38
rs112022631 Chr.X: 38674319-38674319 on Build GRCh38
rs919084545 Chr.X: 38674397-38674397 on Build GRCh38
rs771261230 Chr.X: 38674340-38674340 on Build GRCh38
rs973238571 Chr.X: 38674416-38674416 on Build GRCh38
rs180682082 Chr.X: 38674166-38674166 on Build GRCh38
rs942984370 Chr.X: 38674532-38674532 on Build GRCh38
rs992371106 Chr.X: 38674388-38674388 on Build GRCh38
rs867549941 Chr.X: 38674231-38674231 on Build GRCh38
rs752608096 Chr.X: 38674253-38674253 on Build GRCh38
rs11541004 Chr.X: 38674250-38674250 on Build GRCh38
rs779879103 Chr.X: 38674304-38674304 on Build GRCh38
rs929239011 Chr.X: 38674414-38674414 on Build GRCh38
rs746469605 Chr.X: 38674308-38674308 on Build GRCh38
rs143618058 Chr.X: 38674341-38674341 on Build GRCh38
rs764350033 Chr.X: 38674272-38674272 on Build GRCh38
rs747855104 Chr.X: 38674333-38674333 on Build GRCh38
rs1014679116 Chr.X: 38674398-38674398 on Build GRCh38
rs917286155 Chr.X: 38674556-38674556 on Build GRCh38
rs543698328 Chr.X: 38674583-38674583 on Build GRCh38
rs780412215 Chr.X: 38674519-38674519 on Build GRCh38
rs958667710 Chr.X: 38674503-38674503 on Build GRCh38
rs760136867 Chr.X: 38674241-38674241 on Build GRCh38
rs35879710 Chr.X: 38674516-38674517 on Build GRCh38
rs984704278 Chr.X: 38674445-38674445 on Build GRCh38
rs768015255 Chr.X: 38674353-38674353 on Build GRCh38
rs960589004 Chr.X: 38674352-38674352 on Build GRCh38
rs993127156 Chr.X: 38674506-38674506 on Build GRCh38
rs963241553 Chr.X: 38674415-38674415 on Build GRCh38
rs754183451 Chr.X: 38674277-38674277 on Build GRCh38
rs946033380 Chr.X: 38674563-38674563 on Build GRCh38
rs111440990 Chr.X: 38674316-38674316 on Build GRCh38
rs1041824108 Chr.X: 38674573-38674573 on Build GRCh38
rs747193481 Chr.X: 38674588-38674588 on Build GRCh38
rs927079564 Chr.X: 38674442-38674442 on Build GRCh38
rs767461332 Chr.X: 38674249-38674249 on Build GRCh38
rs778903482 Chr.X: 38674283-38674283 on Build GRCh38
rs771884245 Chr.X: 38674201-38674201 on Build GRCh38
rs149275253 Chr.X: 38674312-38674312 on Build GRCh38
rs201047637 Chr.X: 38674291-38674291 on Build GRCh38
rs375853893 Chr.X: 38674199-38674199 on Build GRCh38
rs908911882 Chr.X: 38674530-38674530 on Build GRCh38
rs111443672 Chr.X: 38674317-38674317 on Build GRCh38

View Full Product Details