Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 112455965-112456475 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZCCHC16
Entrez Gene ID
340595
Gene Name
zinc finger CCHC-type containing 16
Gene Aliases
Mar4, Mart4
UniGene
Hs.368140
Gene Chromosome Location
Chr.X: 112083025-112457245 on Build GRCh38
Transcripts
NM_001004308.2

SNP Detail

SNP ID Chromosome Location
rs774717443 Chr.X: 112456065-112456065 on Build GRCh38
rs917297405 Chr.X: 112455992-112455992 on Build GRCh38
rs988946542 Chr.X: 112456330-112456330 on Build GRCh38
rs923306432 Chr.X: 112456425-112456425 on Build GRCh38
rs993299157 Chr.X: 112456066-112456066 on Build GRCh38
rs865834409 Chr.X: 112456118-112456118 on Build GRCh38
rs200735618 Chr.X: 112456119-112456119 on Build GRCh38
rs778853143 Chr.X: 112456386-112456386 on Build GRCh38
rs941587148 Chr.X: 112456045-112456045 on Build GRCh38
rs927263530 Chr.X: 112456428-112456428 on Build GRCh38
rs753468503 Chr.X: 112456154-112456154 on Build GRCh38
rs769431277 Chr.X: 112456266-112456266 on Build GRCh38
rs760381345 Chr.X: 112456202-112456202 on Build GRCh38
rs980528863 Chr.X: 112456372-112456372 on Build GRCh38
rs368094090 Chr.X: 112456377-112456377 on Build GRCh38
rs151241112 Chr.X: 112456201-112456201 on Build GRCh38
rs1038509185 Chr.X: 112456046-112456046 on Build GRCh38
rs917309668 Chr.X: 112456357-112456357 on Build GRCh38
rs148726996 Chr.X: 112456144-112456144 on Build GRCh38
rs145670125 Chr.X: 112456281-112456281 on Build GRCh38
rs115792396 Chr.X: 112456237-112456237 on Build GRCh38
rs964788939 Chr.X: 112456359-112456359 on Build GRCh38
rs763125765 Chr.X: 112456153-112456153 on Build GRCh38
rs142281620 Chr.X: 112456179-112456179 on Build GRCh38
rs760135202 Chr.X: 112456102-112456102 on Build GRCh38
rs753698878 Chr.X: 112456250-112456250 on Build GRCh38
rs376123847 Chr.X: 112456189-112456189 on Build GRCh38
rs1024003512 Chr.X: 112456020-112456020 on Build GRCh38
rs971446294 Chr.X: 112456369-112456369 on Build GRCh38
rs59057814 Chr.X: 112456117-112456117 on Build GRCh38
rs897288580 Chr.X: 112456063-112456063 on Build GRCh38
rs927531481 Chr.X: 112456036-112456036 on Build GRCh38
rs950086699 Chr.X: 112456007-112456007 on Build GRCh38
rs190684647 Chr.X: 112456010-112456010 on Build GRCh38
rs796473463 Chr.X: 112456118-112456118 on Build GRCh38

View Full Product Details