Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 134887550-134888065 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MOSPD1
Entrez Gene ID
56180
Gene Name
motile sperm domain containing 1
Gene Aliases
DJ473B4
UniGene
Hs.590789
Gene Chromosome Location
Chr.X: 134887626-134915344 on Build GRCh38
Transcripts
NM_001306188.1 NM_019556.2 XM_011531366.2 XM_017029653.1 XM_017029654.1

SNP Detail

SNP ID Chromosome Location
rs868676977 Chr.X: 134887703-134887703 on Build GRCh38
rs945024825 Chr.X: 134887949-134887949 on Build GRCh38
rs752103806 Chr.X: 134887648-134887648 on Build GRCh38
rs188364368 Chr.X: 134887908-134887908 on Build GRCh38
rs1056085364 Chr.X: 134887588-134887588 on Build GRCh38
rs771843878 Chr.X: 134887933-134887933 on Build GRCh38
rs892111409 Chr.X: 134887596-134887596 on Build GRCh38
rs755074254 Chr.X: 134887840-134887840 on Build GRCh38
rs756581701 Chr.X: 134887754-134887754 on Build GRCh38
rs919982252 Chr.X: 134887787-134887787 on Build GRCh38
rs1043451385 Chr.X: 134887602-134887602 on Build GRCh38
rs898043936 Chr.X: 134887709-134887709 on Build GRCh38
rs748261794 Chr.X: 134887914-134887914 on Build GRCh38
rs1027787878 Chr.X: 134887715-134887715 on Build GRCh38
rs747567608 Chr.X: 134887719-134887719 on Build GRCh38
rs1036719295 Chr.X: 134887708-134887708 on Build GRCh38
rs1013000627 Chr.X: 134887726-134887726 on Build GRCh38
rs928792404 Chr.X: 134887900-134887900 on Build GRCh38
rs866600813 Chr.X: 134887702-134887702 on Build GRCh38
rs1009767393 Chr.X: 134887601-134887601 on Build GRCh38
rs768082730 Chr.X: 134887718-134887718 on Build GRCh38
rs996670943 Chr.X: 134887616-134887616 on Build GRCh38
rs183470423 Chr.X: 134887885-134887885 on Build GRCh38
rs752056995 Chr.X: 134887904-134887904 on Build GRCh38
rs764700370 Chr.X: 134887636-134887636 on Build GRCh38
rs771246250 Chr.X: 134887690-134887691 on Build GRCh38
rs780395732 Chr.X: 134887763-134887763 on Build GRCh38
rs1031683093 Chr.X: 134887839-134887839 on Build GRCh38
rs6638293 Chr.X: 134887613-134887613 on Build GRCh38
rs938958855 Chr.X: 134887901-134887901 on Build GRCh38
rs971402153 Chr.X: 134887821-134887821 on Build GRCh38
rs749574506 Chr.X: 134887827-134887827 on Build GRCh38
rs776296253 Chr.X: 134887581-134887581 on Build GRCh38
rs955304436 Chr.X: 134887653-134887653 on Build GRCh38
rs993759209 Chr.X: 134887711-134887711 on Build GRCh38
rs757640145 Chr.X: 134887694-134887694 on Build GRCh38
rs1053293013 Chr.X: 134887902-134887902 on Build GRCh38
rs750767561 Chr.X: 134887740-134887740 on Build GRCh38
rs981823864 Chr.X: 134887826-134887826 on Build GRCh38
rs974570738 Chr.X: 134887771-134887771 on Build GRCh38

View Full Product Details