Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 135169438-135169838 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CT55
Entrez Gene ID
54967
Gene Name
cancer/testis antigen 55
Gene Aliases
BJHCC20A, CXorf48
UniGene
Hs.272804
Gene Chromosome Location
Chr.X: 135156536-135171827 on Build GRCh38
Transcripts
NM_001031705.2 NM_017863.2 XM_005262427.1

SNP Detail

SNP ID Chromosome Location
rs782578766 Chr.X: 135169644-135169644 on Build GRCh38
rs782371793 Chr.X: 135169667-135169667 on Build GRCh38
rs782292242 Chr.X: 135169630-135169630 on Build GRCh38
rs781785637 Chr.X: 135169473-135169473 on Build GRCh38
rs782527240 Chr.X: 135169594-135169594 on Build GRCh38
rs781806839 Chr.X: 135169810-135169810 on Build GRCh38
rs782738818 Chr.X: 135169719-135169719 on Build GRCh38
rs781997682 Chr.X: 135169670-135169670 on Build GRCh38
rs782682774 Chr.X: 135169625-135169625 on Build GRCh38
rs782122552 Chr.X: 135169756-135169756 on Build GRCh38
rs782553752 Chr.X: 135169798-135169798 on Build GRCh38
rs781861495 Chr.X: 135169584-135169584 on Build GRCh38
rs781840816 Chr.X: 135169724-135169724 on Build GRCh38
rs782149979 Chr.X: 135169672-135169672 on Build GRCh38
rs200818862 Chr.X: 135169564-135169564 on Build GRCh38
rs782785117 Chr.X: 135169787-135169787 on Build GRCh38
rs782814553 Chr.X: 135169466-135169466 on Build GRCh38
rs559141080 Chr.X: 135169477-135169477 on Build GRCh38
rs782056530 Chr.X: 135169706-135169706 on Build GRCh38
rs782468519 Chr.X: 135169558-135169558 on Build GRCh38
rs782673658 Chr.X: 135169572-135169572 on Build GRCh38
rs781894647 Chr.X: 135169795-135169795 on Build GRCh38
rs151255483 Chr.X: 135169809-135169809 on Build GRCh38
rs113172956 Chr.X: 135169811-135169811 on Build GRCh38
rs375438569 Chr.X: 135169675-135169675 on Build GRCh38
rs782442061 Chr.X: 135169504-135169504 on Build GRCh38
rs782214720 Chr.X: 135169651-135169651 on Build GRCh38
rs150387330 Chr.X: 135169632-135169632 on Build GRCh38
rs782603164 Chr.X: 135169508-135169508 on Build GRCh38

View Full Product Details