Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 211940694-211941204 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INTS7
Entrez Gene ID
25896
Gene Name
integrator complex subunit 7
Gene Aliases
C1orf73, INT7
UniGene
Hs.369285
Gene Chromosome Location
Chr.1: 211940328-212035660 on Build GRCh38
Transcripts
NM_001199809.1 NM_001199811.1 NM_001199812.1 NM_015434.3 NR_037667.1 XM_011509396.2

SNP Detail

SNP ID Chromosome Location
rs1002959284 Chr.1: 211941167-211941168 on Build GRCh38
rs184783200 Chr.1: 211941166-211941166 on Build GRCh38
rs1053105069 Chr.1: 211940996-211940996 on Build GRCh38
rs566980048 Chr.1: 211940839-211940839 on Build GRCh38
rs1011688458 Chr.1: 211941033-211941033 on Build GRCh38
rs371120153 Chr.1: 211941175-211941175 on Build GRCh38
rs902299561 Chr.1: 211941025-211941025 on Build GRCh38
rs946402730 Chr.1: 211941005-211941005 on Build GRCh38
rs540692209 Chr.1: 211941045-211941045 on Build GRCh38
rs191947881 Chr.1: 211941151-211941151 on Build GRCh38
rs1007105375 Chr.1: 211940883-211940884 on Build GRCh38
rs993850397 Chr.1: 211941106-211941106 on Build GRCh38
rs898737412 Chr.1: 211941111-211941111 on Build GRCh38
rs935088788 Chr.1: 211941052-211941052 on Build GRCh38
rs1025633845 Chr.1: 211941140-211941140 on Build GRCh38
rs553075923 Chr.1: 211940962-211940962 on Build GRCh38
rs972441506 Chr.1: 211940897-211940897 on Build GRCh38
rs182453962 Chr.1: 211941024-211941024 on Build GRCh38
rs559832186 Chr.1: 211941169-211941169 on Build GRCh38
rs549170989 Chr.1: 211940801-211940801 on Build GRCh38
rs375201220 Chr.1: 211940916-211940916 on Build GRCh38
rs571132226 Chr.1: 211940966-211940966 on Build GRCh38
rs974073406 Chr.1: 211940920-211940920 on Build GRCh38
rs187489037 Chr.1: 211941083-211941083 on Build GRCh38
rs1000125620 Chr.1: 211940826-211940826 on Build GRCh38
rs374959133 Chr.1: 211940941-211940944 on Build GRCh38
rs918352581 Chr.1: 211940900-211940900 on Build GRCh38
rs959664465 Chr.1: 211941095-211941095 on Build GRCh38
rs1035347672 Chr.1: 211940827-211940827 on Build GRCh38
rs942603070 Chr.1: 211940944-211940944 on Build GRCh38
rs891899845 Chr.1: 211940999-211940999 on Build GRCh38
rs995939009 Chr.1: 211941114-211941114 on Build GRCh38
rs1043379665 Chr.1: 211941021-211941021 on Build GRCh38
rs139565960 Chr.1: 211941162-211941162 on Build GRCh38
rs959734899 Chr.1: 211940833-211940833 on Build GRCh38
rs77582459 Chr.1: 211941150-211941150 on Build GRCh38
rs1018442941 Chr.1: 211940911-211940914 on Build GRCh38
rs78481394 Chr.1: 211941081-211941081 on Build GRCh38
rs916471393 Chr.1: 211941002-211941002 on Build GRCh38
rs577974977 Chr.1: 211940803-211940803 on Build GRCh38
rs1043605839 Chr.1: 211941070-211941070 on Build GRCh38
rs771514617 Chr.1: 211940788-211940788 on Build GRCh38
rs779578099 Chr.1: 211940824-211940824 on Build GRCh38
rs745406072 Chr.1: 211940719-211940719 on Build GRCh38
rs983920925 Chr.1: 211941157-211941157 on Build GRCh38
rs202231233 Chr.1: 211940943-211940944 on Build GRCh38
rs990752228 Chr.1: 211940968-211940968 on Build GRCh38
rs1021857038 Chr.1: 211940772-211940772 on Build GRCh38
rs970698021 Chr.1: 211940806-211940806 on Build GRCh38
rs573203527 Chr.1: 211940841-211940842 on Build GRCh38
rs984342849 Chr.1: 211940921-211940921 on Build GRCh38
rs768393576 Chr.1: 211940976-211940976 on Build GRCh38
rs1050769563 Chr.1: 211941071-211941071 on Build GRCh38
rs1035950423 Chr.1: 211941094-211941094 on Build GRCh38
rs150575437 Chr.1: 211940925-211940925 on Build GRCh38

View Full Product Details