Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 212989929-212990472 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VASH2
Entrez Gene ID
79805
Gene Name
vasohibin 2
Gene Aliases
-
UniGene
Hs.96885
Gene Chromosome Location
Chr.1: 212935543-212991585 on Build GRCh38
Transcripts
NM_001136474.2 NM_001136475.2 NM_001301056.1 NM_024749.4 XM_011509987.2 XM_011509988.2 XM_011509991.2 XM_011509994.1 XM_011509995.1 XM_017002351.1 XM_017002352.1

SNP Detail

SNP ID Chromosome Location
rs768021069 Chr.1: 212990043-212990043 on Build GRCh38
rs937371146 Chr.1: 212990379-212990379 on Build GRCh38
rs987414215 Chr.1: 212990412-212990413 on Build GRCh38
rs191404316 Chr.1: 212990306-212990306 on Build GRCh38
rs969009867 Chr.1: 212990324-212990324 on Build GRCh38
rs778068525 Chr.1: 212990106-212990106 on Build GRCh38
rs1029453533 Chr.1: 212990231-212990231 on Build GRCh38
rs994047245 Chr.1: 212990075-212990075 on Build GRCh38
rs958899095 Chr.1: 212990284-212990284 on Build GRCh38
rs956442012 Chr.1: 212990267-212990267 on Build GRCh38
rs397965154 Chr.1: 212990075-212990076 on Build GRCh38
rs756974169 Chr.1: 212990056-212990056 on Build GRCh38
rs1003095573 Chr.1: 212990243-212990248 on Build GRCh38
rs550740224 Chr.1: 212990254-212990254 on Build GRCh38
rs115693012 Chr.1: 212990361-212990361 on Build GRCh38
rs927529980 Chr.1: 212990364-212990364 on Build GRCh38
rs997840426 Chr.1: 212990225-212990225 on Build GRCh38
rs869103247 Chr.1: 212990074-212990075 on Build GRCh38
rs555116079 Chr.1: 212990395-212990395 on Build GRCh38
rs988997245 Chr.1: 212990286-212990286 on Build GRCh38
rs1021756070 Chr.1: 212990297-212990297 on Build GRCh38
rs879073944 Chr.1: 212990131-212990131 on Build GRCh38
rs187914837 Chr.1: 212990178-212990178 on Build GRCh38
rs977464049 Chr.1: 212990348-212990348 on Build GRCh38
rs1041386417 Chr.1: 212990051-212990054 on Build GRCh38
rs76312391 Chr.1: 212990060-212990060 on Build GRCh38
rs888975330 Chr.1: 212990181-212990181 on Build GRCh38
rs759720161 Chr.1: 212989966-212989966 on Build GRCh38
rs933239424 Chr.1: 212990121-212990121 on Build GRCh38
rs878992709 Chr.1: 212990396-212990396 on Build GRCh38
rs546851549 Chr.1: 212989993-212989993 on Build GRCh38
rs895891043 Chr.1: 212990057-212990057 on Build GRCh38
rs927268407 Chr.1: 212989962-212989962 on Build GRCh38
rs766171558 Chr.1: 212990217-212990219 on Build GRCh38
rs1035808965 Chr.1: 212990248-212990248 on Build GRCh38
rs1025648725 Chr.1: 212990230-212990230 on Build GRCh38
rs1017974324 Chr.1: 212990133-212990133 on Build GRCh38
rs888370272 Chr.1: 212990096-212990096 on Build GRCh38
rs947083211 Chr.1: 212989967-212989967 on Build GRCh38
rs139796805 Chr.1: 212990081-212990081 on Build GRCh38
rs75620216 Chr.1: 212990063-212990063 on Build GRCh38
rs898492648 Chr.1: 212990218-212990218 on Build GRCh38
rs1043070412 Chr.1: 212989953-212989953 on Build GRCh38
rs1006961807 Chr.1: 212990182-212990182 on Build GRCh38
rs867182327 Chr.1: 212989978-212989978 on Build GRCh38
rs57314416 Chr.1: 212990069-212990070 on Build GRCh38

View Full Product Details