Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 247037468-247037968 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF670
Entrez Gene ID
93474
Gene Name
zinc finger protein 670
Gene Aliases
-
UniGene
Hs.732305
Gene Chromosome Location
Chr.1: 247034638-247078813 on Build GRCh38
Transcripts
NM_001204220.1 NM_033213.4 NR_037894.1

Gene Symbol
ZNF670-ZNF695
Entrez Gene ID
100533111
Gene Name
ZNF670-ZNF695 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.732305
Gene Chromosome Location
Chr.1: 246945547-247078813 on Build GRCh38
Transcripts
NM_001204220.1 NM_033213.4 NR_037894.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2052369 c.1089A>C p.K363N Chr.1: 247037530-247037530 on Build GRCh38
6832258 c.821C>T p.T274M Chr.1: 247037798-247037798 on Build GRCh38
2052374 c.948A>C p.K316N Chr.1: 247037671-247037671 on Build GRCh38
4629945 c.1033T>A p.S345T Chr.1: 247037586-247037586 on Build GRCh38
6832262 c.709delT p.S237fs*48 Chr.1: 247037910-247037910 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775463462 Chr.1: 247037738-247037738 on Build GRCh38
rs768067873 Chr.1: 247037892-247037892 on Build GRCh38
rs780011896 Chr.1: 247037561-247037561 on Build GRCh38
rs899149944 Chr.1: 247037527-247037527 on Build GRCh38
rs774152488 Chr.1: 247037594-247037594 on Build GRCh38
rs978514345 Chr.1: 247037682-247037682 on Build GRCh38
rs763666416 Chr.1: 247037528-247037528 on Build GRCh38
rs375731131 Chr.1: 247037604-247037604 on Build GRCh38
rs975387651 Chr.1: 247037614-247037614 on Build GRCh38
rs752102748 Chr.1: 247037677-247037677 on Build GRCh38
rs770892784 Chr.1: 247037585-247037585 on Build GRCh38
rs778120181 Chr.1: 247037825-247037825 on Build GRCh38
rs757380390 Chr.1: 247037927-247037927 on Build GRCh38
rs750743937 Chr.1: 247037655-247037655 on Build GRCh38
rs879138633 Chr.1: 247037926-247037926 on Build GRCh38
rs768856212 Chr.1: 247037569-247037569 on Build GRCh38
rs150721271 Chr.1: 247037828-247037828 on Build GRCh38
rs142649426 Chr.1: 247037675-247037675 on Build GRCh38
rs752199734 Chr.1: 247037497-247037497 on Build GRCh38
rs755773046 Chr.1: 247037511-247037511 on Build GRCh38
rs778583714 Chr.1: 247037710-247037710 on Build GRCh38
rs527677872 Chr.1: 247037691-247037691 on Build GRCh38
rs547402830 Chr.1: 247037798-247037798 on Build GRCh38
rs949891831 Chr.1: 247037780-247037780 on Build GRCh38
rs780678776 Chr.1: 247037670-247037670 on Build GRCh38
rs769174733 Chr.1: 247037860-247037860 on Build GRCh38
rs746978429 Chr.1: 247037842-247037842 on Build GRCh38
rs764878390 Chr.1: 247037764-247037764 on Build GRCh38
rs138056144 Chr.1: 247037603-247037603 on Build GRCh38
rs372441684 Chr.1: 247037494-247037494 on Build GRCh38
rs765544070 Chr.1: 247037640-247037640 on Build GRCh38
rs1002632866 Chr.1: 247037503-247037503 on Build GRCh38
rs369428325 Chr.1: 247037898-247037898 on Build GRCh38
rs779034323 Chr.1: 247037695-247037695 on Build GRCh38
rs779838721 Chr.1: 247037928-247037928 on Build GRCh38
rs528107236 Chr.1: 247037579-247037579 on Build GRCh38
rs770006873 Chr.1: 247037708-247037708 on Build GRCh38
rs751744636 Chr.1: 247037848-247037848 on Build GRCh38
rs145888979 Chr.1: 247037940-247037940 on Build GRCh38
rs760244659 Chr.1: 247037609-247037609 on Build GRCh38
rs779791671 Chr.1: 247037840-247037840 on Build GRCh38
rs377329417 Chr.1: 247037762-247037762 on Build GRCh38
rs753249219 Chr.1: 247037893-247037893 on Build GRCh38
rs899203739 Chr.1: 247037838-247037838 on Build GRCh38
rs745350235 Chr.1: 247037717-247037718 on Build GRCh38
rs750041002 Chr.1: 247037772-247037772 on Build GRCh38
rs376049479 Chr.1: 247037861-247037861 on Build GRCh38
rs754127403 Chr.1: 247037923-247037923 on Build GRCh38
rs759412331 Chr.1: 247037595-247037595 on Build GRCh38
rs956442318 Chr.1: 247037722-247037722 on Build GRCh38
rs141125753 Chr.1: 247037807-247037807 on Build GRCh38
rs779312869 Chr.1: 247037549-247037549 on Build GRCh38
rs372416787 Chr.1: 247037555-247037555 on Build GRCh38
rs752153935 Chr.1: 247037797-247037797 on Build GRCh38
rs781440328 Chr.1: 247037801-247037801 on Build GRCh38
rs776072379 Chr.1: 247037755-247037755 on Build GRCh38
rs149848448 Chr.1: 247037897-247037897 on Build GRCh38
rs148842493 Chr.1: 247037584-247037584 on Build GRCh38
rs745606608 Chr.1: 247037718-247037718 on Build GRCh38
rs755187559 Chr.1: 247037856-247037856 on Build GRCh38
rs370603456 Chr.1: 247037637-247037637 on Build GRCh38
rs766800445 Chr.1: 247037882-247037882 on Build GRCh38
rs763580814 Chr.1: 247037610-247037610 on Build GRCh38
rs757674232 Chr.1: 247037547-247037547 on Build GRCh38
rs1022657545 Chr.1: 247037639-247037639 on Build GRCh38
rs563040127 Chr.1: 247037562-247037562 on Build GRCh38
rs370919641 Chr.1: 247037777-247037777 on Build GRCh38
rs147292690 Chr.1: 247037883-247037883 on Build GRCh38
rs779567952 Chr.1: 247037848-247037848 on Build GRCh38
rs766634455 Chr.1: 247037795-247037795 on Build GRCh38
rs758891351 Chr.1: 247037559-247037559 on Build GRCh38
rs761453478 Chr.1: 247037857-247037857 on Build GRCh38
rs748414939 Chr.1: 247037700-247037700 on Build GRCh38
rs771756174 Chr.1: 247037727-247037727 on Build GRCh38
rs776125630 Chr.1: 247037853-247037853 on Build GRCh38
rs201836316 Chr.1: 247037638-247037638 on Build GRCh38
rs771668964 Chr.1: 247037720-247037721 on Build GRCh38
rs550074620 Chr.1: 247037517-247037517 on Build GRCh38
rs752764783 Chr.1: 247037806-247037806 on Build GRCh38
rs200652027 Chr.1: 247037891-247037891 on Build GRCh38
rs753535700 Chr.1: 247037619-247037619 on Build GRCh38
rs988974136 Chr.1: 247037753-247037753 on Build GRCh38
rs988557041 Chr.1: 247037649-247037649 on Build GRCh38
rs760582002 Chr.1: 247037746-247037746 on Build GRCh38
rs762486208 Chr.1: 247037879-247037879 on Build GRCh38
rs764056586 Chr.1: 247037901-247037901 on Build GRCh38
rs546352675 Chr.1: 247037689-247037689 on Build GRCh38
rs771586236 Chr.1: 247037506-247037508 on Build GRCh38
rs753484437 Chr.1: 247037543-247037543 on Build GRCh38
rs373991730 Chr.1: 247037662-247037662 on Build GRCh38
rs146950973 Chr.1: 247037627-247037627 on Build GRCh38
rs771950911 Chr.1: 247037829-247037829 on Build GRCh38
rs777055005 Chr.1: 247037573-247037573 on Build GRCh38

View Full Product Details