Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 95911235-95911754 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf131
Entrez Gene ID
100127889
Gene Name
chromosome 10 open reading frame 131
Gene Aliases
bA690P14.3
UniGene
Hs.719076
Gene Chromosome Location
Chr.10: 95907965-95938658 on Build GRCh38
Transcripts
NM_001130446.2 NR_038444.1

Gene Symbol
ENTPD1-AS1
Entrez Gene ID
728558
Gene Name
ENTPD1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.538374
Gene Chromosome Location
Chr.10: 95753206-96090235 on Build GRCh38
Transcripts
NM_001130446.2 NR_038444.1

SNP Detail

SNP ID Chromosome Location
rs941078650 Chr.10: 95911690-95911690 on Build GRCh38
rs773313769 Chr.10: 95911348-95911348 on Build GRCh38
rs922886585 Chr.10: 95911629-95911629 on Build GRCh38
rs905052010 Chr.10: 95911283-95911283 on Build GRCh38
rs578245572 Chr.10: 95911597-95911597 on Build GRCh38
rs866121721 Chr.10: 95911641-95911641 on Build GRCh38
rs540063759 Chr.10: 95911692-95911692 on Build GRCh38
rs923107637 Chr.10: 95911530-95911530 on Build GRCh38
rs11596697 Chr.10: 95911265-95911265 on Build GRCh38
rs530389578 Chr.10: 95911516-95911516 on Build GRCh38
rs1020296219 Chr.10: 95911405-95911405 on Build GRCh38
rs1034982131 Chr.10: 95911309-95911309 on Build GRCh38
rs1038595312 Chr.10: 95911278-95911280 on Build GRCh38
rs980607419 Chr.10: 95911338-95911338 on Build GRCh38
rs142415049 Chr.10: 95911316-95911316 on Build GRCh38
rs774337973 Chr.10: 95911422-95911422 on Build GRCh38
rs925018674 Chr.10: 95911645-95911645 on Build GRCh38
rs1034410297 Chr.10: 95911360-95911360 on Build GRCh38
rs1033587763 Chr.10: 95911591-95911591 on Build GRCh38
rs543789831 Chr.10: 95911618-95911618 on Build GRCh38
rs907827859 Chr.10: 95911416-95911416 on Build GRCh38
rs975988170 Chr.10: 95911526-95911526 on Build GRCh38
rs931808071 Chr.10: 95911537-95911537 on Build GRCh38
rs557645048 Chr.10: 95911648-95911648 on Build GRCh38
rs952050880 Chr.10: 95911366-95911366 on Build GRCh38
rs529932206 Chr.10: 95911695-95911695 on Build GRCh38
rs1014215537 Chr.10: 95911377-95911377 on Build GRCh38
rs908233731 Chr.10: 95911667-95911667 on Build GRCh38
rs967453957 Chr.10: 95911555-95911558 on Build GRCh38
rs375485375 Chr.10: 95911652-95911652 on Build GRCh38
rs978838887 Chr.10: 95911568-95911571 on Build GRCh38
rs777132203 Chr.10: 95911272-95911272 on Build GRCh38
rs940587597 Chr.10: 95911420-95911420 on Build GRCh38
rs971555629 Chr.10: 95911290-95911290 on Build GRCh38
rs377515021 Chr.10: 95911460-95911460 on Build GRCh38
rs984777860 Chr.10: 95911410-95911410 on Build GRCh38
rs564916962 Chr.10: 95911320-95911320 on Build GRCh38
rs559972862 Chr.10: 95911718-95911718 on Build GRCh38

View Full Product Details