Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 100779382-100779927 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAX2
Entrez Gene ID
5076
Gene Name
paired box 2
Gene Aliases
FSGS7, PAPRS
UniGene
Hs.155644
Gene Chromosome Location
Chr.10: 100732940-100829941 on Build GRCh38
Transcripts
NM_000278.4 NM_001304569.1 NM_003987.4 NM_003988.4 NM_003989.4 NM_003990.4 XM_017016281.1 XM_017016282.1 XM_017016283.1 XM_017016284.1 XM_017016285.1 XM_017016286.1 XM_017016287.1 XM_017016288.1 XM_017016289.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5592372 c.441C>T p.F147F Chr.10: 100779528-100779528 on Build GRCh38
5523401 c.452C>T p.P151L Chr.10: 100779539-100779539 on Build GRCh38
6128218 c.443A>T p.H148L Chr.10: 100779530-100779530 on Build GRCh38
5592373 c.441C>T p.F147F Chr.10: 100779528-100779528 on Build GRCh38
4450223 c.496+1G>A p.? Chr.10: 100779584-100779584 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs895636918 Chr.10: 100779519-100779519 on Build GRCh38
rs187133715 Chr.10: 100779682-100779682 on Build GRCh38
rs532486518 Chr.10: 100779863-100779863 on Build GRCh38
rs754769029 Chr.10: 100779544-100779544 on Build GRCh38
rs373723559 Chr.10: 100779536-100779536 on Build GRCh38
rs895689159 Chr.10: 100779833-100779833 on Build GRCh38
rs753431059 Chr.10: 100779543-100779543 on Build GRCh38
rs776057750 Chr.10: 100779623-100779623 on Build GRCh38
rs199876625 Chr.10: 100779540-100779540 on Build GRCh38
rs376030530 Chr.10: 100779464-100779464 on Build GRCh38
rs1043412511 Chr.10: 100779445-100779445 on Build GRCh38
rs776956884 Chr.10: 100779452-100779452 on Build GRCh38
rs139874530 Chr.10: 100779514-100779514 on Build GRCh38
rs550483970 Chr.10: 100779887-100779887 on Build GRCh38
rs201383632 Chr.10: 100779565-100779565 on Build GRCh38
rs907097622 Chr.10: 100779747-100779747 on Build GRCh38
rs796880441 Chr.10: 100779671-100779672 on Build GRCh38
rs770138233 Chr.10: 100779614-100779614 on Build GRCh38
rs747922414 Chr.10: 100779551-100779551 on Build GRCh38
rs925442936 Chr.10: 100779900-100779900 on Build GRCh38
rs751744609 Chr.10: 100779810-100779810 on Build GRCh38
rs868385559 Chr.10: 100779407-100779407 on Build GRCh38
rs1042543739 Chr.10: 100779642-100779642 on Build GRCh38
rs888208200 Chr.10: 100779861-100779861 on Build GRCh38
rs975732199 Chr.10: 100779879-100779879 on Build GRCh38
rs923508893 Chr.10: 100779619-100779619 on Build GRCh38
rs1056966390 Chr.10: 100779801-100779801 on Build GRCh38
rs773645514 Chr.10: 100779867-100779869 on Build GRCh38
rs778742674 Chr.10: 100779547-100779547 on Build GRCh38
rs1005930116 Chr.10: 100779843-100779843 on Build GRCh38
rs541291517 Chr.10: 100779487-100779487 on Build GRCh38
rs565173273 Chr.10: 100779728-100779728 on Build GRCh38
rs573718382 Chr.10: 100779448-100779448 on Build GRCh38
rs1045660915 Chr.10: 100779694-100779694 on Build GRCh38
rs7901210 Chr.10: 100779672-100779672 on Build GRCh38
rs577483360 Chr.10: 100779592-100779592 on Build GRCh38
rs1017362324 Chr.10: 100779862-100779862 on Build GRCh38
rs1013538576 Chr.10: 100779693-100779693 on Build GRCh38
rs887529376 Chr.10: 100779838-100779838 on Build GRCh38
rs372061953 Chr.10: 100779447-100779447 on Build GRCh38
rs993753624 Chr.10: 100779785-100779785 on Build GRCh38
rs563941289 Chr.10: 100779641-100779641 on Build GRCh38
rs889144026 Chr.10: 100779417-100779417 on Build GRCh38
rs961012910 Chr.10: 100779885-100779885 on Build GRCh38
rs199919058 Chr.10: 100779539-100779539 on Build GRCh38
rs865906227 Chr.10: 100779506-100779506 on Build GRCh38
rs138946406 Chr.10: 100779670-100779670 on Build GRCh38
rs1044580702 Chr.10: 100779857-100779857 on Build GRCh38
rs903161039 Chr.10: 100779702-100779702 on Build GRCh38
rs934603878 Chr.10: 100779729-100779729 on Build GRCh38
rs191583108 Chr.10: 100779889-100779889 on Build GRCh38
rs777007561 Chr.10: 100779598-100779598 on Build GRCh38
rs978679028 Chr.10: 100779894-100779894 on Build GRCh38
rs369637148 Chr.10: 100779485-100779485 on Build GRCh38
rs867272170 Chr.10: 100779429-100779429 on Build GRCh38
rs746022143 Chr.10: 100779613-100779613 on Build GRCh38
rs753261521 Chr.10: 100779488-100779488 on Build GRCh38
rs544848152 Chr.10: 100779887-100779889 on Build GRCh38
rs546868338 Chr.10: 100779690-100779690 on Build GRCh38
rs80030703 Chr.10: 100779671-100779671 on Build GRCh38
rs747148327 Chr.10: 100779591-100779591 on Build GRCh38
rs201775091 Chr.10: 100779564-100779564 on Build GRCh38
rs370214925 Chr.10: 100779578-100779578 on Build GRCh38
rs572949603 Chr.10: 100779761-100779761 on Build GRCh38
rs373077911 Chr.10: 100779457-100779457 on Build GRCh38

View Full Product Details