Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 102450795-102451333 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf95
Entrez Gene ID
79946
Gene Name
chromosome 10 open reading frame 95
Gene Aliases
-
UniGene
Hs.225084
Gene Chromosome Location
Chr.10: 102449837-102451543 on Build GRCh38
Transcripts
NM_024886.2 NR_038937.1 NR_038938.1

Gene Symbol
RPARP-AS1
Entrez Gene ID
100505761
Gene Name
RPARP antisense RNA 1
Gene Aliases
-
UniGene
Hs.656457
Gene Chromosome Location
Chr.10: 102449817-102456293 on Build GRCh38
Transcripts
NM_024886.2 NR_038937.1 NR_038938.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6518221 c.329C>T p.P110L Chr.10: 102450902-102450902 on Build GRCh38
53484 c.139T>C p.C47R Chr.10: 102451092-102451092 on Build GRCh38
6570713 c.104C>T p.P35L Chr.10: 102451127-102451127 on Build GRCh38
6613970 c.190_195delCGCAGC p.R64_S65delRS Chr.10: 102451036-102451041 on Build GRCh38
5359728 c.131delG p.G44fs*>214 Chr.10: 102451100-102451100 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758514002 Chr.10: 102450982-102450982 on Build GRCh38
rs769127436 Chr.10: 102451044-102451067 on Build GRCh38
rs377649213 Chr.10: 102451129-102451129 on Build GRCh38
rs762475767 Chr.10: 102451100-102451100 on Build GRCh38
rs748775530 Chr.10: 102450944-102450944 on Build GRCh38
rs930731895 Chr.10: 102451232-102451232 on Build GRCh38
rs759368438 Chr.10: 102451160-102451160 on Build GRCh38
rs765493137 Chr.10: 102451074-102451074 on Build GRCh38
rs1007445497 Chr.10: 102451073-102451073 on Build GRCh38
rs776973795 Chr.10: 102450993-102450993 on Build GRCh38
rs560351019 Chr.10: 102450882-102450882 on Build GRCh38
rs200179377 Chr.10: 102451002-102451002 on Build GRCh38
rs753672910 Chr.10: 102451048-102451048 on Build GRCh38
rs757765766 Chr.10: 102451100-102451100 on Build GRCh38
rs897815404 Chr.10: 102451014-102451014 on Build GRCh38
rs950620359 Chr.10: 102450834-102450834 on Build GRCh38
rs771634162 Chr.10: 102451015-102451015 on Build GRCh38
rs771660367 Chr.10: 102451131-102451131 on Build GRCh38
rs753003860 Chr.10: 102451085-102451085 on Build GRCh38
rs965788520 Chr.10: 102450883-102450883 on Build GRCh38
rs373519518 Chr.10: 102451105-102451105 on Build GRCh38
rs775078501 Chr.10: 102450920-102450920 on Build GRCh38
rs930701254 Chr.10: 102451028-102451028 on Build GRCh38
rs1049534442 Chr.10: 102451040-102451040 on Build GRCh38
rs767289700 Chr.10: 102450947-102450947 on Build GRCh38
rs751654758 Chr.10: 102451096-102451096 on Build GRCh38
rs898527216 Chr.10: 102451103-102451103 on Build GRCh38
rs968202920 Chr.10: 102451127-102451127 on Build GRCh38
rs761322132 Chr.10: 102450945-102450945 on Build GRCh38
rs995956142 Chr.10: 102451251-102451251 on Build GRCh38
rs1042618889 Chr.10: 102451305-102451305 on Build GRCh38
rs889000259 Chr.10: 102451050-102451050 on Build GRCh38
rs752487556 Chr.10: 102451165-102451165 on Build GRCh38
rs760627889 Chr.10: 102451043-102451043 on Build GRCh38
rs200900514 Chr.10: 102450997-102450997 on Build GRCh38
rs766172988 Chr.10: 102450960-102450960 on Build GRCh38
rs73349055 Chr.10: 102451104-102451104 on Build GRCh38
rs1020740985 Chr.10: 102451125-102451125 on Build GRCh38
rs995151539 Chr.10: 102450846-102450846 on Build GRCh38
rs528942640 Chr.10: 102450962-102450962 on Build GRCh38
rs759419410 Chr.10: 102451049-102451049 on Build GRCh38
rs775214184 Chr.10: 102451047-102451067 on Build GRCh38
rs943275917 Chr.10: 102451276-102451276 on Build GRCh38
rs1028742985 Chr.10: 102451262-102451262 on Build GRCh38
rs1018809470 Chr.10: 102451167-102451167 on Build GRCh38
rs994793066 Chr.10: 102450815-102450817 on Build GRCh38
rs189174307 Chr.10: 102450986-102450986 on Build GRCh38
rs531325093 Chr.10: 102451240-102451241 on Build GRCh38
rs1049008247 Chr.10: 102451109-102451109 on Build GRCh38
rs534224224 Chr.10: 102451163-102451163 on Build GRCh38
rs764277599 Chr.10: 102451192-102451192 on Build GRCh38
rs761000976 Chr.10: 102451178-102451179 on Build GRCh38
rs577440295 Chr.10: 102451273-102451273 on Build GRCh38
rs544397683 Chr.10: 102451286-102451286 on Build GRCh38
rs987873901 Chr.10: 102450915-102450915 on Build GRCh38
rs746775352 Chr.10: 102451025-102451041 on Build GRCh38
rs910576283 Chr.10: 102451245-102451245 on Build GRCh38
rs888945401 Chr.10: 102450873-102450873 on Build GRCh38
rs770935419 Chr.10: 102451145-102451145 on Build GRCh38
rs2281878 Chr.10: 102450978-102450978 on Build GRCh38
rs193070947 Chr.10: 102451132-102451132 on Build GRCh38
rs979418971 Chr.10: 102450948-102450948 on Build GRCh38
rs763799657 Chr.10: 102450925-102450925 on Build GRCh38
rs776428690 Chr.10: 102451038-102451039 on Build GRCh38
rs946286076 Chr.10: 102451009-102451009 on Build GRCh38
rs771360324 Chr.10: 102450907-102450907 on Build GRCh38
rs919310021 Chr.10: 102451221-102451221 on Build GRCh38
rs901712519 Chr.10: 102451079-102451079 on Build GRCh38
rs972556520 Chr.10: 102451213-102451213 on Build GRCh38
rs779336748 Chr.10: 102450973-102450973 on Build GRCh38
rs202020850 Chr.10: 102451012-102451012 on Build GRCh38
rs548199586 Chr.10: 102450849-102450849 on Build GRCh38
rs773616380 Chr.10: 102451174-102451175 on Build GRCh38
rs763206795 Chr.10: 102451178-102451178 on Build GRCh38
rs766296419 Chr.10: 102451044-102451044 on Build GRCh38
rs755814882 Chr.10: 102450952-102450952 on Build GRCh38
rs558974227 Chr.10: 102451218-102451218 on Build GRCh38
rs971592561 Chr.10: 102451176-102451176 on Build GRCh38
rs551745793 Chr.10: 102450965-102450965 on Build GRCh38
rs201345996 Chr.10: 102450961-102450961 on Build GRCh38
rs777657843 Chr.10: 102450968-102450969 on Build GRCh38
rs758363775 Chr.10: 102451086-102451086 on Build GRCh38
rs567769933 Chr.10: 102450889-102450889 on Build GRCh38
rs1049029119 Chr.10: 102451234-102451234 on Build GRCh38
rs758297752 Chr.10: 102450823-102450833 on Build GRCh38
rs762559106 Chr.10: 102450922-102450922 on Build GRCh38
rs778978294 Chr.10: 102451196-102451196 on Build GRCh38
rs750405943 Chr.10: 102450950-102450950 on Build GRCh38
rs771031631 Chr.10: 102451039-102451041 on Build GRCh38
rs755249312 Chr.10: 102450971-102450971 on Build GRCh38
rs3740417 Chr.10: 102451301-102451301 on Build GRCh38
rs965743844 Chr.10: 102450911-102450911 on Build GRCh38
rs773785584 Chr.10: 102450854-102450854 on Build GRCh38
rs1040302823 Chr.10: 102451284-102451284 on Build GRCh38
rs772644512 Chr.10: 102451038-102451038 on Build GRCh38
rs767891491 Chr.10: 102451173-102451174 on Build GRCh38
rs776715082 Chr.10: 102451156-102451156 on Build GRCh38
rs778487332 Chr.10: 102451106-102451106 on Build GRCh38
rs777889192 Chr.10: 102451092-102451092 on Build GRCh38
rs1018742554 Chr.10: 102450891-102450891 on Build GRCh38
rs778107247 Chr.10: 102450984-102450984 on Build GRCh38
rs369188406 Chr.10: 102451089-102451089 on Build GRCh38
rs1027607878 Chr.10: 102450862-102450862 on Build GRCh38

View Full Product Details