Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 119043964-119044469 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF3A
Entrez Gene ID
8661
Gene Name
eukaryotic translation initiation factor 3 subunit A
Gene Aliases
EIF3, EIF3S10, P167, TIF32, eIF3-p170, eIF3-theta, p180, p185
UniGene
Hs.523299
Gene Chromosome Location
Chr.10: 119035029-119080822 on Build GRCh38
Transcripts
NM_003750.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7092643 c.2739G>A p.P913P Chr.10: 119044062-119044062 on Build GRCh38
4760701 c.2738C>T p.P913L Chr.10: 119044063-119044063 on Build GRCh38
6333790 c.2738C>A p.P913Q Chr.10: 119044063-119044063 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs943230240 Chr.10: 119044000-119044000 on Build GRCh38
rs375751079 Chr.10: 119044110-119044110 on Build GRCh38
rs943198923 Chr.10: 119044258-119044258 on Build GRCh38
rs1030094243 Chr.10: 119044230-119044230 on Build GRCh38
rs979011017 Chr.10: 119044273-119044273 on Build GRCh38
rs769951056 Chr.10: 119044147-119044147 on Build GRCh38
rs774540261 Chr.10: 119044247-119044247 on Build GRCh38
rs144646420 Chr.10: 119044062-119044062 on Build GRCh38
rs1022747581 Chr.10: 119044159-119044159 on Build GRCh38
rs374120155 Chr.10: 119044181-119044181 on Build GRCh38
rs767722019 Chr.10: 119044427-119044427 on Build GRCh38
rs562995534 Chr.10: 119044358-119044358 on Build GRCh38
rs192962147 Chr.10: 119044271-119044271 on Build GRCh38
rs921857749 Chr.10: 119044239-119044239 on Build GRCh38
rs188092854 Chr.10: 119044026-119044026 on Build GRCh38
rs116819503 Chr.10: 119044027-119044027 on Build GRCh38
rs1014028323 Chr.10: 119044135-119044135 on Build GRCh38
rs747698576 Chr.10: 119044071-119044071 on Build GRCh38
rs773141591 Chr.10: 119044175-119044175 on Build GRCh38
rs1017175449 Chr.10: 119044204-119044204 on Build GRCh38
rs200011068 Chr.10: 119044328-119044329 on Build GRCh38
rs532043228 Chr.10: 119044376-119044376 on Build GRCh38
rs1005761111 Chr.10: 119044198-119044198 on Build GRCh38
rs658352 Chr.10: 119044276-119044276 on Build GRCh38
rs1044164891 Chr.10: 119044379-119044379 on Build GRCh38
rs72509488 Chr.10: 119044395-119044396 on Build GRCh38
rs969177416 Chr.10: 119044422-119044422 on Build GRCh38
rs560317174 Chr.10: 119044304-119044304 on Build GRCh38
rs767388081 Chr.10: 119044162-119044162 on Build GRCh38
rs202038940 Chr.10: 119044055-119044055 on Build GRCh38
rs147876651 Chr.10: 119044114-119044114 on Build GRCh38
rs767476237 Chr.10: 119044040-119044040 on Build GRCh38
rs995645982 Chr.10: 119044318-119044318 on Build GRCh38
rs750172120 Chr.10: 119044048-119044048 on Build GRCh38
rs571506052 Chr.10: 119044439-119044439 on Build GRCh38
rs771329719 Chr.10: 119044115-119044115 on Build GRCh38
rs1001509967 Chr.10: 119044158-119044158 on Build GRCh38
rs1010612378 Chr.10: 119044399-119044399 on Build GRCh38
rs138249872 Chr.10: 119044352-119044352 on Build GRCh38
rs968581810 Chr.10: 119044233-119044233 on Build GRCh38
rs961938092 Chr.10: 119044209-119044209 on Build GRCh38
rs1001729555 Chr.10: 119044331-119044331 on Build GRCh38
rs550925504 Chr.10: 119044240-119044240 on Build GRCh38
rs76966260 Chr.10: 119044401-119044401 on Build GRCh38
rs149601446 Chr.10: 119044353-119044353 on Build GRCh38
rs1053202972 Chr.10: 119044036-119044036 on Build GRCh38
rs898876146 Chr.10: 119044309-119044309 on Build GRCh38
rs897313650 Chr.10: 119044076-119044076 on Build GRCh38
rs74438956 Chr.10: 119044224-119044224 on Build GRCh38
rs758932223 Chr.10: 119044108-119044108 on Build GRCh38
rs373332834 Chr.10: 119044160-119044160 on Build GRCh38
rs906010424 Chr.10: 119044191-119044191 on Build GRCh38
rs377638403 Chr.10: 119044015-119044015 on Build GRCh38
rs1036115357 Chr.10: 119044305-119044305 on Build GRCh38
rs397715112 Chr.10: 119044396-119044397 on Build GRCh38
rs3740553 Chr.10: 119044234-119044234 on Build GRCh38
rs3832705 Chr.10: 119044397-119044398 on Build GRCh38
rs775510275 Chr.10: 119044157-119044157 on Build GRCh38
rs1042952016 Chr.10: 119044017-119044017 on Build GRCh38
rs370541566 Chr.10: 119044173-119044173 on Build GRCh38
rs776924660 Chr.10: 119044009-119044009 on Build GRCh38
rs772653186 Chr.10: 119044330-119044330 on Build GRCh38
rs769492560 Chr.10: 119044063-119044063 on Build GRCh38
rs1027202114 Chr.10: 119044156-119044156 on Build GRCh38
rs913098612 Chr.10: 119044233-119044235 on Build GRCh38
rs934593785 Chr.10: 119044347-119044347 on Build GRCh38
rs755920785 Chr.10: 119044050-119044050 on Build GRCh38
rs746080611 Chr.10: 119044138-119044138 on Build GRCh38
rs776919253 Chr.10: 119044136-119044136 on Build GRCh38
rs765347246 Chr.10: 119044023-119044023 on Build GRCh38

View Full Product Details