Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 57389809-57390313 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRG2
Entrez Gene ID
5553
Gene Name
proteoglycan 2, pro eosinophil major basic protein
Gene Aliases
BMPG, MBP, MBP1, proMBP
UniGene
Hs.512633
Gene Chromosome Location
Chr.11: 57386787-57390657 on Build GRCh38
Transcripts
NM_001243245.2 NM_001302926.1 NM_001302927.1 NM_002728.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
928602 c.49C>A p.L17I Chr.11: 57389896-57389896 on Build GRCh38
3449699 c.9C>T p.L3L Chr.11: 57389936-57389936 on Build GRCh38
5372820 c.43T>C p.S15P Chr.11: 57389902-57389902 on Build GRCh38
1704017 c.11C>T p.P4L Chr.11: 57389934-57389934 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs762921892 Chr.11: 57390212-57390212 on Build GRCh38
rs1009031237 Chr.11: 57390003-57390003 on Build GRCh38
rs555058073 Chr.11: 57390185-57390185 on Build GRCh38
rs145669911 Chr.11: 57390282-57390282 on Build GRCh38
rs551186411 Chr.11: 57390075-57390075 on Build GRCh38
rs952942177 Chr.11: 57390065-57390065 on Build GRCh38
rs764821684 Chr.11: 57390272-57390272 on Build GRCh38
rs149147034 Chr.11: 57389963-57389963 on Build GRCh38
rs964909469 Chr.11: 57390153-57390153 on Build GRCh38
rs777335608 Chr.11: 57389922-57389922 on Build GRCh38
rs1021880996 Chr.11: 57390148-57390148 on Build GRCh38
rs752696273 Chr.11: 57389907-57389907 on Build GRCh38
rs185069188 Chr.11: 57390266-57390266 on Build GRCh38
rs572506753 Chr.11: 57390135-57390135 on Build GRCh38
rs138796776 Chr.11: 57390097-57390097 on Build GRCh38
rs375258376 Chr.11: 57389837-57389837 on Build GRCh38
rs180845023 Chr.11: 57390140-57390140 on Build GRCh38
rs189466317 Chr.11: 57390134-57390134 on Build GRCh38
rs868358738 Chr.11: 57389910-57389910 on Build GRCh38
rs753393646 Chr.11: 57389873-57389874 on Build GRCh38
rs111932870 Chr.11: 57390015-57390015 on Build GRCh38
rs911675803 Chr.11: 57390188-57390188 on Build GRCh38
rs1027300405 Chr.11: 57390043-57390043 on Build GRCh38
rs947889722 Chr.11: 57390053-57390053 on Build GRCh38
rs763325722 Chr.11: 57389887-57389887 on Build GRCh38
rs772466934 Chr.11: 57390206-57390206 on Build GRCh38
rs199751155 Chr.11: 57389992-57389992 on Build GRCh38
rs760976823 Chr.11: 57390225-57390225 on Build GRCh38
rs974529116 Chr.11: 57389914-57389914 on Build GRCh38
rs745655241 Chr.11: 57390124-57390124 on Build GRCh38
rs533292024 Chr.11: 57389872-57389872 on Build GRCh38
rs781351822 Chr.11: 57389943-57389943 on Build GRCh38
rs200430596 Chr.11: 57389905-57389905 on Build GRCh38
rs941683579 Chr.11: 57390255-57390255 on Build GRCh38
rs754407736 Chr.11: 57390003-57390004 on Build GRCh38
rs766686641 Chr.11: 57389890-57389890 on Build GRCh38
rs1039313555 Chr.11: 57390229-57390229 on Build GRCh38
rs765892063 Chr.11: 57389855-57389858 on Build GRCh38
rs201160242 Chr.11: 57390101-57390101 on Build GRCh38
rs142055764 Chr.11: 57390281-57390281 on Build GRCh38
rs773475523 Chr.11: 57389959-57389959 on Build GRCh38
rs755942321 Chr.11: 57389916-57389916 on Build GRCh38
rs490358 Chr.11: 57389932-57389932 on Build GRCh38
rs144962228 Chr.11: 57390035-57390035 on Build GRCh38
rs756948426 Chr.11: 57389933-57389933 on Build GRCh38
rs200225211 Chr.11: 57389987-57389987 on Build GRCh38
rs982726254 Chr.11: 57390115-57390115 on Build GRCh38
rs749360009 Chr.11: 57390160-57390160 on Build GRCh38
rs770420283 Chr.11: 57389949-57389949 on Build GRCh38
rs748744550 Chr.11: 57390198-57390198 on Build GRCh38
rs569334699 Chr.11: 57390088-57390088 on Build GRCh38
rs184588945 Chr.11: 57389956-57389956 on Build GRCh38
rs369791002 Chr.11: 57389964-57389964 on Build GRCh38
rs751442419 Chr.11: 57389893-57389893 on Build GRCh38
rs1002892513 Chr.11: 57390110-57390110 on Build GRCh38
rs769929622 Chr.11: 57389851-57389851 on Build GRCh38
rs745468154 Chr.11: 57389936-57389936 on Build GRCh38
rs774992208 Chr.11: 57390192-57390192 on Build GRCh38
rs759407925 Chr.11: 57389899-57389899 on Build GRCh38
rs759621132 Chr.11: 57390004-57390004 on Build GRCh38
rs895007496 Chr.11: 57390036-57390036 on Build GRCh38
rs764894676 Chr.11: 57390002-57390002 on Build GRCh38

View Full Product Details