Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 60517932-60518479 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MS4A13
Entrez Gene ID
503497
Gene Name
membrane spanning 4-domains A13
Gene Aliases
-
UniGene
Hs.523704
Gene Chromosome Location
Chr.11: 60515382-60543424 on Build GRCh38
Transcripts
NM_001012417.2 NM_001100909.1 NM_001278320.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
138003 c.76G>A p.G26R Chr.11: 60518159-60518159 on Build GRCh38
4034432 c.116T>A p.L39* Chr.11: 60518199-60518199 on Build GRCh38
4034431 c.20T>C p.I7T Chr.11: 60518103-60518103 on Build GRCh38
5612086 c.59_60AA>CT p.Q20P Chr.11: 60518142-60518143 on Build GRCh38
1241449 c.18C>G p.H6Q Chr.11: 60518101-60518101 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375739692 Chr.11: 60518188-60518188 on Build GRCh38
rs764775506 Chr.11: 60518126-60518126 on Build GRCh38
rs902713965 Chr.11: 60518379-60518379 on Build GRCh38
rs55756397 Chr.11: 60518091-60518091 on Build GRCh38
rs902649446 Chr.11: 60518060-60518060 on Build GRCh38
rs776429344 Chr.11: 60518200-60518200 on Build GRCh38
rs761626378 Chr.11: 60518218-60518218 on Build GRCh38
rs551088819 Chr.11: 60518267-60518267 on Build GRCh38
rs758927629 Chr.11: 60518368-60518368 on Build GRCh38
rs78448638 Chr.11: 60517962-60517962 on Build GRCh38
rs932563844 Chr.11: 60518447-60518447 on Build GRCh38
rs573487696 Chr.11: 60518001-60518001 on Build GRCh38
rs141622117 Chr.11: 60518164-60518164 on Build GRCh38
rs535747434 Chr.11: 60518419-60518419 on Build GRCh38
rs79002350 Chr.11: 60518330-60518330 on Build GRCh38
rs946371326 Chr.11: 60518084-60518084 on Build GRCh38
rs756465443 Chr.11: 60518140-60518140 on Build GRCh38
rs895553426 Chr.11: 60517995-60517995 on Build GRCh38
rs983513424 Chr.11: 60518193-60518193 on Build GRCh38
rs138544018 Chr.11: 60518154-60518154 on Build GRCh38
rs756841197 Chr.11: 60518123-60518123 on Build GRCh38
rs554601345 Chr.11: 60518421-60518421 on Build GRCh38
rs749953409 Chr.11: 60518227-60518227 on Build GRCh38
rs771942437 Chr.11: 60518189-60518189 on Build GRCh38
rs548477400 Chr.11: 60518172-60518172 on Build GRCh38
rs941826929 Chr.11: 60518211-60518211 on Build GRCh38
rs752593130 Chr.11: 60518193-60518193 on Build GRCh38
rs199805573 Chr.11: 60518221-60518221 on Build GRCh38
rs755498420 Chr.11: 60518133-60518133 on Build GRCh38
rs79855830 Chr.11: 60517961-60517961 on Build GRCh38
rs189542728 Chr.11: 60518317-60518317 on Build GRCh38
rs10897078 Chr.11: 60518055-60518055 on Build GRCh38
rs370921940 Chr.11: 60518195-60518195 on Build GRCh38
rs568332275 Chr.11: 60518191-60518191 on Build GRCh38
rs372542416 Chr.11: 60518056-60518056 on Build GRCh38
rs755376389 Chr.11: 60518040-60518040 on Build GRCh38
rs116120815 Chr.11: 60518307-60518307 on Build GRCh38
rs370691746 Chr.11: 60518296-60518296 on Build GRCh38
rs374551361 Chr.11: 60518245-60518245 on Build GRCh38
rs778876746 Chr.11: 60518189-60518189 on Build GRCh38
rs759143661 Chr.11: 60518282-60518282 on Build GRCh38
rs753186721 Chr.11: 60518257-60518257 on Build GRCh38
rs892095651 Chr.11: 60518059-60518059 on Build GRCh38
rs573401649 Chr.11: 60518370-60518370 on Build GRCh38
rs756435517 Chr.11: 60518262-60518262 on Build GRCh38
rs767944356 Chr.11: 60518256-60518256 on Build GRCh38
rs994794488 Chr.11: 60518219-60518219 on Build GRCh38
rs774273962 Chr.11: 60518067-60518067 on Build GRCh38
rs749949938 Chr.11: 60518130-60518130 on Build GRCh38
rs895461612 Chr.11: 60518343-60518343 on Build GRCh38
rs386754077 Chr.11: 60517961-60517962 on Build GRCh38
rs980944155 Chr.11: 60517986-60517986 on Build GRCh38
rs758240657 Chr.11: 60518208-60518208 on Build GRCh38
rs770935954 Chr.11: 60518062-60518062 on Build GRCh38
rs867904579 Chr.11: 60518348-60518348 on Build GRCh38
rs781478635 Chr.11: 60518044-60518044 on Build GRCh38
rs748506168 Chr.11: 60518135-60518135 on Build GRCh38
rs529663247 Chr.11: 60518076-60518076 on Build GRCh38
rs969599440 Chr.11: 60518285-60518285 on Build GRCh38
rs759455454 Chr.11: 60518077-60518077 on Build GRCh38
rs563052916 Chr.11: 60518093-60518093 on Build GRCh38
rs184120879 Chr.11: 60518163-60518163 on Build GRCh38
rs775291772 Chr.11: 60518287-60518287 on Build GRCh38
rs777344688 Chr.11: 60518257-60518258 on Build GRCh38
rs75015159 Chr.11: 60518045-60518045 on Build GRCh38
rs769171570 Chr.11: 60517981-60517981 on Build GRCh38
rs1001438489 Chr.11: 60518437-60518437 on Build GRCh38
rs566129943 Chr.11: 60518323-60518323 on Build GRCh38
rs913644431 Chr.11: 60518347-60518347 on Build GRCh38
rs760486787 Chr.11: 60518094-60518094 on Build GRCh38
rs753493897 Chr.11: 60518110-60518110 on Build GRCh38
rs10736706 Chr.11: 60518102-60518102 on Build GRCh38

View Full Product Details