Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61437597-61438117 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SDHAF2
Entrez Gene ID
54949
Gene Name
succinate dehydrogenase complex assembly factor 2
Gene Aliases
C11orf79, PGL2, SDH5
UniGene
Hs.313247
Gene Chromosome Location
Chr.11: 61430125-61446767 on Build GRCh38
Transcripts
NM_017841.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6445669 c.365C>T p.A122V Chr.11: 61438108-61438108 on Build GRCh38
6773773 c.295G>A p.E99K Chr.11: 61438038-61438038 on Build GRCh38
1355237 c.135G>T p.K45N Chr.11: 61437723-61437723 on Build GRCh38
689318 c.307A>G p.N103D Chr.11: 61438050-61438050 on Build GRCh38
6913672 c.363G>A p.W121* Chr.11: 61438106-61438106 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777442412 Chr.11: 61437686-61437686 on Build GRCh38
rs768896122 Chr.11: 61437866-61437866 on Build GRCh38
rs754291669 Chr.11: 61438064-61438064 on Build GRCh38
rs949388785 Chr.11: 61437784-61437784 on Build GRCh38
rs755511528 Chr.11: 61437818-61437818 on Build GRCh38
rs759472787 Chr.11: 61437668-61437668 on Build GRCh38
rs550980414 Chr.11: 61437963-61437963 on Build GRCh38
rs777927167 Chr.11: 61437846-61437846 on Build GRCh38
rs756917732 Chr.11: 61437688-61437688 on Build GRCh38
rs935462449 Chr.11: 61437864-61437864 on Build GRCh38
rs151040226 Chr.11: 61438093-61438093 on Build GRCh38
rs749030246 Chr.11: 61437879-61437879 on Build GRCh38
rs749618577 Chr.11: 61437989-61437989 on Build GRCh38
rs761956866 Chr.11: 61437624-61437624 on Build GRCh38
rs373951663 Chr.11: 61438012-61438012 on Build GRCh38
rs747508289 Chr.11: 61437710-61437710 on Build GRCh38
rs112606325 Chr.11: 61437871-61437871 on Build GRCh38
rs749431351 Chr.11: 61437733-61437735 on Build GRCh38
rs756514954 Chr.11: 61437975-61437975 on Build GRCh38
rs562958122 Chr.11: 61437746-61437746 on Build GRCh38
rs371469299 Chr.11: 61437860-61437860 on Build GRCh38
rs757670376 Chr.11: 61438070-61438070 on Build GRCh38
rs201520416 Chr.11: 61438032-61438032 on Build GRCh38
rs868016844 Chr.11: 61437697-61437697 on Build GRCh38
rs753325226 Chr.11: 61437687-61437687 on Build GRCh38
rs752462796 Chr.11: 61437782-61437782 on Build GRCh38
rs931576045 Chr.11: 61437919-61437919 on Build GRCh38
rs536680634 Chr.11: 61437973-61437974 on Build GRCh38
rs112687476 Chr.11: 61437859-61437859 on Build GRCh38
rs776339422 Chr.11: 61437880-61437880 on Build GRCh38
rs745989557 Chr.11: 61438002-61438002 on Build GRCh38
rs750979204 Chr.11: 61438058-61438058 on Build GRCh38
rs760529473 Chr.11: 61438002-61438005 on Build GRCh38
rs111402137 Chr.11: 61437727-61437727 on Build GRCh38
rs952830677 Chr.11: 61437684-61437684 on Build GRCh38
rs982336783 Chr.11: 61437836-61437836 on Build GRCh38
rs758481098 Chr.11: 61438077-61438077 on Build GRCh38
rs751530375 Chr.11: 61437664-61437664 on Build GRCh38
rs368945911 Chr.11: 61437721-61437721 on Build GRCh38
rs897080048 Chr.11: 61438011-61438011 on Build GRCh38
rs376941346 Chr.11: 61437882-61437882 on Build GRCh38
rs756825771 Chr.11: 61437795-61437795 on Build GRCh38
rs746968831 Chr.11: 61438008-61438008 on Build GRCh38
rs747994812 Chr.11: 61438095-61438095 on Build GRCh38
rs1002894711 Chr.11: 61437631-61437631 on Build GRCh38
rs755605816 Chr.11: 61438029-61438029 on Build GRCh38
rs969049910 Chr.11: 61437726-61437726 on Build GRCh38
rs745899580 Chr.11: 61437863-61437863 on Build GRCh38
rs769950627 Chr.11: 61437741-61437741 on Build GRCh38
rs371977724 Chr.11: 61437784-61437784 on Build GRCh38
rs987209720 Chr.11: 61437870-61437870 on Build GRCh38
rs771133722 Chr.11: 61437751-61437751 on Build GRCh38
rs767970905 Chr.11: 61437967-61437967 on Build GRCh38
rs779335034 Chr.11: 61438073-61438073 on Build GRCh38
rs774508076 Chr.11: 61437753-61437753 on Build GRCh38
rs184314049 Chr.11: 61438052-61438052 on Build GRCh38
rs780593356 Chr.11: 61437705-61437705 on Build GRCh38
rs535627239 Chr.11: 61438063-61438063 on Build GRCh38
rs147928545 Chr.11: 61438016-61438016 on Build GRCh38
rs753554501 Chr.11: 61438048-61438049 on Build GRCh38
rs780194184 Chr.11: 61438080-61438080 on Build GRCh38
rs753474292 Chr.11: 61437794-61437794 on Build GRCh38
rs773466310 Chr.11: 61437744-61437744 on Build GRCh38
rs112578506 Chr.11: 61437754-61437754 on Build GRCh38
rs766468929 Chr.11: 61437654-61437654 on Build GRCh38
rs200911550 Chr.11: 61437640-61437640 on Build GRCh38
rs770012985 Chr.11: 61437633-61437633 on Build GRCh38
rs920160582 Chr.11: 61437913-61437913 on Build GRCh38
rs773213740 Chr.11: 61437982-61437982 on Build GRCh38
rs772177974 Chr.11: 61438003-61438003 on Build GRCh38
rs368333259 Chr.11: 61437973-61437973 on Build GRCh38
rs775356110 Chr.11: 61438005-61438005 on Build GRCh38
rs376560419 Chr.11: 61437827-61437827 on Build GRCh38
rs876658350 Chr.11: 61438090-61438090 on Build GRCh38
rs757580722 Chr.11: 61437993-61437993 on Build GRCh38
rs778397152 Chr.11: 61437804-61437804 on Build GRCh38
rs761478064 Chr.11: 61437881-61437881 on Build GRCh38
rs548371183 Chr.11: 61437930-61437930 on Build GRCh38
rs774626235 Chr.11: 61437865-61437866 on Build GRCh38
rs143087398 Chr.11: 61437932-61437932 on Build GRCh38
rs191513932 Chr.11: 61437651-61437651 on Build GRCh38
rs762441285 Chr.11: 61438037-61438037 on Build GRCh38
rs113307092 Chr.11: 61437780-61437780 on Build GRCh38
rs1007869247 Chr.11: 61437937-61437937 on Build GRCh38
rs144867876 Chr.11: 61437685-61437685 on Build GRCh38
rs748505431 Chr.11: 61437729-61437729 on Build GRCh38
rs201884652 Chr.11: 61437874-61437874 on Build GRCh38
rs752477657 Chr.11: 61437681-61437681 on Build GRCh38
rs532255760 Chr.11: 61437793-61437793 on Build GRCh38
rs756492007 Chr.11: 61437843-61437843 on Build GRCh38
rs778298433 Chr.11: 61437983-61437983 on Build GRCh38
rs764919936 Chr.11: 61438031-61438031 on Build GRCh38
rs113560320 Chr.11: 61437820-61437820 on Build GRCh38
rs771217603 Chr.11: 61437852-61437852 on Build GRCh38
rs779242759 Chr.11: 61437996-61437996 on Build GRCh38
rs747022200 Chr.11: 61438084-61438084 on Build GRCh38
rs767436409 Chr.11: 61437759-61437759 on Build GRCh38
rs749527870 Chr.11: 61437849-61437849 on Build GRCh38
rs772204945 Chr.11: 61437976-61437977 on Build GRCh38
rs766035180 Chr.11: 61438006-61438006 on Build GRCh38
rs140191819 Chr.11: 61438062-61438062 on Build GRCh38
rs145616631 Chr.11: 61438074-61438074 on Build GRCh38
rs760482747 Chr.11: 61437786-61437786 on Build GRCh38
rs759556749 Chr.11: 61437757-61437757 on Build GRCh38
rs879647 Chr.11: 61437962-61437962 on Build GRCh38
rs781702725 Chr.11: 61437823-61437823 on Build GRCh38
rs749887667 Chr.11: 61437810-61437810 on Build GRCh38

View Full Product Details