Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62193307-62193802 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCGB1D1
Entrez Gene ID
10648
Gene Name
secretoglobin family 1D member 1
Gene Aliases
LIPA, LPHA, LPNA
UniGene
Hs.202686
Gene Chromosome Location
Chr.11: 62190238-62193537 on Build GRCh38
Transcripts
NM_006552.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
147260 c.253G>A p.A85T Chr.11: 62193408-62193408 on Build GRCh38
929579 c.258G>A p.E86E Chr.11: 62193413-62193413 on Build GRCh38
1676092 c.269G>A p.R90H Chr.11: 62193424-62193424 on Build GRCh38
1741695 c.244-7A>G p.? Chr.11: 62193392-62193392 on Build GRCh38
689253 c.250A>G p.I84V Chr.11: 62193405-62193405 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs969676894 Chr.11: 62193676-62193676 on Build GRCh38
rs766448972 Chr.11: 62193424-62193424 on Build GRCh38
rs772437701 Chr.11: 62193361-62193361 on Build GRCh38
rs1051367910 Chr.11: 62193551-62193551 on Build GRCh38
rs1023334058 Chr.11: 62193655-62193655 on Build GRCh38
rs7951859 Chr.11: 62193337-62193337 on Build GRCh38
rs748460592 Chr.11: 62193356-62193356 on Build GRCh38
rs755231866 Chr.11: 62193348-62193348 on Build GRCh38
rs565996668 Chr.11: 62193709-62193709 on Build GRCh38
rs530995185 Chr.11: 62193561-62193561 on Build GRCh38
rs187897966 Chr.11: 62193644-62193644 on Build GRCh38
rs778743170 Chr.11: 62193355-62193356 on Build GRCh38
rs774162033 Chr.11: 62193393-62193393 on Build GRCh38
rs903382575 Chr.11: 62193554-62193554 on Build GRCh38
rs919734962 Chr.11: 62193693-62193693 on Build GRCh38
rs551105532 Chr.11: 62193573-62193573 on Build GRCh38
rs369847324 Chr.11: 62193448-62193448 on Build GRCh38
rs141461828 Chr.11: 62193399-62193399 on Build GRCh38
rs116716071 Chr.11: 62193475-62193475 on Build GRCh38
rs753978316 Chr.11: 62193349-62193350 on Build GRCh38
rs973988476 Chr.11: 62193681-62193681 on Build GRCh38
rs1008630504 Chr.11: 62193435-62193435 on Build GRCh38
rs765945771 Chr.11: 62193541-62193541 on Build GRCh38
rs2232942 Chr.11: 62193507-62193507 on Build GRCh38
rs374753073 Chr.11: 62193451-62193451 on Build GRCh38
rs373191839 Chr.11: 62193425-62193425 on Build GRCh38
rs1038603702 Chr.11: 62193378-62193378 on Build GRCh38
rs377082018 Chr.11: 62193515-62193515 on Build GRCh38
rs768033512 Chr.11: 62193461-62193461 on Build GRCh38
rs758390063 Chr.11: 62193540-62193542 on Build GRCh38
rs577027339 Chr.11: 62193423-62193423 on Build GRCh38
rs368993364 Chr.11: 62193377-62193377 on Build GRCh38
rs560035602 Chr.11: 62193452-62193452 on Build GRCh38
rs774682190 Chr.11: 62193433-62193433 on Build GRCh38
rs959382091 Chr.11: 62193750-62193750 on Build GRCh38
rs1047196276 Chr.11: 62193428-62193428 on Build GRCh38
rs774907928 Chr.11: 62193369-62193369 on Build GRCh38
rs547754674 Chr.11: 62193738-62193738 on Build GRCh38
rs900569169 Chr.11: 62193523-62193524 on Build GRCh38
rs992375050 Chr.11: 62193771-62193771 on Build GRCh38
rs755033281 Chr.11: 62193352-62193354 on Build GRCh38
rs762586249 Chr.11: 62193372-62193372 on Build GRCh38
rs773697623 Chr.11: 62193363-62193363 on Build GRCh38
rs767486935 Chr.11: 62193420-62193420 on Build GRCh38
rs763791852 Chr.11: 62193387-62193387 on Build GRCh38
rs929806777 Chr.11: 62193696-62193696 on Build GRCh38
rs370083682 Chr.11: 62193473-62193473 on Build GRCh38
rs375732265 Chr.11: 62193445-62193445 on Build GRCh38
rs536771504 Chr.11: 62193364-62193364 on Build GRCh38
rs562089258 Chr.11: 62193509-62193509 on Build GRCh38
rs2232941 Chr.11: 62193355-62193355 on Build GRCh38
rs117229148 Chr.11: 62193737-62193737 on Build GRCh38
rs754622641 Chr.11: 62193626-62193626 on Build GRCh38
rs1015426289 Chr.11: 62193578-62193578 on Build GRCh38
rs748153624 Chr.11: 62193400-62193401 on Build GRCh38
rs944174757 Chr.11: 62193778-62193778 on Build GRCh38
rs936629880 Chr.11: 62193514-62193514 on Build GRCh38
rs545978030 Chr.11: 62193553-62193553 on Build GRCh38
rs761617791 Chr.11: 62193476-62193476 on Build GRCh38
rs758754645 Chr.11: 62193434-62193434 on Build GRCh38
rs748751797 Chr.11: 62193468-62193468 on Build GRCh38
rs552175513 Chr.11: 62193658-62193658 on Build GRCh38
rs771885129 Chr.11: 62193440-62193440 on Build GRCh38
rs1042630782 Chr.11: 62193501-62193501 on Build GRCh38
rs955450371 Chr.11: 62193362-62193362 on Build GRCh38
rs973796180 Chr.11: 62193375-62193375 on Build GRCh38
rs117865826 Chr.11: 62193408-62193408 on Build GRCh38
rs918089178 Chr.11: 62193780-62193780 on Build GRCh38
rs779047079 Chr.11: 62193430-62193430 on Build GRCh38
rs962319026 Chr.11: 62193367-62193367 on Build GRCh38
rs768288434 Chr.11: 62193470-62193470 on Build GRCh38
rs992233470 Chr.11: 62193647-62193647 on Build GRCh38
rs563424682 Chr.11: 62193604-62193604 on Build GRCh38

View Full Product Details