Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 63946609-63947108 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NAA40
Entrez Gene ID
79829
Gene Name
N(alpha)-acetyltransferase 40, NatD catalytic subunit
Gene Aliases
NAT11, PATT1
UniGene
Hs.523753
Gene Chromosome Location
Chr.11: 63938970-63957322 on Build GRCh38
Transcripts
NM_001300800.1 NM_024771.3 XM_006718689.3 XM_011545254.2 XM_011545255.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2162851 c.153C>T p.N51N Chr.11: 63947001-63947001 on Build GRCh38
3451350 c.107G>A p.G36E Chr.11: 63946955-63946955 on Build GRCh38
1492661 c.130G>A p.V44M Chr.11: 63946978-63946978 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs527295072 Chr.11: 63946904-63946904 on Build GRCh38
rs966428225 Chr.11: 63946811-63946811 on Build GRCh38
rs760582058 Chr.11: 63946843-63946843 on Build GRCh38
rs776574821 Chr.11: 63946914-63946914 on Build GRCh38
rs927901155 Chr.11: 63946820-63946820 on Build GRCh38
rs568918338 Chr.11: 63946691-63946691 on Build GRCh38
rs578137147 Chr.11: 63946633-63946633 on Build GRCh38
rs761175873 Chr.11: 63946815-63946815 on Build GRCh38
rs569949857 Chr.11: 63947040-63947040 on Build GRCh38
rs763227684 Chr.11: 63946941-63946941 on Build GRCh38
rs1037812165 Chr.11: 63946900-63946900 on Build GRCh38
rs1010688268 Chr.11: 63946774-63946774 on Build GRCh38
rs747805431 Chr.11: 63946907-63946907 on Build GRCh38
rs757942241 Chr.11: 63946648-63946648 on Build GRCh38
rs966577337 Chr.11: 63946657-63946657 on Build GRCh38
rs11231651 Chr.11: 63946950-63946950 on Build GRCh38
rs140180729 Chr.11: 63946670-63946670 on Build GRCh38
rs770797218 Chr.11: 63946913-63946913 on Build GRCh38
rs752487246 Chr.11: 63946940-63946940 on Build GRCh38
rs150307692 Chr.11: 63946745-63946745 on Build GRCh38
rs993693907 Chr.11: 63946658-63946658 on Build GRCh38
rs763492521 Chr.11: 63946758-63946759 on Build GRCh38
rs757422545 Chr.11: 63946962-63946962 on Build GRCh38
rs772638094 Chr.11: 63946840-63946840 on Build GRCh38
rs757838142 Chr.11: 63946869-63946869 on Build GRCh38
rs762663667 Chr.11: 63947050-63947050 on Build GRCh38
rs778105492 Chr.11: 63946772-63946772 on Build GRCh38
rs981730707 Chr.11: 63946732-63946732 on Build GRCh38
rs934462902 Chr.11: 63946746-63946746 on Build GRCh38
rs915951580 Chr.11: 63946700-63946700 on Build GRCh38
rs910988144 Chr.11: 63947074-63947074 on Build GRCh38
rs771750598 Chr.11: 63946992-63946992 on Build GRCh38
rs752995073 Chr.11: 63946852-63946852 on Build GRCh38
rs1023063235 Chr.11: 63946924-63946924 on Build GRCh38
rs4980500 Chr.11: 63946847-63946847 on Build GRCh38
rs750995420 Chr.11: 63946975-63946975 on Build GRCh38
rs1017745378 Chr.11: 63946766-63946766 on Build GRCh38
rs765395229 Chr.11: 63946851-63946851 on Build GRCh38
rs914308227 Chr.11: 63946770-63946770 on Build GRCh38
rs776274621 Chr.11: 63946796-63946796 on Build GRCh38
rs199849714 Chr.11: 63946903-63946903 on Build GRCh38
rs764402608 Chr.11: 63946850-63946850 on Build GRCh38
rs947227898 Chr.11: 63947068-63947068 on Build GRCh38
rs560383199 Chr.11: 63946689-63946689 on Build GRCh38
rs753564667 Chr.11: 63946845-63946845 on Build GRCh38
rs780441867 Chr.11: 63946983-63946983 on Build GRCh38
rs531918277 Chr.11: 63946777-63946777 on Build GRCh38
rs567264827 Chr.11: 63946910-63946910 on Build GRCh38
rs778387198 Chr.11: 63946906-63946906 on Build GRCh38
rs776160425 Chr.11: 63947019-63947019 on Build GRCh38
rs758592648 Chr.11: 63946863-63946863 on Build GRCh38
rs756725489 Chr.11: 63946980-63946980 on Build GRCh38
rs548158062 Chr.11: 63946868-63946868 on Build GRCh38
rs370163040 Chr.11: 63946963-63946963 on Build GRCh38
rs897861114 Chr.11: 63946672-63946672 on Build GRCh38
rs780746195 Chr.11: 63946947-63946947 on Build GRCh38
rs550030510 Chr.11: 63947020-63947020 on Build GRCh38
rs985144390 Chr.11: 63947073-63947073 on Build GRCh38
rs766129845 Chr.11: 63946677-63946677 on Build GRCh38
rs368956834 Chr.11: 63947014-63947014 on Build GRCh38
rs777126069 Chr.11: 63946784-63946784 on Build GRCh38
rs943727937 Chr.11: 63946838-63946838 on Build GRCh38
rs970432429 Chr.11: 63946971-63946971 on Build GRCh38
rs767084669 Chr.11: 63946835-63946835 on Build GRCh38
rs747523982 Chr.11: 63946773-63946773 on Build GRCh38
rs538974147 Chr.11: 63947080-63947080 on Build GRCh38
rs769477305 Chr.11: 63947002-63947002 on Build GRCh38
rs942088446 Chr.11: 63946646-63946646 on Build GRCh38
rs374448283 Chr.11: 63947001-63947001 on Build GRCh38
rs758528468 Chr.11: 63946769-63946769 on Build GRCh38
rs762030423 Chr.11: 63946941-63946941 on Build GRCh38
rs777891116 Chr.11: 63946866-63946866 on Build GRCh38
rs765053069 Chr.11: 63946936-63946936 on Build GRCh38
rs1052045464 Chr.11: 63946741-63946741 on Build GRCh38
rs137857294 Chr.11: 63946984-63946984 on Build GRCh38
rs759288365 Chr.11: 63946923-63946923 on Build GRCh38
rs755190876 Chr.11: 63946718-63946718 on Build GRCh38
rs1026379809 Chr.11: 63946663-63946663 on Build GRCh38
rs769680130 Chr.11: 63947034-63947034 on Build GRCh38
rs751618856 Chr.11: 63946956-63946956 on Build GRCh38
rs774062329 Chr.11: 63946812-63946812 on Build GRCh38
rs562812641 Chr.11: 63946751-63946751 on Build GRCh38
rs1021121808 Chr.11: 63946805-63946805 on Build GRCh38
rs766359736 Chr.11: 63946844-63946844 on Build GRCh38
rs746299022 Chr.11: 63946791-63946791 on Build GRCh38
rs774807395 Chr.11: 63946935-63946937 on Build GRCh38
rs367619278 Chr.11: 63946908-63946908 on Build GRCh38
rs748119661 Chr.11: 63946761-63946761 on Build GRCh38

View Full Product Details