Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64317996-64318488 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRDX5
Entrez Gene ID
25824
Gene Name
peroxiredoxin 5
Gene Aliases
ACR1, AOEB166, B166, HEL-S-55, PLP, PMP20, PRDX6, PRXV, SBBI10, prx-V
UniGene
Hs.502823
Gene Chromosome Location
Chr.11: 64318088-64321823 on Build GRCh38
Transcripts
NM_012094.4 NM_181651.2 NM_181652.2 XM_005273882.4 NM_001286082.1 NM_001286084.1 NM_016404.2

Gene Symbol
TRMT112
Entrez Gene ID
51504
Gene Name
tRNA methyltransferase 11-2 homolog (S. cerevisiae)
Gene Aliases
HSPC152, HSPC170, TRM112, TRMT11-2, hTrm112
UniGene
Hs.333579
Gene Chromosome Location
Chr.11: 64316492-64318084 on Build GRCh38
Transcripts
NM_012094.4 NM_181651.2 NM_181652.2 XM_005273882.4 NM_001286082.1 NM_001286084.1 NM_016404.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4574620 c.47T>C p.I16T Chr.11: 64318262-64318262 on Build GRCh38
4541419 c.29G>A p.R10K Chr.11: 64318244-64318244 on Build GRCh38
1355679 c.77C>T p.A26V Chr.11: 64318292-64318292 on Build GRCh38
429533 c.98A>G p.Y33C Chr.11: 64318313-64318313 on Build GRCh38
1322273 c.106G>A p.G36R Chr.11: 64318321-64318321 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1016607893 Chr.11: 64318138-64318138 on Build GRCh38
rs546358756 Chr.11: 64318338-64318338 on Build GRCh38
rs939149289 Chr.11: 64318241-64318241 on Build GRCh38
rs1025284913 Chr.11: 64318155-64318155 on Build GRCh38
rs776283715 Chr.11: 64318399-64318399 on Build GRCh38
rs1056746543 Chr.11: 64318114-64318114 on Build GRCh38
rs1039103217 Chr.11: 64318078-64318078 on Build GRCh38
rs911910557 Chr.11: 64318063-64318063 on Build GRCh38
rs764027175 Chr.11: 64318353-64318353 on Build GRCh38
rs563893744 Chr.11: 64318296-64318296 on Build GRCh38
rs771579508 Chr.11: 64318166-64318166 on Build GRCh38
rs915221912 Chr.11: 64318215-64318215 on Build GRCh38
rs748462954 Chr.11: 64318393-64318393 on Build GRCh38
rs1038272111 Chr.11: 64318441-64318441 on Build GRCh38
rs749893679 Chr.11: 64318424-64318434 on Build GRCh38
rs767112440 Chr.11: 64318206-64318206 on Build GRCh38
rs749654926 Chr.11: 64318308-64318308 on Build GRCh38
rs936012017 Chr.11: 64318142-64318153 on Build GRCh38
rs1007973441 Chr.11: 64318367-64318367 on Build GRCh38
rs755560689 Chr.11: 64318210-64318210 on Build GRCh38
rs962476920 Chr.11: 64318143-64318143 on Build GRCh38
rs781169297 Chr.11: 64318289-64318289 on Build GRCh38
rs752749547 Chr.11: 64318345-64318345 on Build GRCh38
rs372637504 Chr.11: 64318259-64318259 on Build GRCh38
rs148589018 Chr.11: 64318292-64318292 on Build GRCh38
rs963835814 Chr.11: 64318382-64318382 on Build GRCh38
rs573251178 Chr.11: 64318107-64318107 on Build GRCh38
rs943333293 Chr.11: 64318076-64318076 on Build GRCh38
rs754924766 Chr.11: 64318229-64318229 on Build GRCh38
rs925303403 Chr.11: 64318086-64318086 on Build GRCh38
rs911072358 Chr.11: 64318060-64318060 on Build GRCh38
rs376203241 Chr.11: 64318404-64318404 on Build GRCh38
rs978210752 Chr.11: 64318389-64318389 on Build GRCh38
rs756150774 Chr.11: 64318377-64318377 on Build GRCh38
rs7938623 Chr.11: 64318313-64318313 on Build GRCh38
rs759109432 Chr.11: 64318401-64318401 on Build GRCh38
rs929596894 Chr.11: 64318458-64318458 on Build GRCh38
rs950924466 Chr.11: 64318147-64318147 on Build GRCh38
rs769297245 Chr.11: 64318430-64318430 on Build GRCh38
rs758202043 Chr.11: 64318277-64318277 on Build GRCh38
rs760310200 Chr.11: 64318269-64318269 on Build GRCh38
rs539515053 Chr.11: 64318084-64318084 on Build GRCh38
rs905393225 Chr.11: 64318145-64318145 on Build GRCh38
rs919402272 Chr.11: 64318049-64318049 on Build GRCh38
rs557025404 Chr.11: 64318055-64318055 on Build GRCh38
rs553564429 Chr.11: 64318160-64318160 on Build GRCh38
rs973636594 Chr.11: 64318047-64318047 on Build GRCh38
rs1016662005 Chr.11: 64318019-64318019 on Build GRCh38
rs757529407 Chr.11: 64318288-64318288 on Build GRCh38
rs760363588 Chr.11: 64318317-64318317 on Build GRCh38
rs761278277 Chr.11: 64318290-64318291 on Build GRCh38
rs761979554 Chr.11: 64318146-64318146 on Build GRCh38
rs573099270 Chr.11: 64318226-64318226 on Build GRCh38
rs777214046 Chr.11: 64318170-64318170 on Build GRCh38
rs761642493 Chr.11: 64318411-64318411 on Build GRCh38
rs140015234 Chr.11: 64318346-64318346 on Build GRCh38
rs774678310 Chr.11: 64318314-64318314 on Build GRCh38
rs896542446 Chr.11: 64318128-64318128 on Build GRCh38
rs11551856 Chr.11: 64318349-64318349 on Build GRCh38
rs978559577 Chr.11: 64318232-64318232 on Build GRCh38
rs146856761 Chr.11: 64318420-64318420 on Build GRCh38
rs994310512 Chr.11: 64318144-64318144 on Build GRCh38
rs192518475 Chr.11: 64318440-64318440 on Build GRCh38
rs771199353 Chr.11: 64318245-64318245 on Build GRCh38
rs371507976 Chr.11: 64318264-64318264 on Build GRCh38
rs1052442141 Chr.11: 64318108-64318108 on Build GRCh38
rs770303491 Chr.11: 64318256-64318256 on Build GRCh38
rs529087378 Chr.11: 64318418-64318418 on Build GRCh38
rs1019065444 Chr.11: 64318321-64318321 on Build GRCh38
rs766981427 Chr.11: 64318318-64318320 on Build GRCh38
rs760081876 Chr.11: 64318177-64318177 on Build GRCh38
rs767636968 Chr.11: 64318413-64318413 on Build GRCh38
rs200384441 Chr.11: 64318421-64318421 on Build GRCh38
rs143054032 Chr.11: 64318303-64318303 on Build GRCh38
rs751745501 Chr.11: 64318117-64318117 on Build GRCh38
rs143723178 Chr.11: 64318381-64318381 on Build GRCh38
rs750481771 Chr.11: 64318372-64318372 on Build GRCh38
rs942159937 Chr.11: 64318293-64318293 on Build GRCh38
rs886617487 Chr.11: 64318310-64318310 on Build GRCh38
rs201329476 Chr.11: 64318209-64318209 on Build GRCh38
rs7941847 Chr.11: 64318466-64318466 on Build GRCh38
rs747092163 Chr.11: 64318239-64318239 on Build GRCh38
rs369360548 Chr.11: 64318181-64318181 on Build GRCh38
rs779991462 Chr.11: 64318380-64318380 on Build GRCh38
rs562055323 Chr.11: 64318237-64318237 on Build GRCh38
rs560433484 Chr.11: 64318386-64318386 on Build GRCh38
rs751417950 Chr.11: 64318408-64318408 on Build GRCh38
rs764280746 Chr.11: 64318112-64318112 on Build GRCh38
rs765091459 Chr.11: 64318343-64318343 on Build GRCh38
rs751210390 Chr.11: 64318249-64318275 on Build GRCh38
rs1011956005 Chr.11: 64318121-64318121 on Build GRCh38
rs1042245068 Chr.11: 64318097-64318097 on Build GRCh38
rs779282514 Chr.11: 64318387-64318387 on Build GRCh38
rs759029280 Chr.11: 64318339-64318339 on Build GRCh38
rs777708768 Chr.11: 64318279-64318279 on Build GRCh38
rs765576811 Chr.11: 64318194-64318194 on Build GRCh38
rs955109438 Chr.11: 64318157-64318157 on Build GRCh38
rs988098997 Chr.11: 64318059-64318059 on Build GRCh38
rs149410533 Chr.11: 64318254-64318254 on Build GRCh38
rs764078661 Chr.11: 64318407-64318407 on Build GRCh38
rs368539290 Chr.11: 64318238-64318238 on Build GRCh38
rs201569388 Chr.11: 64318255-64318255 on Build GRCh38
rs769406922 Chr.11: 64318403-64318403 on Build GRCh38
rs962446162 Chr.11: 64318036-64318036 on Build GRCh38
rs898162032 Chr.11: 64318448-64318448 on Build GRCh38
rs964258104 Chr.11: 64318198-64318198 on Build GRCh38
rs577288827 Chr.11: 64318082-64318082 on Build GRCh38
rs987743524 Chr.11: 64318156-64318156 on Build GRCh38
rs779537653 Chr.11: 64318213-64318213 on Build GRCh38
rs368851243 Chr.11: 64318416-64318416 on Build GRCh38
rs1022265075 Chr.11: 64318354-64318354 on Build GRCh38
rs772782767 Chr.11: 64318267-64318267 on Build GRCh38
rs942617519 Chr.11: 64318062-64318062 on Build GRCh38
rs755497893 Chr.11: 64318419-64318419 on Build GRCh38
rs375334607 Chr.11: 64318029-64318029 on Build GRCh38
rs374200354 Chr.11: 64318270-64318270 on Build GRCh38
rs373094666 Chr.11: 64318422-64318422 on Build GRCh38
rs750723219 Chr.11: 64318414-64318414 on Build GRCh38
rs1052830049 Chr.11: 64318020-64318020 on Build GRCh38
rs371600628 Chr.11: 64318368-64318368 on Build GRCh38
rs946797783 Chr.11: 64318089-64318089 on Build GRCh38
rs955139141 Chr.11: 64318048-64318048 on Build GRCh38
rs753082757 Chr.11: 64318197-64318197 on Build GRCh38
rs761408760 Chr.11: 64318203-64318203 on Build GRCh38
rs1003482317 Chr.11: 64318111-64318111 on Build GRCh38
rs745582013 Chr.11: 64318426-64318426 on Build GRCh38
rs369628643 Chr.11: 64318283-64318283 on Build GRCh38
rs770507349 Chr.11: 64318394-64318394 on Build GRCh38
rs993766702 Chr.11: 64318152-64318152 on Build GRCh38
rs781426413 Chr.11: 64318371-64318371 on Build GRCh38
rs202108463 Chr.11: 64318230-64318230 on Build GRCh38
rs765980723 Chr.11: 64318327-64318327 on Build GRCh38
rs144769220 Chr.11: 64318276-64318276 on Build GRCh38
rs1025775108 Chr.11: 64318309-64318309 on Build GRCh38
rs935278655 Chr.11: 64318092-64318092 on Build GRCh38

View Full Product Details