Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82459522-82460083 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NARF
Entrez Gene ID
26502
Gene Name
nuclear prelamin A recognition factor
Gene Aliases
IOP2
UniGene
Hs.256526
Gene Chromosome Location
Chr.17: 82458184-82490537 on Build GRCh38
Transcripts
NM_001038618.2 NM_001083608.1 NM_012336.3 NM_031968.2 XM_005256340.3 XM_006722278.3 XM_011523541.2 XM_011523543.2 XM_011523544.2 XM_017024449.1 XM_017024450.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
708579 c.39G>T p.K13N Chr.17: 82460003-82460003 on Build GRCh38
5780314 c.96G>C p.Q32H Chr.17: 82460060-82460060 on Build GRCh38
2806833 c.84G>A p.P28P Chr.17: 82460048-82460048 on Build GRCh38
5118452 c.63G>T p.E21D Chr.17: 82460027-82460027 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs886737264 Chr.17: 82459698-82459698 on Build GRCh38
rs998927086 Chr.17: 82459954-82459954 on Build GRCh38
rs968788221 Chr.17: 82459758-82459758 on Build GRCh38
rs72859113 Chr.17: 82459567-82459567 on Build GRCh38
rs149245781 Chr.17: 82459686-82459686 on Build GRCh38
rs762569549 Chr.17: 82459870-82459873 on Build GRCh38
rs186734283 Chr.17: 82459891-82459891 on Build GRCh38
rs770313123 Chr.17: 82459951-82459951 on Build GRCh38
rs143558028 Chr.17: 82459919-82459919 on Build GRCh38
rs748630510 Chr.17: 82460040-82460040 on Build GRCh38
rs995229323 Chr.17: 82459840-82459840 on Build GRCh38
rs142068937 Chr.17: 82459995-82459995 on Build GRCh38
rs368518853 Chr.17: 82460039-82460039 on Build GRCh38
rs138884508 Chr.17: 82459992-82459992 on Build GRCh38
rs78720068 Chr.17: 82459853-82459853 on Build GRCh38
rs533946277 Chr.17: 82459749-82459749 on Build GRCh38
rs374424351 Chr.17: 82459950-82459950 on Build GRCh38
rs1015236555 Chr.17: 82459811-82459811 on Build GRCh38
rs779374834 Chr.17: 82459949-82459949 on Build GRCh38
rs1008937860 Chr.17: 82459699-82459699 on Build GRCh38
rs943708057 Chr.17: 82459869-82459869 on Build GRCh38
rs766527529 Chr.17: 82460027-82460027 on Build GRCh38
rs796964335 Chr.17: 82459872-82459872 on Build GRCh38
rs977435964 Chr.17: 82459769-82459769 on Build GRCh38
rs144725155 Chr.17: 82460023-82460023 on Build GRCh38
rs762778688 Chr.17: 82459987-82459987 on Build GRCh38
rs549876246 Chr.17: 82459676-82459676 on Build GRCh38
rs373163516 Chr.17: 82459576-82459576 on Build GRCh38
rs913219407 Chr.17: 82459667-82459667 on Build GRCh38
rs944058876 Chr.17: 82459652-82459652 on Build GRCh38
rs1031671861 Chr.17: 82459776-82459776 on Build GRCh38
rs568496348 Chr.17: 82459617-82459617 on Build GRCh38
rs917820373 Chr.17: 82459815-82459815 on Build GRCh38
rs997480639 Chr.17: 82459947-82459947 on Build GRCh38
rs954237325 Chr.17: 82459849-82459849 on Build GRCh38
rs558459268 Chr.17: 82459784-82459784 on Build GRCh38
rs1021725504 Chr.17: 82459738-82459738 on Build GRCh38
rs772812562 Chr.17: 82459972-82459972 on Build GRCh38
rs181683539 Chr.17: 82459664-82459664 on Build GRCh38
rs973774969 Chr.17: 82459560-82459560 on Build GRCh38
rs192005315 Chr.17: 82459656-82459656 on Build GRCh38
rs997006792 Chr.17: 82459737-82459737 on Build GRCh38
rs992513837 Chr.17: 82459804-82459804 on Build GRCh38
rs900028939 Chr.17: 82459718-82459718 on Build GRCh38
rs534077201 Chr.17: 82459945-82459945 on Build GRCh38
rs112789881 Chr.17: 82459842-82459842 on Build GRCh38
rs544467463 Chr.17: 82460054-82460054 on Build GRCh38
rs886810858 Chr.17: 82459868-82459868 on Build GRCh38
rs917776792 Chr.17: 82459653-82459653 on Build GRCh38
rs774698710 Chr.17: 82459614-82459614 on Build GRCh38
rs764679850 Chr.17: 82459946-82459946 on Build GRCh38
rs912764644 Chr.17: 82459611-82459611 on Build GRCh38
rs1019859159 Chr.17: 82459703-82459703 on Build GRCh38
rs947940397 Chr.17: 82459822-82459822 on Build GRCh38
rs766362469 Chr.17: 82460047-82460047 on Build GRCh38
rs745308576 Chr.17: 82460056-82460056 on Build GRCh38
rs896301095 Chr.17: 82460014-82460014 on Build GRCh38
rs913250403 Chr.17: 82459899-82459899 on Build GRCh38
rs980641341 Chr.17: 82459825-82459825 on Build GRCh38
rs1045238087 Chr.17: 82459661-82459661 on Build GRCh38
rs557428182 Chr.17: 82459861-82459861 on Build GRCh38
rs371797405 Chr.17: 82460048-82460048 on Build GRCh38
rs896429764 Chr.17: 82459736-82459736 on Build GRCh38
rs745526721 Chr.17: 82459973-82459973 on Build GRCh38
rs142922828 Chr.17: 82460025-82460025 on Build GRCh38
rs12051807 Chr.17: 82459546-82459546 on Build GRCh38
rs924587781 Chr.17: 82459568-82459568 on Build GRCh38
rs148570791 Chr.17: 82460024-82460024 on Build GRCh38
rs12952969 Chr.17: 82459690-82459690 on Build GRCh38
rs147160209 Chr.17: 82460037-82460037 on Build GRCh38
rs761996941 Chr.17: 82460005-82460005 on Build GRCh38
rs565115638 Chr.17: 82460031-82460031 on Build GRCh38
rs931055220 Chr.17: 82459844-82459844 on Build GRCh38
rs57897512 Chr.17: 82459866-82459866 on Build GRCh38
rs186869853 Chr.17: 82459597-82459597 on Build GRCh38
rs769447874 Chr.17: 82459977-82459977 on Build GRCh38
rs947348834 Chr.17: 82459912-82459912 on Build GRCh38
rs903705270 Chr.17: 82459695-82459695 on Build GRCh38
rs960334508 Chr.17: 82460008-82460008 on Build GRCh38
rs776392943 Chr.17: 82459970-82459970 on Build GRCh38

View Full Product Details