Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 71786197-71786722 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB21
Entrez Gene ID
23011
Gene Name
RAB21, member RAS oncogene family
Gene Aliases
-
UniGene
Hs.524590
Gene Chromosome Location
Chr.12: 71754863-71793476 on Build GRCh38
Transcripts
NM_014999.3

SNP Detail

SNP ID Chromosome Location
rs966395643 Chr.12: 71786601-71786601 on Build GRCh38
rs929932294 Chr.12: 71786486-71786486 on Build GRCh38
rs888335142 Chr.12: 71786513-71786513 on Build GRCh38
rs1021321587 Chr.12: 71786355-71786355 on Build GRCh38
rs901232117 Chr.12: 71786317-71786317 on Build GRCh38
rs911800014 Chr.12: 71786322-71786326 on Build GRCh38
rs1031194284 Chr.12: 71786612-71786612 on Build GRCh38
rs328742 Chr.12: 71786392-71786392 on Build GRCh38
rs79888023 Chr.12: 71786407-71786407 on Build GRCh38
rs886493446 Chr.12: 71786402-71786402 on Build GRCh38
rs1031551390 Chr.12: 71786339-71786339 on Build GRCh38
rs930736018 Chr.12: 71786368-71786368 on Build GRCh38
rs562542679 Chr.12: 71786409-71786409 on Build GRCh38
rs550288536 Chr.12: 71786680-71786680 on Build GRCh38
rs998355137 Chr.12: 71786321-71786321 on Build GRCh38
rs1048429647 Chr.12: 71786261-71786261 on Build GRCh38
rs1034504173 Chr.12: 71786530-71786530 on Build GRCh38
rs938537631 Chr.12: 71786227-71786227 on Build GRCh38
rs897080052 Chr.12: 71786242-71786242 on Build GRCh38
rs1048774779 Chr.12: 71786502-71786502 on Build GRCh38
rs867278103 Chr.12: 71786369-71786369 on Build GRCh38
rs966446243 Chr.12: 71786586-71786586 on Build GRCh38
rs984598232 Chr.12: 71786678-71786678 on Build GRCh38
rs888429736 Chr.12: 71786267-71786267 on Build GRCh38
rs1046189441 Chr.12: 71786700-71786700 on Build GRCh38
rs775642010 Chr.12: 71786543-71786543 on Build GRCh38
rs990123069 Chr.12: 71786353-71786353 on Build GRCh38
rs978168756 Chr.12: 71786591-71786591 on Build GRCh38
rs921789029 Chr.12: 71786606-71786606 on Build GRCh38
rs188009079 Chr.12: 71786305-71786305 on Build GRCh38
rs376273883 Chr.12: 71786335-71786336 on Build GRCh38
rs549192926 Chr.12: 71786370-71786370 on Build GRCh38
rs1011049082 Chr.12: 71786673-71786673 on Build GRCh38
rs537620139 Chr.12: 71786271-71786271 on Build GRCh38
rs1057330106 Chr.12: 71786240-71786240 on Build GRCh38
rs537592723 Chr.12: 71786360-71786360 on Build GRCh38
rs918413824 Chr.12: 71786480-71786480 on Build GRCh38
rs1039685097 Chr.12: 71786599-71786599 on Build GRCh38
rs901263136 Chr.12: 71786600-71786600 on Build GRCh38
rs546723150 Chr.12: 71786627-71786627 on Build GRCh38
rs570117980 Chr.12: 71786342-71786342 on Build GRCh38
rs944605851 Chr.12: 71786325-71786325 on Build GRCh38
rs141431614 Chr.12: 71786605-71786605 on Build GRCh38
rs747118914 Chr.12: 71786657-71786657 on Build GRCh38
rs184690521 Chr.12: 71786554-71786554 on Build GRCh38
rs940780299 Chr.12: 71786694-71786694 on Build GRCh38
rs992709833 Chr.12: 71786434-71786434 on Build GRCh38
rs769719470 Chr.12: 71786533-71786533 on Build GRCh38
rs574991880 Chr.12: 71786664-71786664 on Build GRCh38
rs1045699952 Chr.12: 71786462-71786462 on Build GRCh38
rs537047583 Chr.12: 71786606-71786607 on Build GRCh38
rs929876157 Chr.12: 71786254-71786254 on Build GRCh38
rs181304002 Chr.12: 71786481-71786481 on Build GRCh38
rs56100239 Chr.12: 71786584-71786584 on Build GRCh38
rs893293475 Chr.12: 71786541-71786541 on Build GRCh38
rs938400492 Chr.12: 71786415-71786415 on Build GRCh38
rs1022523439 Chr.12: 71786695-71786695 on Build GRCh38

View Full Product Details