Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 119178984-119179486 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPB8
Entrez Gene ID
26353
Gene Name
heat shock protein family B (small) member 8
Gene Aliases
CMT2L, DHMN2, E2IG1, H11, HMN2, HMN2A, HSP22
UniGene
Hs.400095
Gene Chromosome Location
Chr.12: 119178790-119194746 on Build GRCh38
Transcripts
NM_014365.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6537841 c.134C>G p.T45R Chr.12: 119179446-119179446 on Build GRCh38
1359244 c.126C>T p.D42D Chr.12: 119179438-119179438 on Build GRCh38
4760658 c.50G>A p.R17H Chr.12: 119179362-119179362 on Build GRCh38
7317765 c.97G>A p.D33N Chr.12: 119179409-119179409 on Build GRCh38
1322724 c.52C>G p.R18G Chr.12: 119179364-119179364 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771827550 Chr.12: 119179428-119179428 on Build GRCh38
rs1036726530 Chr.12: 119179351-119179351 on Build GRCh38
rs773298891 Chr.12: 119179399-119179399 on Build GRCh38
rs903417106 Chr.12: 119179202-119179202 on Build GRCh38
rs141871482 Chr.12: 119179426-119179426 on Build GRCh38
rs35832069 Chr.12: 119179376-119179377 on Build GRCh38
rs752348447 Chr.12: 119179356-119179356 on Build GRCh38
rs146997263 Chr.12: 119179362-119179362 on Build GRCh38
rs763658767 Chr.12: 119179438-119179438 on Build GRCh38
rs759005659 Chr.12: 119179364-119179364 on Build GRCh38
rs886049025 Chr.12: 119179141-119179141 on Build GRCh38
rs971836129 Chr.12: 119179073-119179073 on Build GRCh38
rs147945133 Chr.12: 119179405-119179405 on Build GRCh38
rs530083542 Chr.12: 119179329-119179329 on Build GRCh38
rs758781718 Chr.12: 119179283-119179283 on Build GRCh38
rs775152699 Chr.12: 119179429-119179429 on Build GRCh38
rs887591853 Chr.12: 119179054-119179054 on Build GRCh38
rs146900850 Chr.12: 119179326-119179326 on Build GRCh38
rs769926309 Chr.12: 119179310-119179310 on Build GRCh38
rs764259720 Chr.12: 119179112-119179112 on Build GRCh38
rs982621631 Chr.12: 119179033-119179033 on Build GRCh38
rs774233360 Chr.12: 119179337-119179337 on Build GRCh38
rs377690814 Chr.12: 119179317-119179317 on Build GRCh38
rs1026783882 Chr.12: 119179012-119179012 on Build GRCh38
rs750990822 Chr.12: 119179201-119179201 on Build GRCh38
rs1057101638 Chr.12: 119179239-119179239 on Build GRCh38
rs894473202 Chr.12: 119179261-119179261 on Build GRCh38
rs374549932 Chr.12: 119179307-119179307 on Build GRCh38
rs762887808 Chr.12: 119179322-119179322 on Build GRCh38
rs201594365 Chr.12: 119179287-119179287 on Build GRCh38
rs952653956 Chr.12: 119179016-119179016 on Build GRCh38
rs765762114 Chr.12: 119179050-119179050 on Build GRCh38
rs1041936869 Chr.12: 119179164-119179164 on Build GRCh38
rs952834680 Chr.12: 119179321-119179321 on Build GRCh38
rs779546616 Chr.12: 119179262-119179262 on Build GRCh38
rs747381453 Chr.12: 119179377-119179377 on Build GRCh38
rs978083858 Chr.12: 119179156-119179157 on Build GRCh38
rs548748659 Chr.12: 119179155-119179155 on Build GRCh38
rs755359241 Chr.12: 119179379-119179379 on Build GRCh38
rs752076364 Chr.12: 119179460-119179460 on Build GRCh38
rs747163439 Chr.12: 119179301-119179301 on Build GRCh38
rs146732479 Chr.12: 119179076-119179076 on Build GRCh38
rs371236491 Chr.12: 119179459-119179459 on Build GRCh38
rs759360759 Chr.12: 119179427-119179427 on Build GRCh38
rs759289686 Chr.12: 119179344-119179344 on Build GRCh38
rs529120786 Chr.12: 119179154-119179154 on Build GRCh38
rs755586270 Chr.12: 119179360-119179360 on Build GRCh38
rs200120006 Chr.12: 119179361-119179361 on Build GRCh38
rs748320300 Chr.12: 119179397-119179397 on Build GRCh38
rs1015406788 Chr.12: 119179330-119179330 on Build GRCh38
rs760427734 Chr.12: 119179437-119179437 on Build GRCh38
rs995989266 Chr.12: 119179052-119179052 on Build GRCh38
rs761462898 Chr.12: 119179440-119179440 on Build GRCh38
rs1002901893 Chr.12: 119179421-119179421 on Build GRCh38
rs185294734 Chr.12: 119179248-119179248 on Build GRCh38
rs920948681 Chr.12: 119179096-119179096 on Build GRCh38
rs1027479328 Chr.12: 119179293-119179293 on Build GRCh38
rs920906527 Chr.12: 119179433-119179433 on Build GRCh38
rs1040134521 Chr.12: 119179005-119179005 on Build GRCh38
rs907098896 Chr.12: 119179407-119179407 on Build GRCh38
rs377372380 Chr.12: 119179304-119179304 on Build GRCh38
rs200874667 Chr.12: 119179335-119179335 on Build GRCh38
rs1014805620 Chr.12: 119179065-119179065 on Build GRCh38
rs944410140 Chr.12: 119179183-119179183 on Build GRCh38
rs1005228743 Chr.12: 119179059-119179059 on Build GRCh38
rs931748228 Chr.12: 119179258-119179258 on Build GRCh38
rs11549401 Chr.12: 119179120-119179120 on Build GRCh38
rs780751241 Chr.12: 119179376-119179376 on Build GRCh38
rs779704770 Chr.12: 119179206-119179206 on Build GRCh38
rs781475312 Chr.12: 119179383-119179383 on Build GRCh38
rs1039998051 Chr.12: 119179197-119179197 on Build GRCh38
rs374254847 Chr.12: 119179461-119179461 on Build GRCh38
rs770823712 Chr.12: 119179420-119179420 on Build GRCh38
rs769707671 Chr.12: 119179268-119179275 on Build GRCh38
rs377147867 Chr.12: 119179278-119179278 on Build GRCh38
rs767174743 Chr.12: 119179354-119179354 on Build GRCh38
rs758136546 Chr.12: 119179159-119179159 on Build GRCh38
rs762394807 Chr.12: 119179039-119179039 on Build GRCh38
rs958762419 Chr.12: 119179108-119179108 on Build GRCh38
rs373146924 Chr.12: 119179300-119179300 on Build GRCh38

View Full Product Details