Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 41834878-41835382 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JMJD7
Entrez Gene ID
100137047
Gene Name
jumonji domain containing 7
Gene Aliases
-
UniGene
Hs.198161
Gene Chromosome Location
Chr.15: 41828085-41837587 on Build GRCh38
Transcripts
NM_001114632.1 NM_001198588.1 NM_005090.3

Gene Symbol
JMJD7-PLA2G4B
Entrez Gene ID
8681
Gene Name
JMJD7-PLA2G4B readthrough
Gene Aliases
HsT16992, cPLA2-beta
UniGene
Hs.198161
Gene Chromosome Location
Chr.15: 41828085-41848148 on Build GRCh38
Transcripts
NM_001114632.1 NM_001198588.1 NM_005090.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7137938 c.218G>C p.R73T Chr.15: 41834893-41834893 on Build GRCh38
961570 c.403C>A p.L135M Chr.15: 41835154-41835154 on Build GRCh38
961565 c.333C>T p.F111F Chr.15: 41835084-41835084 on Build GRCh38
5473614 c.411C>T p.S137S Chr.15: 41835162-41835162 on Build GRCh38
961571 c.403C>A p.L135M Chr.15: 41835154-41835154 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771301136 Chr.15: 41835236-41835236 on Build GRCh38
rs759601576 Chr.15: 41834980-41834980 on Build GRCh38
rs2241523 Chr.15: 41835245-41835245 on Build GRCh38
rs763236711 Chr.15: 41835152-41835152 on Build GRCh38
rs766813591 Chr.15: 41835090-41835090 on Build GRCh38
rs758469784 Chr.15: 41834933-41834933 on Build GRCh38
rs780167106 Chr.15: 41834934-41834934 on Build GRCh38
rs898749507 Chr.15: 41835314-41835314 on Build GRCh38
rs779337927 Chr.15: 41834904-41834904 on Build GRCh38
rs1028782543 Chr.15: 41835077-41835077 on Build GRCh38
rs148739127 Chr.15: 41835195-41835195 on Build GRCh38
rs112203400 Chr.15: 41835257-41835257 on Build GRCh38
rs925587595 Chr.15: 41834954-41834954 on Build GRCh38
rs759973342 Chr.15: 41835242-41835242 on Build GRCh38
rs772845928 Chr.15: 41835233-41835233 on Build GRCh38
rs776366285 Chr.15: 41835103-41835103 on Build GRCh38
rs143398561 Chr.15: 41835179-41835179 on Build GRCh38
rs371335846 Chr.15: 41834902-41834902 on Build GRCh38
rs781407834 Chr.15: 41835031-41835031 on Build GRCh38
rs747177498 Chr.15: 41834939-41834939 on Build GRCh38
rs1045907819 Chr.15: 41835251-41835251 on Build GRCh38
rs768201038 Chr.15: 41835122-41835122 on Build GRCh38
rs760597072 Chr.15: 41835064-41835064 on Build GRCh38
rs779978452 Chr.15: 41835117-41835117 on Build GRCh38
rs1032669808 Chr.15: 41835350-41835350 on Build GRCh38
rs200624748 Chr.15: 41834913-41834913 on Build GRCh38
rs1015736503 Chr.15: 41834908-41834908 on Build GRCh38
rs781222742 Chr.15: 41835214-41835214 on Build GRCh38
rs756281013 Chr.15: 41835173-41835173 on Build GRCh38
rs1001121060 Chr.15: 41835328-41835328 on Build GRCh38
rs575931750 Chr.15: 41835296-41835296 on Build GRCh38
rs780685788 Chr.15: 41835130-41835131 on Build GRCh38
rs200054658 Chr.15: 41835295-41835295 on Build GRCh38
rs145753099 Chr.15: 41835023-41835023 on Build GRCh38
rs1037730411 Chr.15: 41835276-41835276 on Build GRCh38
rs752091465 Chr.15: 41835162-41835162 on Build GRCh38
rs776739162 Chr.15: 41834949-41834949 on Build GRCh38
rs755078404 Chr.15: 41835100-41835100 on Build GRCh38
rs746244300 Chr.15: 41835073-41835073 on Build GRCh38
rs75094146 Chr.15: 41835222-41835222 on Build GRCh38
rs766168095 Chr.15: 41835260-41835260 on Build GRCh38
rs1055909666 Chr.15: 41834975-41834975 on Build GRCh38
rs935807183 Chr.15: 41834976-41834976 on Build GRCh38
rs554013827 Chr.15: 41835253-41835253 on Build GRCh38
rs746943774 Chr.15: 41835203-41835203 on Build GRCh38
rs979753126 Chr.15: 41835125-41835125 on Build GRCh38
rs374779486 Chr.15: 41835024-41835024 on Build GRCh38
rs142916967 Chr.15: 41835077-41835077 on Build GRCh38
rs751624183 Chr.15: 41835033-41835034 on Build GRCh38
rs755026008 Chr.15: 41835030-41835030 on Build GRCh38
rs992870458 Chr.15: 41834928-41834928 on Build GRCh38
rs80284393 Chr.15: 41835094-41835094 on Build GRCh38
rs764338229 Chr.15: 41835252-41835252 on Build GRCh38
rs746989390 Chr.15: 41835120-41835120 on Build GRCh38
rs768058716 Chr.15: 41835065-41835065 on Build GRCh38
rs374618704 Chr.15: 41835169-41835169 on Build GRCh38
rs781183887 Chr.15: 41835101-41835101 on Build GRCh38
rs150789634 Chr.15: 41835084-41835084 on Build GRCh38
rs925902618 Chr.15: 41834960-41834960 on Build GRCh38
rs969684743 Chr.15: 41835115-41835115 on Build GRCh38
rs56276001 Chr.15: 41835281-41835281 on Build GRCh38
rs957315623 Chr.15: 41835145-41835145 on Build GRCh38
rs775176223 Chr.15: 41834993-41834993 on Build GRCh38
rs576647114 Chr.15: 41834945-41834945 on Build GRCh38
rs768883637 Chr.15: 41834947-41834947 on Build GRCh38
rs770851869 Chr.15: 41834959-41834959 on Build GRCh38
rs761423879 Chr.15: 41835124-41835124 on Build GRCh38
rs371042555 Chr.15: 41835107-41835107 on Build GRCh38
rs757517732 Chr.15: 41834920-41834921 on Build GRCh38
rs932615267 Chr.15: 41835238-41835238 on Build GRCh38
rs754121935 Chr.15: 41835186-41835186 on Build GRCh38
rs77521398 Chr.15: 41834958-41834958 on Build GRCh38
rs747598273 Chr.15: 41835219-41835219 on Build GRCh38
rs765659372 Chr.15: 41835021-41835021 on Build GRCh38
rs758265428 Chr.15: 41835190-41835190 on Build GRCh38
rs760007240 Chr.15: 41835163-41835163 on Build GRCh38
rs35873017 Chr.15: 41835352-41835353 on Build GRCh38
rs769398219 Chr.15: 41835127-41835127 on Build GRCh38
rs867603436 Chr.15: 41835091-41835091 on Build GRCh38
rs370469363 Chr.15: 41834963-41834963 on Build GRCh38
rs377033075 Chr.15: 41835131-41835131 on Build GRCh38
rs146955165 Chr.15: 41835004-41835004 on Build GRCh38
rs774615941 Chr.15: 41835239-41835239 on Build GRCh38
rs75995450 Chr.15: 41835289-41835289 on Build GRCh38
rs748383621 Chr.15: 41835042-41835042 on Build GRCh38
rs901154958 Chr.15: 41835043-41835043 on Build GRCh38
rs751272288 Chr.15: 41835264-41835264 on Build GRCh38
rs767110569 Chr.15: 41834989-41834989 on Build GRCh38
rs752675352 Chr.15: 41835171-41835171 on Build GRCh38
rs185919788 Chr.15: 41835318-41835318 on Build GRCh38
rs756440690 Chr.15: 41835106-41835106 on Build GRCh38
rs548174849 Chr.15: 41835213-41835213 on Build GRCh38
rs984318531 Chr.15: 41834903-41834903 on Build GRCh38
rs368638530 Chr.15: 41835244-41835244 on Build GRCh38
rs768369402 Chr.15: 41835069-41835069 on Build GRCh38
rs533259141 Chr.15: 41835058-41835058 on Build GRCh38
rs775317128 Chr.15: 41835062-41835062 on Build GRCh38
rs772010631 Chr.15: 41835113-41835113 on Build GRCh38
rs745613438 Chr.15: 41835108-41835108 on Build GRCh38
rs562707007 Chr.15: 41835085-41835093 on Build GRCh38
rs747062485 Chr.15: 41835028-41835028 on Build GRCh38
rs373782042 Chr.15: 41835022-41835022 on Build GRCh38
rs1028550403 Chr.15: 41835050-41835050 on Build GRCh38
rs955478533 Chr.15: 41835092-41835092 on Build GRCh38
rs760138153 Chr.15: 41835003-41835003 on Build GRCh38
rs796173415 Chr.15: 41835126-41835126 on Build GRCh38
rs541847469 Chr.15: 41835316-41835316 on Build GRCh38
rs138958721 Chr.15: 41835051-41835051 on Build GRCh38
rs754825604 Chr.15: 41835265-41835265 on Build GRCh38
rs199904907 Chr.15: 41835009-41835009 on Build GRCh38
rs775344075 Chr.15: 41835044-41835044 on Build GRCh38
rs765565225 Chr.15: 41835075-41835075 on Build GRCh38
rs539179690 Chr.15: 41835277-41835277 on Build GRCh38
rs776344544 Chr.15: 41835068-41835068 on Build GRCh38
rs550115704 Chr.15: 41835085-41835085 on Build GRCh38

View Full Product Details