Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 21678220-21678721 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USP48
Entrez Gene ID
84196
Gene Name
ubiquitin specific peptidase 48
Gene Aliases
RAP1GA1, USP31
UniGene
Hs.467524
Gene Chromosome Location
Chr.1: 21678298-21783620 on Build GRCh38
Transcripts
NM_032236.5

SNP Detail

SNP ID Chromosome Location
rs111506041 Chr.1: 21678287-21678287 on Build GRCh38
rs943484579 Chr.1: 21678553-21678553 on Build GRCh38
rs368701503 Chr.1: 21678435-21678435 on Build GRCh38
rs192131755 Chr.1: 21678369-21678369 on Build GRCh38
rs920505911 Chr.1: 21678454-21678454 on Build GRCh38
rs1012185265 Chr.1: 21678418-21678418 on Build GRCh38
rs943367752 Chr.1: 21678296-21678297 on Build GRCh38
rs766051149 Chr.1: 21678603-21678603 on Build GRCh38
rs959454543 Chr.1: 21678442-21678442 on Build GRCh38
rs748123518 Chr.1: 21678421-21678421 on Build GRCh38
rs577092432 Chr.1: 21678339-21678339 on Build GRCh38
rs1009636365 Chr.1: 21678359-21678359 on Build GRCh38
rs932808250 Chr.1: 21678590-21678590 on Build GRCh38
rs750697010 Chr.1: 21678288-21678289 on Build GRCh38
rs187194916 Chr.1: 21678269-21678269 on Build GRCh38
rs909297506 Chr.1: 21678527-21678527 on Build GRCh38
rs549114889 Chr.1: 21678287-21678288 on Build GRCh38
rs944104070 Chr.1: 21678533-21678533 on Build GRCh38
rs868035727 Chr.1: 21678686-21678686 on Build GRCh38
rs200312469 Chr.1: 21678301-21678301 on Build GRCh38
rs565211315 Chr.1: 21678624-21678624 on Build GRCh38
rs1032986433 Chr.1: 21678338-21678338 on Build GRCh38
rs796196003 Chr.1: 21678287-21678288 on Build GRCh38
rs997081737 Chr.1: 21678286-21678286 on Build GRCh38
rs909319388 Chr.1: 21678294-21678295 on Build GRCh38
rs894013542 Chr.1: 21678634-21678634 on Build GRCh38
rs397727492 Chr.1: 21678300-21678301 on Build GRCh38
rs933542975 Chr.1: 21678651-21678651 on Build GRCh38
rs550267567 Chr.1: 21678664-21678664 on Build GRCh38
rs1009237757 Chr.1: 21678650-21678650 on Build GRCh38
rs532857237 Chr.1: 21678633-21678633 on Build GRCh38
rs10917034 Chr.1: 21678436-21678436 on Build GRCh38
rs11428215 Chr.1: 21678286-21678287 on Build GRCh38
rs147153186 Chr.1: 21678674-21678674 on Build GRCh38
rs908638047 Chr.1: 21678503-21678503 on Build GRCh38
rs1053190343 Chr.1: 21678374-21678374 on Build GRCh38
rs970425460 Chr.1: 21678415-21678415 on Build GRCh38
rs891974750 Chr.1: 21678375-21678379 on Build GRCh38
rs113663369 Chr.1: 21678499-21678499 on Build GRCh38
rs1026123457 Chr.1: 21678460-21678461 on Build GRCh38
rs1042051448 Chr.1: 21678663-21678663 on Build GRCh38
rs561088332 Chr.1: 21678523-21678523 on Build GRCh38
rs371062651 Chr.1: 21678443-21678443 on Build GRCh38
rs950567251 Chr.1: 21678477-21678479 on Build GRCh38
rs933585034 Chr.1: 21678354-21678354 on Build GRCh38
rs760474034 Chr.1: 21678578-21678578 on Build GRCh38
rs528648699 Chr.1: 21678559-21678559 on Build GRCh38
rs931366106 Chr.1: 21678458-21678458 on Build GRCh38
rs929353004 Chr.1: 21678268-21678268 on Build GRCh38
rs563714151 Chr.1: 21678403-21678403 on Build GRCh38
rs546819189 Chr.1: 21678575-21678575 on Build GRCh38
rs868488590 Chr.1: 21678466-21678466 on Build GRCh38
rs867955108 Chr.1: 21678685-21678685 on Build GRCh38
rs1041213260 Chr.1: 21678557-21678557 on Build GRCh38
rs893510356 Chr.1: 21678357-21678357 on Build GRCh38
rs973368181 Chr.1: 21678446-21678446 on Build GRCh38
rs558873622 Chr.1: 21678300-21678300 on Build GRCh38

View Full Product Details