Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 21679009-21679517 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USP48
Entrez Gene ID
84196
Gene Name
ubiquitin specific peptidase 48
Gene Aliases
RAP1GA1, USP31
UniGene
Hs.467524
Gene Chromosome Location
Chr.1: 21678298-21783620 on Build GRCh38
Transcripts
NM_032236.5 XM_005246009.2 XM_006710951.2 XM_006710955.3 XM_011542262.2 XM_011542263.2 XM_011542264.1 XM_011542265.2 XM_011542267.2 XM_011542268.2 XM_017002495.1 XM_017002497.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4781296 c.1562A>G p.H521R Chr.1: 21679421-21679421 on Build GRCh38
3705528 c.3104A>G p.H1035R Chr.1: 21679421-21679421 on Build GRCh38
291760 c.3090T>A p.T1030T Chr.1: 21679435-21679435 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs867217351 Chr.1: 21679045-21679045 on Build GRCh38
rs763178187 Chr.1: 21679465-21679465 on Build GRCh38
rs199842068 Chr.1: 21679441-21679441 on Build GRCh38
rs766518075 Chr.1: 21679467-21679467 on Build GRCh38
rs745580593 Chr.1: 21679395-21679395 on Build GRCh38
rs945509704 Chr.1: 21679126-21679126 on Build GRCh38
rs1018823123 Chr.1: 21679283-21679283 on Build GRCh38
rs186995427 Chr.1: 21679336-21679336 on Build GRCh38
rs1048791991 Chr.1: 21679268-21679268 on Build GRCh38
rs1059218 Chr.1: 21679172-21679172 on Build GRCh38
rs563078410 Chr.1: 21679460-21679460 on Build GRCh38
rs187285308 Chr.1: 21679081-21679081 on Build GRCh38
rs770810159 Chr.1: 21679457-21679457 on Build GRCh38
rs976685011 Chr.1: 21679063-21679063 on Build GRCh38
rs776570889 Chr.1: 21679456-21679456 on Build GRCh38
rs368352580 Chr.1: 21679407-21679407 on Build GRCh38
rs768595908 Chr.1: 21679449-21679449 on Build GRCh38
rs768662799 Chr.1: 21679290-21679290 on Build GRCh38
rs771466002 Chr.1: 21679401-21679401 on Build GRCh38
rs949125143 Chr.1: 21679346-21679346 on Build GRCh38
rs886299680 Chr.1: 21679275-21679275 on Build GRCh38
rs565484355 Chr.1: 21679296-21679296 on Build GRCh38
rs978378715 Chr.1: 21679157-21679157 on Build GRCh38
rs756584043 Chr.1: 21679452-21679453 on Build GRCh38
rs1004662822 Chr.1: 21679280-21679280 on Build GRCh38
rs780247231 Chr.1: 21679115-21679115 on Build GRCh38
rs368579224 Chr.1: 21679470-21679470 on Build GRCh38
rs751623588 Chr.1: 21679469-21679469 on Build GRCh38
rs781759822 Chr.1: 21679482-21679482 on Build GRCh38
rs965381235 Chr.1: 21679058-21679058 on Build GRCh38
rs954244275 Chr.1: 21679080-21679080 on Build GRCh38
rs201522478 Chr.1: 21679429-21679429 on Build GRCh38
rs773166264 Chr.1: 21679333-21679333 on Build GRCh38
rs1047355027 Chr.1: 21679418-21679418 on Build GRCh38
rs937474730 Chr.1: 21679259-21679259 on Build GRCh38
rs1028712099 Chr.1: 21679384-21679384 on Build GRCh38
rs78452980 Chr.1: 21679055-21679055 on Build GRCh38
rs200662556 Chr.1: 21679432-21679432 on Build GRCh38
rs989994129 Chr.1: 21679468-21679468 on Build GRCh38
rs939007486 Chr.1: 21679179-21679179 on Build GRCh38
rs140288196 Chr.1: 21679295-21679295 on Build GRCh38
rs182665037 Chr.1: 21679188-21679188 on Build GRCh38
rs368722392 Chr.1: 21679090-21679090 on Build GRCh38
rs199557120 Chr.1: 21679427-21679427 on Build GRCh38
rs372358243 Chr.1: 21679421-21679421 on Build GRCh38
rs200354902 Chr.1: 21679426-21679426 on Build GRCh38
rs551252073 Chr.1: 21679434-21679434 on Build GRCh38
rs192015805 Chr.1: 21679092-21679092 on Build GRCh38
rs998213927 Chr.1: 21679340-21679340 on Build GRCh38
rs746901254 Chr.1: 21679360-21679360 on Build GRCh38
rs930362163 Chr.1: 21679254-21679254 on Build GRCh38
rs778674564 Chr.1: 21679387-21679387 on Build GRCh38
rs183514931 Chr.1: 21679070-21679070 on Build GRCh38
rs113660412 Chr.1: 21679218-21679218 on Build GRCh38
rs1055159949 Chr.1: 21679184-21679184 on Build GRCh38
rs774137482 Chr.1: 21679288-21679288 on Build GRCh38
rs753294190 Chr.1: 21679483-21679483 on Build GRCh38
rs556233716 Chr.1: 21679046-21679046 on Build GRCh38
rs533887228 Chr.1: 21679050-21679050 on Build GRCh38
rs376642031 Chr.1: 21679458-21679458 on Build GRCh38
rs1027879338 Chr.1: 21679040-21679040 on Build GRCh38
rs778254076 Chr.1: 21679479-21679480 on Build GRCh38

View Full Product Details