Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 79457211-79457705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1024
Entrez Gene ID
23251
Gene Name
KIAA1024
Gene Aliases
-
UniGene
Hs.301654
Gene Chromosome Location
Chr.15: 79411557-79472304 on Build GRCh38
Transcripts
NM_015206.2 XM_011521392.1 XM_011521393.2 XM_011521395.2 XM_011521396.2 XM_017022027.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6077965 c.1286C>A p.A429E Chr.15: 79457433-79457433 on Build GRCh38
965484 c.1441G>A p.V481M Chr.15: 79457588-79457588 on Build GRCh38
2011539 c.1232G>A p.R411H Chr.15: 79457379-79457379 on Build GRCh38
965486 c.1546G>A p.D516N Chr.15: 79457693-79457693 on Build GRCh38
4459898 c.1145C>T p.S382F Chr.15: 79457292-79457292 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs181865672 Chr.15: 79457384-79457384 on Build GRCh38
rs762572890 Chr.15: 79457417-79457417 on Build GRCh38
rs897509423 Chr.15: 79457237-79457237 on Build GRCh38
rs746759999 Chr.15: 79457448-79457449 on Build GRCh38
rs369507901 Chr.15: 79457650-79457650 on Build GRCh38
rs749272522 Chr.15: 79457301-79457301 on Build GRCh38
rs769231383 Chr.15: 79457302-79457302 on Build GRCh38
rs79651143 Chr.15: 79457360-79457360 on Build GRCh38
rs201404813 Chr.15: 79457582-79457582 on Build GRCh38
rs549288323 Chr.15: 79457429-79457429 on Build GRCh38
rs373558330 Chr.15: 79457288-79457288 on Build GRCh38
rs779526675 Chr.15: 79457377-79457377 on Build GRCh38
rs779745959 Chr.15: 79457342-79457342 on Build GRCh38
rs764131947 Chr.15: 79457332-79457332 on Build GRCh38
rs746183055 Chr.15: 79457679-79457679 on Build GRCh38
rs148542288 Chr.15: 79457378-79457378 on Build GRCh38
rs374349090 Chr.15: 79457318-79457318 on Build GRCh38
rs201473692 Chr.15: 79457494-79457494 on Build GRCh38
rs758362431 Chr.15: 79457246-79457246 on Build GRCh38
rs759259983 Chr.15: 79457322-79457322 on Build GRCh38
rs780110040 Chr.15: 79457293-79457293 on Build GRCh38
rs147416682 Chr.15: 79457567-79457567 on Build GRCh38
rs765443010 Chr.15: 79457649-79457649 on Build GRCh38
rs759682450 Chr.15: 79457640-79457640 on Build GRCh38
rs371598881 Chr.15: 79457309-79457309 on Build GRCh38
rs777521578 Chr.15: 79457257-79457257 on Build GRCh38
rs139919189 Chr.15: 79457619-79457619 on Build GRCh38
rs751429691 Chr.15: 79457344-79457344 on Build GRCh38
rs772542731 Chr.15: 79457315-79457315 on Build GRCh38
rs199834575 Chr.15: 79457285-79457285 on Build GRCh38
rs769600562 Chr.15: 79457411-79457411 on Build GRCh38
rs935105217 Chr.15: 79457584-79457584 on Build GRCh38
rs749420225 Chr.15: 79457372-79457372 on Build GRCh38
rs544980722 Chr.15: 79457633-79457633 on Build GRCh38
rs773717135 Chr.15: 79457398-79457398 on Build GRCh38
rs755865839 Chr.15: 79457474-79457474 on Build GRCh38
rs766667683 Chr.15: 79457484-79457484 on Build GRCh38
rs763498870 Chr.15: 79457423-79457423 on Build GRCh38
rs941899412 Chr.15: 79457562-79457562 on Build GRCh38
rs749563921 Chr.15: 79457566-79457566 on Build GRCh38
rs755143664 Chr.15: 79457514-79457514 on Build GRCh38
rs149369510 Chr.15: 79457286-79457286 on Build GRCh38
rs779813346 Chr.15: 79457580-79457580 on Build GRCh38
rs145637309 Chr.15: 79457651-79457651 on Build GRCh38
rs757097711 Chr.15: 79457352-79457352 on Build GRCh38
rs780092656 Chr.15: 79457368-79457368 on Build GRCh38
rs753264951 Chr.15: 79457362-79457362 on Build GRCh38
rs780425765 Chr.15: 79457546-79457546 on Build GRCh38
rs767382134 Chr.15: 79457448-79457448 on Build GRCh38
rs61740913 Chr.15: 79457617-79457617 on Build GRCh38
rs781411019 Chr.15: 79457678-79457678 on Build GRCh38
rs371873445 Chr.15: 79457433-79457433 on Build GRCh38
rs770555397 Chr.15: 79457671-79457671 on Build GRCh38
rs368033943 Chr.15: 79457289-79457289 on Build GRCh38
rs778472194 Chr.15: 79457654-79457654 on Build GRCh38
rs771177539 Chr.15: 79457579-79457579 on Build GRCh38
rs371812078 Chr.15: 79457434-79457434 on Build GRCh38
rs758002743 Chr.15: 79457257-79457257 on Build GRCh38
rs77230768 Chr.15: 79457397-79457397 on Build GRCh38
rs367894983 Chr.15: 79457373-79457373 on Build GRCh38
rs768284783 Chr.15: 79457587-79457587 on Build GRCh38
rs761261007 Chr.15: 79457607-79457607 on Build GRCh38
rs751204505 Chr.15: 79457520-79457520 on Build GRCh38
rs1044931661 Chr.15: 79457442-79457442 on Build GRCh38
rs758122997 Chr.15: 79457578-79457578 on Build GRCh38
rs772031549 Chr.15: 79457612-79457612 on Build GRCh38
rs762328772 Chr.15: 79457330-79457330 on Build GRCh38
rs777262791 Chr.15: 79457620-79457620 on Build GRCh38
rs35576815 Chr.15: 79457594-79457594 on Build GRCh38
rs370387568 Chr.15: 79457272-79457272 on Build GRCh38
rs981840229 Chr.15: 79457379-79457380 on Build GRCh38
rs141252372 Chr.15: 79457379-79457379 on Build GRCh38
rs764918499 Chr.15: 79457328-79457328 on Build GRCh38
rs750243493 Chr.15: 79457473-79457473 on Build GRCh38
rs778855436 Chr.15: 79457523-79457523 on Build GRCh38
rs748590849 Chr.15: 79457314-79457314 on Build GRCh38
rs758989301 Chr.15: 79457559-79457559 on Build GRCh38
rs773744096 Chr.15: 79457598-79457598 on Build GRCh38
rs752098586 Chr.15: 79457666-79457666 on Build GRCh38
rs747321344 Chr.15: 79457401-79457401 on Build GRCh38
rs372335362 Chr.15: 79457329-79457329 on Build GRCh38
rs767292002 Chr.15: 79457353-79457353 on Build GRCh38
rs774973465 Chr.15: 79457414-79457414 on Build GRCh38
rs577612145 Chr.15: 79457356-79457356 on Build GRCh38
rs756339829 Chr.15: 79457361-79457361 on Build GRCh38
rs752644862 Chr.15: 79457234-79457234 on Build GRCh38

View Full Product Details