Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 19040239-19040748 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMC7
Entrez Gene ID
79905
Gene Name
transmembrane channel like 7
Gene Aliases
-
UniGene
Hs.187377
Gene Chromosome Location
Chr.16: 18983934-19063942 on Build GRCh38
Transcripts
NM_001160364.2 NM_001300732.1 NM_001324263.1 NM_001324265.1 NM_001324268.1 NM_024847.4 NR_136733.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5593464 c.1277C>T p.A426V Chr.16: 19040386-19040386 on Build GRCh38
3506635 c.1239G>A p.V413V Chr.16: 19040348-19040348 on Build GRCh38
6402206 c.1268T>C p.M423T Chr.16: 19040377-19040377 on Build GRCh38
3506637 c.1284C>T p.I428I Chr.16: 19040393-19040393 on Build GRCh38
3506636 c.1242C>T p.I414I Chr.16: 19040351-19040351 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs879047546 Chr.16: 19040523-19040523 on Build GRCh38
rs144562384 Chr.16: 19040420-19040420 on Build GRCh38
rs749839774 Chr.16: 19040295-19040295 on Build GRCh38
rs200081538 Chr.16: 19040376-19040376 on Build GRCh38
rs894324777 Chr.16: 19040381-19040381 on Build GRCh38
rs772314010 Chr.16: 19040309-19040309 on Build GRCh38
rs940991347 Chr.16: 19040590-19040590 on Build GRCh38
rs763752623 Chr.16: 19040473-19040473 on Build GRCh38
rs920633113 Chr.16: 19040674-19040674 on Build GRCh38
rs553212676 Chr.16: 19040522-19040522 on Build GRCh38
rs759357346 Chr.16: 19040430-19040430 on Build GRCh38
rs752432481 Chr.16: 19040341-19040341 on Build GRCh38
rs771463397 Chr.16: 19040299-19040299 on Build GRCh38
rs960279884 Chr.16: 19040396-19040396 on Build GRCh38
rs770845593 Chr.16: 19040440-19040440 on Build GRCh38
rs112947023 Chr.16: 19040347-19040347 on Build GRCh38
rs751578885 Chr.16: 19040429-19040429 on Build GRCh38
rs73528934 Chr.16: 19040419-19040419 on Build GRCh38
rs775850366 Chr.16: 19040311-19040311 on Build GRCh38
rs761944868 Chr.16: 19040328-19040328 on Build GRCh38
rs771079685 Chr.16: 19040536-19040536 on Build GRCh38
rs150991752 Chr.16: 19040398-19040398 on Build GRCh38
rs764319642 Chr.16: 19040324-19040324 on Build GRCh38
rs61297988 Chr.16: 19040500-19040501 on Build GRCh38
rs753478403 Chr.16: 19040478-19040478 on Build GRCh38
rs1049525958 Chr.16: 19040534-19040534 on Build GRCh38
rs376911484 Chr.16: 19040330-19040330 on Build GRCh38
rs772528907 Chr.16: 19040397-19040397 on Build GRCh38
rs370812589 Chr.16: 19040336-19040336 on Build GRCh38
rs1005243920 Chr.16: 19040628-19040628 on Build GRCh38
rs567252665 Chr.16: 19040606-19040606 on Build GRCh38
rs572384371 Chr.16: 19040539-19040539 on Build GRCh38
rs375939261 Chr.16: 19040391-19040391 on Build GRCh38
rs774776409 Chr.16: 19040304-19040304 on Build GRCh38
rs767945224 Chr.16: 19040403-19040403 on Build GRCh38
rs201669442 Chr.16: 19040280-19040280 on Build GRCh38
rs924595791 Chr.16: 19040475-19040475 on Build GRCh38
rs751392976 Chr.16: 19040338-19040338 on Build GRCh38
rs373626023 Chr.16: 19040594-19040594 on Build GRCh38
rs759961434 Chr.16: 19040305-19040305 on Build GRCh38
rs755738299 Chr.16: 19040354-19040354 on Build GRCh38
rs576081562 Chr.16: 19040655-19040655 on Build GRCh38
rs554690926 Chr.16: 19040597-19040597 on Build GRCh38
rs896811808 Chr.16: 19040675-19040675 on Build GRCh38
rs972707131 Chr.16: 19040660-19040660 on Build GRCh38
rs374074934 Chr.16: 19040472-19040472 on Build GRCh38
rs570742136 Chr.16: 19040508-19040508 on Build GRCh38
rs748806025 Chr.16: 19040371-19040371 on Build GRCh38
rs565917949 Chr.16: 19040534-19040536 on Build GRCh38
rs779534768 Chr.16: 19040377-19040377 on Build GRCh38
rs746859346 Chr.16: 19040399-19040399 on Build GRCh38
rs529696735 Chr.16: 19040315-19040315 on Build GRCh38
rs767391187 Chr.16: 19040443-19040443 on Build GRCh38
rs777352678 Chr.16: 19040367-19040367 on Build GRCh38
rs752452812 Chr.16: 19040459-19040459 on Build GRCh38
rs912038651 Chr.16: 19040564-19040564 on Build GRCh38
rs975162081 Chr.16: 19040499-19040499 on Build GRCh38
rs796311265 Chr.16: 19040504-19040505 on Build GRCh38
rs182457659 Chr.16: 19040519-19040519 on Build GRCh38
rs372654268 Chr.16: 19040325-19040325 on Build GRCh38
rs369239891 Chr.16: 19040339-19040339 on Build GRCh38
rs953566713 Chr.16: 19040509-19040509 on Build GRCh38
rs542604688 Chr.16: 19040570-19040570 on Build GRCh38

View Full Product Details